Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 10539910-10540409 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NAPG
Entrez Gene ID
8774
Gene Name
NSF attachment protein gamma
Gene Aliases
GAMMASNAP
UniGene
Hs.464622
Gene Chromosome Location
Chr.18: 10525876-10552769 on Build GRCh38
Transcripts
NM_003826.2 XM_011525754.2 XM_011525756.2 XM_017026063.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
986504 c.393G>T p.E131D Chr.18: 10540012-10540012 on Build GRCh38
2883203 c.477A>G p.K159K Chr.18: 10540370-10540370 on Build GRCh38
6721893 c.452G>A p.R151Q Chr.18: 10540345-10540345 on Build GRCh38
4749877 c.485G>A p.R162K Chr.18: 10540378-10540378 on Build GRCh38
1258708 c.422C>A p.A141D Chr.18: 10540041-10540041 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs75947355 Chr.18: 10540161-10540161 on Build GRCh38
rs762743655 Chr.18: 10540250-10540250 on Build GRCh38
rs766771509 Chr.18: 10540338-10540338 on Build GRCh38
rs745682963 Chr.18: 10540357-10540357 on Build GRCh38
rs762630023 Chr.18: 10540011-10540011 on Build GRCh38
rs1038454728 Chr.18: 10540086-10540086 on Build GRCh38
rs370847718 Chr.18: 10540265-10540265 on Build GRCh38
rs556265347 Chr.18: 10540382-10540382 on Build GRCh38
rs1045624279 Chr.18: 10540179-10540179 on Build GRCh38
rs372649678 Chr.18: 10540339-10540339 on Build GRCh38
rs767213842 Chr.18: 10540071-10540073 on Build GRCh38
rs1017355639 Chr.18: 10540031-10540031 on Build GRCh38
rs756574984 Chr.18: 10540326-10540326 on Build GRCh38
rs941430520 Chr.18: 10540081-10540081 on Build GRCh38
rs759601297 Chr.18: 10539999-10539999 on Build GRCh38
rs769763423 Chr.18: 10540378-10540378 on Build GRCh38
rs759418288 Chr.18: 10540070-10540070 on Build GRCh38
rs577651667 Chr.18: 10540304-10540304 on Build GRCh38
rs776702439 Chr.18: 10539967-10539967 on Build GRCh38
rs764779608 Chr.18: 10540007-10540007 on Build GRCh38
rs752455021 Chr.18: 10540080-10540083 on Build GRCh38
rs763703055 Chr.18: 10540313-10540313 on Build GRCh38
rs929966591 Chr.18: 10540362-10540362 on Build GRCh38
rs753063751 Chr.18: 10539941-10539941 on Build GRCh38
rs773010123 Chr.18: 10539960-10539960 on Build GRCh38
rs749347505 Chr.18: 10540078-10540078 on Build GRCh38
rs778971906 Chr.18: 10540287-10540287 on Build GRCh38
rs781092504 Chr.18: 10540354-10540354 on Build GRCh38
rs750916988 Chr.18: 10540039-10540039 on Build GRCh38
rs200300448 Chr.18: 10540345-10540345 on Build GRCh38
rs571432637 Chr.18: 10540203-10540203 on Build GRCh38
rs550215417 Chr.18: 10540175-10540175 on Build GRCh38
rs185443801 Chr.18: 10540172-10540172 on Build GRCh38
rs1009222131 Chr.18: 10539981-10539981 on Build GRCh38
rs373778163 Chr.18: 10540341-10540341 on Build GRCh38
rs17546495 Chr.18: 10540302-10540302 on Build GRCh38
rs762351095 Chr.18: 10540312-10540312 on Build GRCh38
rs768007837 Chr.18: 10539938-10539938 on Build GRCh38
rs545043862 Chr.18: 10540019-10540019 on Build GRCh38
rs17849546 Chr.18: 10540334-10540334 on Build GRCh38
rs892096239 Chr.18: 10539964-10539964 on Build GRCh38
rs770833199 Chr.18: 10540292-10540292 on Build GRCh38
rs1044501886 Chr.18: 10540275-10540275 on Build GRCh38
rs138392728 Chr.18: 10540164-10540164 on Build GRCh38
rs761001321 Chr.18: 10540018-10540018 on Build GRCh38
rs999972029 Chr.18: 10539946-10539946 on Build GRCh38
rs780116556 Chr.18: 10540047-10540047 on Build GRCh38
rs527973210 Chr.18: 10540153-10540153 on Build GRCh38
rs757590309 Chr.18: 10540024-10540024 on Build GRCh38
rs766599630 Chr.18: 10540218-10540218 on Build GRCh38
rs11664051 Chr.18: 10540226-10540226 on Build GRCh38
rs1056697599 Chr.18: 10540171-10540171 on Build GRCh38
rs749471455 Chr.18: 10539951-10539951 on Build GRCh38
rs560217810 Chr.18: 10540066-10540066 on Build GRCh38
rs540177296 Chr.18: 10540316-10540316 on Build GRCh38
rs768767626 Chr.18: 10539958-10539958 on Build GRCh38
rs965850649 Chr.18: 10540046-10540046 on Build GRCh38
rs770888654 Chr.18: 10539966-10539966 on Build GRCh38
rs922514402 Chr.18: 10539994-10539994 on Build GRCh38
rs781604699 Chr.18: 10540034-10540034 on Build GRCh38
rs750792428 Chr.18: 10540325-10540325 on Build GRCh38
rs984808169 Chr.18: 10540197-10540197 on Build GRCh38
rs562577262 Chr.18: 10539961-10539961 on Build GRCh38
rs752149996 Chr.18: 10540009-10540009 on Build GRCh38
rs758695906 Chr.18: 10540348-10540348 on Build GRCh38
rs756628278 Chr.18: 10540043-10540043 on Build GRCh38
rs745730053 Chr.18: 10540303-10540303 on Build GRCh38
rs565778752 Chr.18: 10540262-10540262 on Build GRCh38
rs985874941 Chr.18: 10540025-10540025 on Build GRCh38
rs538778933 Chr.18: 10540219-10540219 on Build GRCh38
rs768828491 Chr.18: 10540089-10540089 on Build GRCh38
rs181135025 Chr.18: 10540017-10540017 on Build GRCh38
rs76471731 Chr.18: 10540319-10540319 on Build GRCh38
rs779103386 Chr.18: 10540344-10540344 on Build GRCh38
rs761512058 Chr.18: 10540314-10540314 on Build GRCh38

View Full Product Details