Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 24065877-24066390 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTC39C
Entrez Gene ID
125488
Gene Name
tetratricopeptide repeat domain 39C
Gene Aliases
C18orf17, HsT2697
UniGene
Hs.128576
Gene Chromosome Location
Chr.18: 23992773-24135952 on Build GRCh38
Transcripts
NM_001135993.1 NM_153211.3 XM_005258198.4 XM_006722392.3 XM_011525814.2 XM_017025550.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6841010 c.162+1G>A p.? Chr.18: 24066141-24066141 on Build GRCh38
1268856 c.141C>G p.I47M Chr.18: 24066119-24066119 on Build GRCh38
6003313 c.112A>G p.S38G Chr.18: 24066090-24066090 on Build GRCh38
473693 c.136A>G p.T46A Chr.18: 24066114-24066114 on Build GRCh38
3371019 c.121G>A p.A41T Chr.18: 24066099-24066099 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs749554835 Chr.18: 24066047-24066047 on Build GRCh38
rs865897706 Chr.18: 24066083-24066083 on Build GRCh38
rs917768695 Chr.18: 24066286-24066286 on Build GRCh38
rs1045424720 Chr.18: 24066358-24066358 on Build GRCh38
rs1031693591 Chr.18: 24065975-24065975 on Build GRCh38
rs774002518 Chr.18: 24066098-24066098 on Build GRCh38
rs781150730 Chr.18: 24066170-24066170 on Build GRCh38
rs769514098 Chr.18: 24066084-24066084 on Build GRCh38
rs769615780 Chr.18: 24066188-24066188 on Build GRCh38
rs1044972521 Chr.18: 24066032-24066032 on Build GRCh38
rs767706688 Chr.18: 24066018-24066018 on Build GRCh38
rs8096969 Chr.18: 24066219-24066219 on Build GRCh38
rs370006797 Chr.18: 24066144-24066144 on Build GRCh38
rs200341803 Chr.18: 24066178-24066178 on Build GRCh38
rs756471825 Chr.18: 24066141-24066141 on Build GRCh38
rs768092769 Chr.18: 24066134-24066134 on Build GRCh38
rs925032923 Chr.18: 24066209-24066209 on Build GRCh38
rs1052665547 Chr.18: 24065906-24065909 on Build GRCh38
rs1013899324 Chr.18: 24065932-24065932 on Build GRCh38
rs756243061 Chr.18: 24066038-24066038 on Build GRCh38
rs775201911 Chr.18: 24066085-24066085 on Build GRCh38
rs762751675 Chr.18: 24066088-24066088 on Build GRCh38
rs779026654 Chr.18: 24066054-24066054 on Build GRCh38
rs369691037 Chr.18: 24065915-24065918 on Build GRCh38
rs780006835 Chr.18: 24066189-24066189 on Build GRCh38
rs8083357 Chr.18: 24066125-24066125 on Build GRCh38
rs549868577 Chr.18: 24066147-24066147 on Build GRCh38
rs752098333 Chr.18: 24066000-24066000 on Build GRCh38
rs767460455 Chr.18: 24066269-24066269 on Build GRCh38
rs755209601 Chr.18: 24066151-24066151 on Build GRCh38
rs906682725 Chr.18: 24066281-24066281 on Build GRCh38
rs761419820 Chr.18: 24065912-24065916 on Build GRCh38
rs896417408 Chr.18: 24065908-24065908 on Build GRCh38
rs866704830 Chr.18: 24066252-24066252 on Build GRCh38
rs199669752 Chr.18: 24066211-24066211 on Build GRCh38
rs752792788 Chr.18: 24066159-24066159 on Build GRCh38
rs370210776 Chr.18: 24065997-24065997 on Build GRCh38
rs929915905 Chr.18: 24066174-24066174 on Build GRCh38
rs988011953 Chr.18: 24066279-24066279 on Build GRCh38
rs149349073 Chr.18: 24066053-24066053 on Build GRCh38
rs149089517 Chr.18: 24065914-24065917 on Build GRCh38
rs750359133 Chr.18: 24065976-24065977 on Build GRCh38
rs151171824 Chr.18: 24066074-24066074 on Build GRCh38
rs1052629886 Chr.18: 24066361-24066361 on Build GRCh38
rs750831590 Chr.18: 24066036-24066036 on Build GRCh38
rs746895142 Chr.18: 24066055-24066055 on Build GRCh38
rs770928753 Chr.18: 24066066-24066066 on Build GRCh38
rs559310528 Chr.18: 24066303-24066303 on Build GRCh38
rs138298971 Chr.18: 24066095-24066095 on Build GRCh38
rs372849552 Chr.18: 24066168-24066168 on Build GRCh38
rs201044353 Chr.18: 24066046-24066046 on Build GRCh38
rs61734884 Chr.18: 24066140-24066140 on Build GRCh38
rs778901933 Chr.18: 24066152-24066152 on Build GRCh38
rs776235873 Chr.18: 24066072-24066072 on Build GRCh38
rs1018520881 Chr.18: 24066166-24066166 on Build GRCh38
rs757852263 Chr.18: 24066003-24066003 on Build GRCh38
rs905891514 Chr.18: 24066037-24066037 on Build GRCh38

View Full Product Details