Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 25062637-25063150 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF521
Entrez Gene ID
25925
Gene Name
zinc finger protein 521
Gene Aliases
EHZF, Evi3
UniGene
Hs.116935
Gene Chromosome Location
Chr.18: 25061924-25352250 on Build GRCh38
Transcripts
NM_001308225.1 NM_015461.2 XM_011525909.2 XM_011525911.1 XM_017025697.1 XM_017025698.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
987202 c.3916A>C p.M1306L Chr.18: 25062732-25062732 on Build GRCh38
6444946 c.3914C>A p.T1305K Chr.18: 25062734-25062734 on Build GRCh38
6444947 c.3907-1G>A p.? Chr.18: 25062742-25062742 on Build GRCh38
3890830 c.3910C>T p.H1304Y Chr.18: 25062738-25062738 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs28510250 Chr.18: 25062751-25062751 on Build GRCh38
rs796347418 Chr.18: 25062784-25062785 on Build GRCh38
rs986286481 Chr.18: 25063000-25063000 on Build GRCh38
rs878886146 Chr.18: 25062801-25062801 on Build GRCh38
rs752539881 Chr.18: 25063111-25063111 on Build GRCh38
rs747129379 Chr.18: 25062693-25062693 on Build GRCh38
rs138058454 Chr.18: 25063030-25063030 on Build GRCh38
rs78938383 Chr.18: 25063086-25063086 on Build GRCh38
rs760223426 Chr.18: 25062755-25062755 on Build GRCh38
rs751977382 Chr.18: 25062750-25062751 on Build GRCh38
rs895698891 Chr.18: 25063098-25063098 on Build GRCh38
rs374855508 Chr.18: 25062662-25062662 on Build GRCh38
rs201903257 Chr.18: 25062745-25062745 on Build GRCh38
rs976864197 Chr.18: 25062920-25062920 on Build GRCh38
rs777211923 Chr.18: 25062788-25062788 on Build GRCh38
rs988953315 Chr.18: 25062910-25062910 on Build GRCh38
rs1044358813 Chr.18: 25063107-25063107 on Build GRCh38
rs200500027 Chr.18: 25062680-25062680 on Build GRCh38
rs377709383 Chr.18: 25062749-25062749 on Build GRCh38
rs1035602851 Chr.18: 25062813-25062813 on Build GRCh38
rs60933974 Chr.18: 25062784-25062785 on Build GRCh38
rs754428499 Chr.18: 25062667-25062667 on Build GRCh38
rs928385163 Chr.18: 25062875-25062875 on Build GRCh38
rs11406899 Chr.18: 25062880-25062881 on Build GRCh38
rs765619728 Chr.18: 25062791-25062791 on Build GRCh38
rs530199615 Chr.18: 25062683-25062683 on Build GRCh38
rs766546540 Chr.18: 25062868-25062868 on Build GRCh38
rs755337049 Chr.18: 25062670-25062670 on Build GRCh38
rs981072875 Chr.18: 25062790-25062790 on Build GRCh38
rs538061871 Chr.18: 25062863-25062863 on Build GRCh38
rs1043108335 Chr.18: 25063048-25063048 on Build GRCh38
rs760027650 Chr.18: 25062789-25062789 on Build GRCh38
rs571943526 Chr.18: 25062901-25062901 on Build GRCh38
rs397696557 Chr.18: 25062881-25062882 on Build GRCh38
rs754522497 Chr.18: 25062753-25062765 on Build GRCh38
rs766010777 Chr.18: 25062762-25062762 on Build GRCh38
rs369573527 Chr.18: 25062690-25062690 on Build GRCh38
rs747720714 Chr.18: 25062784-25062788 on Build GRCh38
rs999289746 Chr.18: 25062689-25062689 on Build GRCh38
rs778531054 Chr.18: 25062753-25062769 on Build GRCh38
rs775552635 Chr.18: 25062749-25062750 on Build GRCh38
rs983856137 Chr.18: 25062902-25062902 on Build GRCh38
rs947539051 Chr.18: 25063015-25063015 on Build GRCh38
rs895758503 Chr.18: 25062856-25062856 on Build GRCh38
rs923508550 Chr.18: 25062927-25062928 on Build GRCh38
rs914779265 Chr.18: 25062916-25062916 on Build GRCh38
rs538787662 Chr.18: 25062860-25062860 on Build GRCh38
rs867322760 Chr.18: 25062786-25062786 on Build GRCh38
rs796379466 Chr.18: 25062860-25062862 on Build GRCh38
rs547857184 Chr.18: 25062781-25062785 on Build GRCh38
rs961066619 Chr.18: 25062913-25062913 on Build GRCh38
rs554128875 Chr.18: 25062994-25062994 on Build GRCh38
rs933576842 Chr.18: 25062954-25062954 on Build GRCh38
rs552091500 Chr.18: 25062862-25062862 on Build GRCh38
rs768141326 Chr.18: 25062753-25062767 on Build GRCh38
rs184884110 Chr.18: 25063074-25063074 on Build GRCh38
rs1015350577 Chr.18: 25063118-25063120 on Build GRCh38
rs757354821 Chr.18: 25062732-25062732 on Build GRCh38
rs768434349 Chr.18: 25062742-25062742 on Build GRCh38
rs765418490 Chr.18: 25062749-25062752 on Build GRCh38
rs749240724 Chr.18: 25062740-25062740 on Build GRCh38
rs746815089 Chr.18: 25062787-25062789 on Build GRCh38
rs534630955 Chr.18: 25062923-25062923 on Build GRCh38
rs116522879 Chr.18: 25062735-25062735 on Build GRCh38
rs142078909 Chr.18: 25063105-25063105 on Build GRCh38
rs533530170 Chr.18: 25063046-25063046 on Build GRCh38
rs190028001 Chr.18: 25063075-25063075 on Build GRCh38
rs757778956 Chr.18: 25062753-25062768 on Build GRCh38
rs924730667 Chr.18: 25063063-25063063 on Build GRCh38
rs760718188 Chr.18: 25062687-25062687 on Build GRCh38
rs10540123 Chr.18: 25062782-25062785 on Build GRCh38
rs758098597 Chr.18: 25062785-25062786 on Build GRCh38
rs1006173987 Chr.18: 25062781-25062781 on Build GRCh38
rs565359617 Chr.18: 25063104-25063104 on Build GRCh38
rs944055178 Chr.18: 25063068-25063068 on Build GRCh38
rs887658953 Chr.18: 25063116-25063116 on Build GRCh38
rs199507114 Chr.18: 25062752-25062752 on Build GRCh38
rs1022811101 Chr.18: 25062865-25062865 on Build GRCh38
rs751037723 Chr.18: 25062753-25062770 on Build GRCh38
rs774083397 Chr.18: 25062744-25062744 on Build GRCh38
rs951959562 Chr.18: 25062778-25062778 on Build GRCh38
rs568471979 Chr.18: 25062753-25062756 on Build GRCh38

View Full Product Details