Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 26038462-26038997 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SS18
Entrez Gene ID
6760
Gene Name
SS18, nBAF chromatin remodeling complex subunit
Gene Aliases
SSXT, SYT
UniGene
Hs.129261
Gene Chromosome Location
Chr.18: 26016253-26091217 on Build GRCh38
Transcripts
NM_001007559.2 NM_001308201.1 NM_005637.3 XM_006722527.2 XM_011526145.1 XM_011526147.2 XM_011526148.2 XM_011526149.2 XM_011526150.2 XM_011526151.2 XM_011526152.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2149707 c.815G>A p.G272E Chr.18: 26038620-26038620 on Build GRCh38
5562074 c.875C>T p.P292L Chr.18: 26038560-26038560 on Build GRCh38
6220777 c.814G>T p.G272W Chr.18: 26038621-26038621 on Build GRCh38
6790652 c.831T>C p.H277H Chr.18: 26038604-26038604 on Build GRCh38
6148982 c.872A>T p.Y291F Chr.18: 26038563-26038563 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1057145196 Chr.18: 26038705-26038705 on Build GRCh38
rs749801259 Chr.18: 26038541-26038541 on Build GRCh38
rs201169007 Chr.18: 26038623-26038623 on Build GRCh38
rs547058660 Chr.18: 26038571-26038571 on Build GRCh38
rs1028690406 Chr.18: 26038932-26038932 on Build GRCh38
rs372046167 Chr.18: 26038615-26038615 on Build GRCh38
rs927934674 Chr.18: 26038945-26038945 on Build GRCh38
rs1020599833 Chr.18: 26038973-26038973 on Build GRCh38
rs376643548 Chr.18: 26038690-26038690 on Build GRCh38
rs781000745 Chr.18: 26038509-26038509 on Build GRCh38
rs72878218 Chr.18: 26038940-26038940 on Build GRCh38
rs112231498 Chr.18: 26038687-26038687 on Build GRCh38
rs755654274 Chr.18: 26038684-26038684 on Build GRCh38
rs181859383 Chr.18: 26038812-26038812 on Build GRCh38
rs750168351 Chr.18: 26038680-26038680 on Build GRCh38
rs779096800 Chr.18: 26038550-26038550 on Build GRCh38
rs201516574 Chr.18: 26038518-26038518 on Build GRCh38
rs761818331 Chr.18: 26038511-26038511 on Build GRCh38
rs755398142 Chr.18: 26038694-26038694 on Build GRCh38
rs756933111 Chr.18: 26038507-26038507 on Build GRCh38
rs937415239 Chr.18: 26038778-26038778 on Build GRCh38
rs596353 Chr.18: 26038920-26038920 on Build GRCh38
rs1010160553 Chr.18: 26038745-26038745 on Build GRCh38
rs944159184 Chr.18: 26038841-26038843 on Build GRCh38
rs868426161 Chr.18: 26038839-26038839 on Build GRCh38
rs1021795560 Chr.18: 26038502-26038502 on Build GRCh38
rs767268320 Chr.18: 26038678-26038678 on Build GRCh38
rs904751040 Chr.18: 26038760-26038760 on Build GRCh38
rs761718161 Chr.18: 26038670-26038670 on Build GRCh38
rs747074442 Chr.18: 26038602-26038602 on Build GRCh38
rs779477357 Chr.18: 26038698-26038698 on Build GRCh38
rs73401882 Chr.18: 26038781-26038781 on Build GRCh38
rs1021594935 Chr.18: 26038759-26038759 on Build GRCh38
rs8095178 Chr.18: 26038807-26038807 on Build GRCh38
rs751396323 Chr.18: 26038506-26038506 on Build GRCh38
rs576149101 Chr.18: 26038862-26038862 on Build GRCh38
rs371755482 Chr.18: 26038520-26038520 on Build GRCh38
rs909765369 Chr.18: 26038626-26038626 on Build GRCh38
rs981511487 Chr.18: 26038749-26038749 on Build GRCh38
rs149281254 Chr.18: 26038568-26038568 on Build GRCh38
rs9953041 Chr.18: 26038834-26038834 on Build GRCh38
rs1018877598 Chr.18: 26038533-26038533 on Build GRCh38
rs746861568 Chr.18: 26038587-26038587 on Build GRCh38
rs376758443 Chr.18: 26038515-26038515 on Build GRCh38
rs965599039 Chr.18: 26038535-26038535 on Build GRCh38
rs951117458 Chr.18: 26038849-26038849 on Build GRCh38
rs1011492415 Chr.18: 26038950-26038950 on Build GRCh38
rs114077471 Chr.18: 26038766-26038766 on Build GRCh38
rs1010234319 Chr.18: 26038498-26038498 on Build GRCh38
rs983757883 Chr.18: 26038859-26038859 on Build GRCh38
rs965714895 Chr.18: 26038595-26038595 on Build GRCh38
rs770979393 Chr.18: 26038605-26038605 on Build GRCh38
rs148462448 Chr.18: 26038656-26038656 on Build GRCh38
rs115330039 Chr.18: 26038622-26038622 on Build GRCh38
rs996324465 Chr.18: 26038903-26038903 on Build GRCh38
rs947768359 Chr.18: 26038905-26038905 on Build GRCh38
rs369084111 Chr.18: 26038704-26038704 on Build GRCh38
rs111379182 Chr.18: 26038872-26038872 on Build GRCh38
rs891886625 Chr.18: 26038732-26038732 on Build GRCh38
rs775464140 Chr.18: 26038621-26038621 on Build GRCh38
rs367950926 Chr.18: 26038647-26038647 on Build GRCh38
rs769045094 Chr.18: 26038545-26038545 on Build GRCh38
rs780607182 Chr.18: 26038527-26038527 on Build GRCh38
rs368021147 Chr.18: 26038596-26038596 on Build GRCh38
rs985619528 Chr.18: 26038516-26038516 on Build GRCh38
rs947978441 Chr.18: 26038682-26038682 on Build GRCh38

View Full Product Details