Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 33226694-33227239 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC178
Entrez Gene ID
374864
Gene Name
coiled-coil domain containing 178
Gene Aliases
C18orf34
UniGene
Hs.115461
Gene Chromosome Location
Chr.18: 32937402-33441040 on Build GRCh38
Transcripts
NM_001105528.1 NM_001308126.1 NM_198995.2 XM_011525948.1 XM_011525951.1 XM_017025721.1 XM_017025722.1 XM_017025723.1 XM_017025724.1 XM_017025725.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5854093 c.1648G>A p.V550M Chr.18: 33226801-33226801 on Build GRCh38
3525365 c.1620C>T p.L540L Chr.18: 33226829-33226829 on Build GRCh38
5431101 c.1594G>T p.G532C Chr.18: 33226855-33226855 on Build GRCh38
6500067 c.1626A>T p.Q542H Chr.18: 33226823-33226823 on Build GRCh38
5987285 c.1600G>A p.E534K Chr.18: 33226849-33226849 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200722018 Chr.18: 33226881-33226881 on Build GRCh38
rs141065788 Chr.18: 33226978-33226978 on Build GRCh38
rs375622157 Chr.18: 33226858-33226858 on Build GRCh38
rs373415111 Chr.18: 33226781-33226781 on Build GRCh38
rs577731150 Chr.18: 33226983-33226983 on Build GRCh38
rs549340060 Chr.18: 33227134-33227134 on Build GRCh38
rs145471109 Chr.18: 33226900-33226900 on Build GRCh38
rs777409252 Chr.18: 33226827-33226827 on Build GRCh38
rs746489224 Chr.18: 33226831-33226831 on Build GRCh38
rs989310109 Chr.18: 33227100-33227100 on Build GRCh38
rs764289278 Chr.18: 33226762-33226762 on Build GRCh38
rs972681128 Chr.18: 33226772-33226772 on Build GRCh38
rs182116500 Chr.18: 33227024-33227024 on Build GRCh38
rs576890207 Chr.18: 33227178-33227178 on Build GRCh38
rs72949112 Chr.18: 33226916-33226916 on Build GRCh38
rs915027126 Chr.18: 33227118-33227118 on Build GRCh38
rs752410492 Chr.18: 33226809-33226809 on Build GRCh38
rs560625452 Chr.18: 33227016-33227016 on Build GRCh38
rs993669042 Chr.18: 33227039-33227039 on Build GRCh38
rs1026923967 Chr.18: 33227085-33227085 on Build GRCh38
rs76533735 Chr.18: 33226932-33226932 on Build GRCh38
rs796831752 Chr.18: 33226932-33226933 on Build GRCh38
rs954755991 Chr.18: 33227092-33227092 on Build GRCh38
rs35564091 Chr.18: 33226941-33226942 on Build GRCh38
rs551885502 Chr.18: 33226855-33226855 on Build GRCh38
rs899446512 Chr.18: 33227027-33227027 on Build GRCh38
rs1017585069 Chr.18: 33227201-33227201 on Build GRCh38
rs202154858 Chr.18: 33226932-33226932 on Build GRCh38
rs953458653 Chr.18: 33227007-33227007 on Build GRCh38
rs753783178 Chr.18: 33226879-33226879 on Build GRCh38
rs763299285 Chr.18: 33226860-33226860 on Build GRCh38
rs200666101 Chr.18: 33226825-33226825 on Build GRCh38
rs1053393995 Chr.18: 33226942-33226942 on Build GRCh38
rs1002427905 Chr.18: 33226724-33226724 on Build GRCh38
rs755465630 Chr.18: 33227019-33227020 on Build GRCh38
rs765655693 Chr.18: 33226750-33226750 on Build GRCh38
rs765138709 Chr.18: 33226895-33226895 on Build GRCh38
rs529855944 Chr.18: 33227017-33227017 on Build GRCh38
rs893519257 Chr.18: 33226856-33226856 on Build GRCh38
rs756083136 Chr.18: 33226780-33226780 on Build GRCh38
rs77111689 Chr.18: 33226933-33226933 on Build GRCh38
rs761906679 Chr.18: 33226872-33226872 on Build GRCh38
rs1016815664 Chr.18: 33226731-33226731 on Build GRCh38
rs949826555 Chr.18: 33226952-33226952 on Build GRCh38
rs147687658 Chr.18: 33226941-33226941 on Build GRCh38
rs563391165 Chr.18: 33227034-33227034 on Build GRCh38
rs529200507 Chr.18: 33227057-33227057 on Build GRCh38
rs925737098 Chr.18: 33226908-33226908 on Build GRCh38
rs895555233 Chr.18: 33226946-33226946 on Build GRCh38
rs763026405 Chr.18: 33226755-33226755 on Build GRCh38
rs1044014440 Chr.18: 33226986-33226986 on Build GRCh38
rs768725119 Chr.18: 33226861-33226861 on Build GRCh38
rs754801622 Chr.18: 33226793-33226793 on Build GRCh38
rs943957480 Chr.18: 33226747-33226747 on Build GRCh38
rs535034484 Chr.18: 33226926-33226926 on Build GRCh38
rs935151621 Chr.18: 33227044-33227044 on Build GRCh38
rs954806929 Chr.18: 33226823-33226823 on Build GRCh38
rs760809489 Chr.18: 33226999-33226999 on Build GRCh38
rs140434666 Chr.18: 33227020-33227024 on Build GRCh38
rs963582477 Chr.18: 33226740-33226740 on Build GRCh38
rs774461330 Chr.18: 33226871-33226871 on Build GRCh38
rs751583306 Chr.18: 33226769-33226769 on Build GRCh38
rs746309219 Chr.18: 33226836-33226836 on Build GRCh38
rs775784940 Chr.18: 33226753-33226753 on Build GRCh38
rs1003779574 Chr.18: 33226931-33226931 on Build GRCh38
rs201359481 Chr.18: 33226876-33226876 on Build GRCh38
rs535112513 Chr.18: 33226805-33226805 on Build GRCh38
rs771616282 Chr.18: 33226832-33226832 on Build GRCh38
rs752432011 Chr.18: 33226896-33226896 on Build GRCh38
rs369257368 Chr.18: 33227024-33227028 on Build GRCh38
rs923817457 Chr.18: 33227041-33227041 on Build GRCh38
rs778510066 Chr.18: 33226808-33226808 on Build GRCh38
rs757975403 Chr.18: 33226821-33226821 on Build GRCh38
rs201782989 Chr.18: 33226833-33226833 on Build GRCh38
rs376435257 Chr.18: 33226784-33226784 on Build GRCh38

View Full Product Details