Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 35991527-35992025 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPRD1A
Entrez Gene ID
55197
Gene Name
regulation of nuclear pre-mRNA domain containing 1A
Gene Aliases
HsT3101, P15RS
UniGene
Hs.464912
Gene Chromosome Location
Chr.18: 35982385-36067594 on Build GRCh38
Transcripts
NM_001303411.1 NM_001303412.1 NM_018170.4 XM_011526048.1 XM_011526049.1 XM_011526050.1

SNP Detail

SNP ID Chromosome Location
rs569342644 Chr.18: 35991757-35991757 on Build GRCh38
rs1053599881 Chr.18: 35991574-35991574 on Build GRCh38
rs554968431 Chr.18: 35991759-35991759 on Build GRCh38
rs762608353 Chr.18: 35991556-35991556 on Build GRCh38
rs954335769 Chr.18: 35991764-35991764 on Build GRCh38
rs923870427 Chr.18: 35991806-35991806 on Build GRCh38
rs991210191 Chr.18: 35991993-35991993 on Build GRCh38
rs973989825 Chr.18: 35991614-35991614 on Build GRCh38
rs1039524569 Chr.18: 35991801-35991801 on Build GRCh38
rs895020754 Chr.18: 35991827-35991827 on Build GRCh38
rs894961188 Chr.18: 35991591-35991591 on Build GRCh38
rs572694916 Chr.18: 35991875-35991875 on Build GRCh38
rs947873700 Chr.18: 35991837-35991837 on Build GRCh38
rs141717307 Chr.18: 35991868-35991868 on Build GRCh38
rs982444264 Chr.18: 35991653-35991653 on Build GRCh38
rs537802108 Chr.18: 35991758-35991758 on Build GRCh38
rs762323703 Chr.18: 35991876-35991879 on Build GRCh38
rs539286193 Chr.18: 35991598-35991598 on Build GRCh38
rs1018023655 Chr.18: 35991580-35991580 on Build GRCh38
rs902396239 Chr.18: 35991551-35991551 on Build GRCh38
rs567048556 Chr.18: 35991561-35991561 on Build GRCh38
rs1054096138 Chr.18: 35991822-35991822 on Build GRCh38
rs752798093 Chr.18: 35991754-35991754 on Build GRCh38
rs530195907 Chr.18: 35991814-35991814 on Build GRCh38
rs991660638 Chr.18: 35991712-35991712 on Build GRCh38
rs544815017 Chr.18: 35991958-35991958 on Build GRCh38
rs746184084 Chr.18: 35991697-35991698 on Build GRCh38
rs961584645 Chr.18: 35991699-35991699 on Build GRCh38
rs1015802472 Chr.18: 35991694-35991694 on Build GRCh38
rs985266174 Chr.18: 35991765-35991765 on Build GRCh38
rs114328900 Chr.18: 35991626-35991626 on Build GRCh38
rs951364322 Chr.18: 35991846-35991846 on Build GRCh38
rs1015519842 Chr.18: 35991934-35991934 on Build GRCh38
rs897149603 Chr.18: 35991967-35991967 on Build GRCh38
rs766213672 Chr.18: 35991741-35991741 on Build GRCh38
rs912305658 Chr.18: 35991770-35991770 on Build GRCh38
rs964325425 Chr.18: 35991606-35991606 on Build GRCh38
rs1002796353 Chr.18: 35991679-35991679 on Build GRCh38
rs551383518 Chr.18: 35991798-35991798 on Build GRCh38
rs959796455 Chr.18: 35991876-35991876 on Build GRCh38
rs1013027384 Chr.18: 35991594-35991594 on Build GRCh38
rs1044469147 Chr.18: 35991865-35991865 on Build GRCh38
rs1023469388 Chr.18: 35991608-35991608 on Build GRCh38
rs943691154 Chr.18: 35991778-35991778 on Build GRCh38
rs1027305162 Chr.18: 35991616-35991616 on Build GRCh38
rs1008024530 Chr.18: 35991562-35991562 on Build GRCh38
rs928406187 Chr.18: 35991657-35991657 on Build GRCh38
rs995726950 Chr.18: 35991817-35991817 on Build GRCh38

View Full Product Details