Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 42743222-42743726 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RIT2
Entrez Gene ID
6014
Gene Name
Ras like without CAAX 2
Gene Aliases
RIBA, RIN, ROC2
UniGene
Hs.464985
Gene Chromosome Location
Chr.18: 42743218-43115692 on Build GRCh38
Transcripts
NM_001272077.1 NM_002930.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7246606 c.623C>G p.S208C Chr.18: 42743524-42743524 on Build GRCh38
5639673 c.583C>A p.L195M Chr.18: 42743564-42743564 on Build GRCh38
6556980 c.558C>A p.S186S Chr.18: 42743589-42743589 on Build GRCh38
1680343 c.545G>A p.R182H Chr.18: 42743602-42743602 on Build GRCh38
374271 c.496C>A p.L166I Chr.18: 42743651-42743651 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776068581 Chr.18: 42743565-42743565 on Build GRCh38
rs756266761 Chr.18: 42743496-42743496 on Build GRCh38
rs983274355 Chr.18: 42743610-42743610 on Build GRCh38
rs920649676 Chr.18: 42743601-42743601 on Build GRCh38
rs1001250399 Chr.18: 42743433-42743433 on Build GRCh38
rs756527558 Chr.18: 42743332-42743332 on Build GRCh38
rs559678016 Chr.18: 42743546-42743546 on Build GRCh38
rs896786438 Chr.18: 42743404-42743404 on Build GRCh38
rs753906491 Chr.18: 42743609-42743609 on Build GRCh38
rs374009419 Chr.18: 42743491-42743491 on Build GRCh38
rs777127147 Chr.18: 42743687-42743687 on Build GRCh38
rs190327842 Chr.18: 42743528-42743528 on Build GRCh38
rs1048070075 Chr.18: 42743415-42743415 on Build GRCh38
rs1012397945 Chr.18: 42743400-42743400 on Build GRCh38
rs886321674 Chr.18: 42743334-42743334 on Build GRCh38
rs771385813 Chr.18: 42743684-42743684 on Build GRCh38
rs17851608 Chr.18: 42743549-42743549 on Build GRCh38
rs749404972 Chr.18: 42743504-42743504 on Build GRCh38
rs756254750 Chr.18: 42743478-42743478 on Build GRCh38
rs748387427 Chr.18: 42743645-42743645 on Build GRCh38
rs1804730 Chr.18: 42743532-42743532 on Build GRCh38
rs767551861 Chr.18: 42743506-42743509 on Build GRCh38
rs754174204 Chr.18: 42743666-42743667 on Build GRCh38
rs944282718 Chr.18: 42743357-42743357 on Build GRCh38
rs746661774 Chr.18: 42743510-42743510 on Build GRCh38
rs904709319 Chr.18: 42743429-42743429 on Build GRCh38
rs753459794 Chr.18: 42743460-42743460 on Build GRCh38
rs759077601 Chr.18: 42743568-42743568 on Build GRCh38
rs775199995 Chr.18: 42743700-42743700 on Build GRCh38
rs1003242280 Chr.18: 42743350-42743350 on Build GRCh38
rs750549822 Chr.18: 42743510-42743512 on Build GRCh38
rs762549730 Chr.18: 42743703-42743703 on Build GRCh38
rs756280371 Chr.18: 42743550-42743552 on Build GRCh38
rs199574261 Chr.18: 42743674-42743674 on Build GRCh38
rs1019350553 Chr.18: 42743511-42743511 on Build GRCh38
rs186313364 Chr.18: 42743468-42743468 on Build GRCh38
rs200268352 Chr.18: 42743540-42743540 on Build GRCh38
rs183755532 Chr.18: 42743308-42743308 on Build GRCh38
rs912736533 Chr.18: 42743344-42743344 on Build GRCh38
rs912014565 Chr.18: 42743254-42743254 on Build GRCh38
rs972962024 Chr.18: 42743637-42743637 on Build GRCh38
rs1049909605 Chr.18: 42743320-42743320 on Build GRCh38
rs144676190 Chr.18: 42743587-42743587 on Build GRCh38
rs770404405 Chr.18: 42743559-42743559 on Build GRCh38
rs550440630 Chr.18: 42743580-42743580 on Build GRCh38
rs769241989 Chr.18: 42743691-42743691 on Build GRCh38
rs764891178 Chr.18: 42743464-42743464 on Build GRCh38
rs1056137158 Chr.18: 42743359-42743359 on Build GRCh38
rs1047679055 Chr.18: 42743327-42743327 on Build GRCh38
rs375571309 Chr.18: 42743461-42743461 on Build GRCh38
rs145853095 Chr.18: 42743261-42743261 on Build GRCh38
rs544036881 Chr.18: 42743293-42743293 on Build GRCh38
rs77976328 Chr.18: 42743557-42743557 on Build GRCh38
rs944213504 Chr.18: 42743612-42743612 on Build GRCh38
rs765941930 Chr.18: 42743450-42743450 on Build GRCh38
rs775014461 Chr.18: 42743586-42743586 on Build GRCh38
rs750986801 Chr.18: 42743303-42743303 on Build GRCh38
rs1000281921 Chr.18: 42743448-42743448 on Build GRCh38
rs367551216 Chr.18: 42743635-42743635 on Build GRCh38
rs551452606 Chr.18: 42743679-42743679 on Build GRCh38
rs75711569 Chr.18: 42743418-42743418 on Build GRCh38
rs780496841 Chr.18: 42743360-42743360 on Build GRCh38
rs755126715 Chr.18: 42743622-42743622 on Build GRCh38
rs543688490 Chr.18: 42743386-42743386 on Build GRCh38
rs763858354 Chr.18: 42743592-42743592 on Build GRCh38
rs140706543 Chr.18: 42743469-42743469 on Build GRCh38
rs771016378 Chr.18: 42743613-42743613 on Build GRCh38
rs148544378 Chr.18: 42743602-42743602 on Build GRCh38
rs757214621 Chr.18: 42743603-42743603 on Build GRCh38
rs995574615 Chr.18: 42743405-42743408 on Build GRCh38
rs939233556 Chr.18: 42743638-42743638 on Build GRCh38
rs573478561 Chr.18: 42743292-42743292 on Build GRCh38
rs531935933 Chr.18: 42743538-42743538 on Build GRCh38
rs142876352 Chr.18: 42743688-42743688 on Build GRCh38
rs772379372 Chr.18: 42743544-42743544 on Build GRCh38
rs758760009 Chr.18: 42743648-42743648 on Build GRCh38
rs766593005 Chr.18: 42743595-42743597 on Build GRCh38
rs1056164595 Chr.18: 42743676-42743676 on Build GRCh38
rs376192514 Chr.18: 42743428-42743428 on Build GRCh38

View Full Product Details