Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 46122885-46123429 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAUS1
Entrez Gene ID
115106
Gene Name
HAUS augmin like complex subunit 1
Gene Aliases
CCDC5, HEI-C, HEIC, HsT1461
UniGene
Hs.436617
Gene Chromosome Location
Chr.18: 46104332-46128333 on Build GRCh38
Transcripts
NM_138443.3 NR_026978.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
709480 c.618A>T p.R206S Chr.18: 46123316-46123316 on Build GRCh38
4932282 c.622A>G p.M208V Chr.18: 46123320-46123320 on Build GRCh38
4072307 c.647C>A p.S216Y Chr.18: 46123345-46123345 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs776531269 Chr.18: 46123279-46123279 on Build GRCh38
rs745737659 Chr.18: 46123368-46123368 on Build GRCh38
rs755518669 Chr.18: 46122950-46122950 on Build GRCh38
rs368127982 Chr.18: 46123295-46123295 on Build GRCh38
rs945123410 Chr.18: 46122934-46122934 on Build GRCh38
rs187643717 Chr.18: 46123020-46123020 on Build GRCh38
rs1013932584 Chr.18: 46123005-46123005 on Build GRCh38
rs966608338 Chr.18: 46123167-46123167 on Build GRCh38
rs367567427 Chr.18: 46123084-46123084 on Build GRCh38
rs758032050 Chr.18: 46123354-46123354 on Build GRCh38
rs201384968 Chr.18: 46123317-46123317 on Build GRCh38
rs965336664 Chr.18: 46123280-46123280 on Build GRCh38
rs867366571 Chr.18: 46123205-46123205 on Build GRCh38
rs528173956 Chr.18: 46123008-46123008 on Build GRCh38
rs930107209 Chr.18: 46123154-46123154 on Build GRCh38
rs1003542072 Chr.18: 46122994-46122994 on Build GRCh38
rs966012771 Chr.18: 46123356-46123356 on Build GRCh38
rs545741771 Chr.18: 46123358-46123358 on Build GRCh38
rs766760587 Chr.18: 46123326-46123326 on Build GRCh38
rs1029397514 Chr.18: 46123204-46123204 on Build GRCh38
rs574465394 Chr.18: 46123088-46123088 on Build GRCh38
rs1055101274 Chr.18: 46123119-46123119 on Build GRCh38
rs943564484 Chr.18: 46122954-46122954 on Build GRCh38
rs889349370 Chr.18: 46122951-46122951 on Build GRCh38
rs767785590 Chr.18: 46123264-46123264 on Build GRCh38
rs190809322 Chr.18: 46123062-46123062 on Build GRCh38
rs747254552 Chr.18: 46123039-46123039 on Build GRCh38
rs570316705 Chr.18: 46123076-46123076 on Build GRCh38
rs778012726 Chr.18: 46123028-46123028 on Build GRCh38
rs1001823325 Chr.18: 46122966-46122966 on Build GRCh38
rs896086380 Chr.18: 46123194-46123194 on Build GRCh38
rs978164184 Chr.18: 46122941-46122941 on Build GRCh38
rs951546492 Chr.18: 46123393-46123393 on Build GRCh38
rs374877700 Chr.18: 46123373-46123373 on Build GRCh38
rs775533628 Chr.18: 46123386-46123386 on Build GRCh38
rs920841898 Chr.18: 46123328-46123328 on Build GRCh38
rs12954742 Chr.18: 46123257-46123257 on Build GRCh38
rs748573541 Chr.18: 46123349-46123349 on Build GRCh38
rs746638363 Chr.18: 46123264-46123264 on Build GRCh38
rs1038276697 Chr.18: 46123079-46123079 on Build GRCh38
rs1041410490 Chr.18: 46123210-46123211 on Build GRCh38
rs530715221 Chr.18: 46122993-46122993 on Build GRCh38
rs895059315 Chr.18: 46123157-46123157 on Build GRCh38
rs567949483 Chr.18: 46123040-46123040 on Build GRCh38
rs933652315 Chr.18: 46122949-46122949 on Build GRCh38
rs767551599 Chr.18: 46123314-46123314 on Build GRCh38
rs915001949 Chr.18: 46122913-46122913 on Build GRCh38
rs1047766339 Chr.18: 46122987-46122987 on Build GRCh38
rs1041410186 Chr.18: 46122932-46122932 on Build GRCh38
rs540185464 Chr.18: 46123110-46123111 on Build GRCh38
rs754358648 Chr.18: 46122940-46122940 on Build GRCh38
rs369042263 Chr.18: 46122974-46122974 on Build GRCh38
rs762850547 Chr.18: 46123400-46123400 on Build GRCh38
rs910818603 Chr.18: 46123071-46123071 on Build GRCh38
rs746691730 Chr.18: 46123364-46123364 on Build GRCh38
rs535407004 Chr.18: 46123054-46123054 on Build GRCh38
rs955067040 Chr.18: 46123067-46123067 on Build GRCh38
rs142850678 Chr.18: 46123236-46123236 on Build GRCh38
rs553396640 Chr.18: 46123142-46123142 on Build GRCh38
rs111641910 Chr.18: 46123001-46123001 on Build GRCh38
rs745869821 Chr.18: 46123166-46123166 on Build GRCh38
rs570669385 Chr.18: 46123007-46123007 on Build GRCh38
rs779530710 Chr.18: 46123335-46123335 on Build GRCh38
rs920738288 Chr.18: 46123083-46123083 on Build GRCh38
rs1010790994 Chr.18: 46123159-46123159 on Build GRCh38
rs985079966 Chr.18: 46123068-46123068 on Build GRCh38
rs78026441 Chr.18: 46122986-46122986 on Build GRCh38
rs375684095 Chr.18: 46123325-46123325 on Build GRCh38
rs1055941791 Chr.18: 46123168-46123168 on Build GRCh38
rs534791581 Chr.18: 46123136-46123136 on Build GRCh38
rs930028486 Chr.18: 46122946-46122946 on Build GRCh38
rs199705963 Chr.18: 46123286-46123286 on Build GRCh38
rs202180722 Chr.18: 46123365-46123365 on Build GRCh38
rs376842842 Chr.18: 46122914-46122914 on Build GRCh38
rs1007360607 Chr.18: 46123293-46123293 on Build GRCh38
rs755453289 Chr.18: 46123332-46123332 on Build GRCh38
rs1037843354 Chr.18: 46122955-46122955 on Build GRCh38
rs368410779 Chr.18: 46123211-46123211 on Build GRCh38
rs752549162 Chr.18: 46123077-46123077 on Build GRCh38
rs754162187 Chr.18: 46123329-46123329 on Build GRCh38
rs922237509 Chr.18: 46122943-46122943 on Build GRCh38
rs1003573193 Chr.18: 46123109-46123109 on Build GRCh38
rs758744585 Chr.18: 46123003-46123003 on Build GRCh38
rs955098120 Chr.18: 46123229-46123229 on Build GRCh38
rs966327785 Chr.18: 46123103-46123103 on Build GRCh38
rs796600608 Chr.18: 46123197-46123197 on Build GRCh38
rs546704664 Chr.18: 46123038-46123038 on Build GRCh38
rs568551031 Chr.18: 46123104-46123104 on Build GRCh38

View Full Product Details