Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 59903772-59904319 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PMAIP1
Entrez Gene ID
5366
Gene Name
phorbol-12-myristate-13-acetate-induced protein 1
Gene Aliases
APR, NOXA
UniGene
Hs.96
Gene Chromosome Location
Chr.18: 59899960-59904306 on Build GRCh38
Transcripts
NM_021127.2

SNP Detail

SNP ID Chromosome Location
rs765366709 Chr.18: 59903908-59903908 on Build GRCh38
rs965194626 Chr.18: 59904204-59904204 on Build GRCh38
rs921210068 Chr.18: 59903833-59903833 on Build GRCh38
rs575723889 Chr.18: 59903912-59903912 on Build GRCh38
rs2849431 Chr.18: 59904236-59904236 on Build GRCh38
rs138203769 Chr.18: 59904180-59904180 on Build GRCh38
rs112680252 Chr.18: 59903931-59903934 on Build GRCh38
rs560808893 Chr.18: 59904179-59904179 on Build GRCh38
rs931293805 Chr.18: 59903890-59903890 on Build GRCh38
rs758854744 Chr.18: 59904108-59904108 on Build GRCh38
rs894097632 Chr.18: 59904092-59904092 on Build GRCh38
rs958815749 Chr.18: 59904090-59904090 on Build GRCh38
rs544391956 Chr.18: 59903945-59903945 on Build GRCh38
rs1033745590 Chr.18: 59904079-59904079 on Build GRCh38
rs558758218 Chr.18: 59903859-59903859 on Build GRCh38
rs574941137 Chr.18: 59904103-59904103 on Build GRCh38
rs1040916120 Chr.18: 59903857-59903857 on Build GRCh38
rs892738351 Chr.18: 59903901-59903901 on Build GRCh38
rs961996968 Chr.18: 59904126-59904126 on Build GRCh38
rs190664610 Chr.18: 59904235-59904235 on Build GRCh38
rs7245349 Chr.18: 59904135-59904135 on Build GRCh38
rs7227934 Chr.18: 59904051-59904051 on Build GRCh38
rs975277630 Chr.18: 59904225-59904225 on Build GRCh38
rs74255491 Chr.18: 59903798-59903798 on Build GRCh38
rs903845497 Chr.18: 59904009-59904009 on Build GRCh38
rs1009056612 Chr.18: 59903837-59903837 on Build GRCh38
rs985873459 Chr.18: 59904237-59904237 on Build GRCh38
rs1021673738 Chr.18: 59904120-59904120 on Build GRCh38
rs1022310831 Chr.18: 59903916-59903916 on Build GRCh38
rs11376390 Chr.18: 59904179-59904180 on Build GRCh38
rs1024534136 Chr.18: 59904121-59904121 on Build GRCh38
rs34120135 Chr.18: 59904132-59904133 on Build GRCh38
rs551426467 Chr.18: 59904200-59904200 on Build GRCh38
rs778258920 Chr.18: 59904180-59904180 on Build GRCh38
rs573004852 Chr.18: 59903900-59903901 on Build GRCh38

View Full Product Details