Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 63660955-63661500 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SERPINB3
Entrez Gene ID
6317
Gene Name
serpin family B member 3
Gene Aliases
HsT1196, SCC, SCCA-1, SCCA-PD, SCCA1, SSCA1, T4-A
UniGene
Hs.227948
Gene Chromosome Location
Chr.18: 63655197-63661963 on Build GRCh38
Transcripts
NM_006919.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4522765 c.117G>A p.G39G Chr.18: 63661100-63661100 on Build GRCh38
709132 c.27C>T p.T9T Chr.18: 63661190-63661190 on Build GRCh38
3527065 c.109G>A p.A37T Chr.18: 63661108-63661108 on Build GRCh38
6149198 c.10C>G p.L4V Chr.18: 63661207-63661207 on Build GRCh38
338580 c.79A>T p.I27F Chr.18: 63661138-63661138 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148776258 Chr.18: 63661177-63661177 on Build GRCh38
rs149471122 Chr.18: 63661322-63661322 on Build GRCh38
rs965936349 Chr.18: 63661314-63661314 on Build GRCh38
rs764830431 Chr.18: 63661226-63661226 on Build GRCh38
rs200505823 Chr.18: 63661107-63661107 on Build GRCh38
rs751332591 Chr.18: 63661055-63661055 on Build GRCh38
rs749347006 Chr.18: 63661182-63661182 on Build GRCh38
rs370684728 Chr.18: 63661002-63661002 on Build GRCh38
rs771519957 Chr.18: 63661037-63661037 on Build GRCh38
rs367779002 Chr.18: 63661035-63661035 on Build GRCh38
rs777222567 Chr.18: 63661123-63661123 on Build GRCh38
rs746360608 Chr.18: 63661125-63661125 on Build GRCh38
rs759031235 Chr.18: 63661222-63661222 on Build GRCh38
rs188823598 Chr.18: 63661421-63661421 on Build GRCh38
rs1041035002 Chr.18: 63660984-63660984 on Build GRCh38
rs768620699 Chr.18: 63661183-63661183 on Build GRCh38
rs772234230 Chr.18: 63661259-63661259 on Build GRCh38
rs938533763 Chr.18: 63661348-63661348 on Build GRCh38
rs367856187 Chr.18: 63661001-63661001 on Build GRCh38
rs771428386 Chr.18: 63661118-63661118 on Build GRCh38
rs368725939 Chr.18: 63661100-63661100 on Build GRCh38
rs757066825 Chr.18: 63661062-63661062 on Build GRCh38
rs114356590 Chr.18: 63661361-63661361 on Build GRCh38
rs894120894 Chr.18: 63661227-63661227 on Build GRCh38
rs145324165 Chr.18: 63661102-63661102 on Build GRCh38
rs546374165 Chr.18: 63661204-63661204 on Build GRCh38
rs754481412 Chr.18: 63661165-63661165 on Build GRCh38
rs767312438 Chr.18: 63661138-63661138 on Build GRCh38
rs767400496 Chr.18: 63661063-63661063 on Build GRCh38
rs192414964 Chr.18: 63661276-63661276 on Build GRCh38
rs780428071 Chr.18: 63660990-63660990 on Build GRCh38
rs377113169 Chr.18: 63661473-63661478 on Build GRCh38
rs998601539 Chr.18: 63661297-63661297 on Build GRCh38
rs756465955 Chr.18: 63661244-63661244 on Build GRCh38
rs764328137 Chr.18: 63661129-63661129 on Build GRCh38
rs537410928 Chr.18: 63661300-63661300 on Build GRCh38
rs144541327 Chr.18: 63661083-63661083 on Build GRCh38
rs977548708 Chr.18: 63661105-63661105 on Build GRCh38
rs753391938 Chr.18: 63661476-63661476 on Build GRCh38
rs377476734 Chr.18: 63661251-63661251 on Build GRCh38
rs919055145 Chr.18: 63661346-63661346 on Build GRCh38
rs758354721 Chr.18: 63661132-63661134 on Build GRCh38
rs762989240 Chr.18: 63661128-63661128 on Build GRCh38
rs375182717 Chr.18: 63661174-63661174 on Build GRCh38
rs564722374 Chr.18: 63661042-63661042 on Build GRCh38
rs774496420 Chr.18: 63661193-63661193 on Build GRCh38
rs998527929 Chr.18: 63661406-63661406 on Build GRCh38
rs530217444 Chr.18: 63661372-63661372 on Build GRCh38
rs560685327 Chr.18: 63661424-63661424 on Build GRCh38
rs369196968 Chr.18: 63661214-63661214 on Build GRCh38
rs984102670 Chr.18: 63661330-63661330 on Build GRCh38
rs762019199 Chr.18: 63661194-63661194 on Build GRCh38
rs773454926 Chr.18: 63661261-63661261 on Build GRCh38
rs749772472 Chr.18: 63661252-63661252 on Build GRCh38
rs781685229 Chr.18: 63661175-63661175 on Build GRCh38
rs779504509 Chr.18: 63661003-63661003 on Build GRCh38
rs530476455 Chr.18: 63661044-63661044 on Build GRCh38
rs560362906 Chr.18: 63661012-63661012 on Build GRCh38
rs1037124071 Chr.18: 63661341-63661341 on Build GRCh38
rs760454101 Chr.18: 63661264-63661264 on Build GRCh38
rs3180048 Chr.18: 63661179-63661179 on Build GRCh38
rs977007613 Chr.18: 63661317-63661317 on Build GRCh38
rs775604979 Chr.18: 63661127-63661127 on Build GRCh38
rs748276719 Chr.18: 63661258-63661258 on Build GRCh38
rs897686194 Chr.18: 63661210-63661210 on Build GRCh38
rs758089735 Chr.18: 63661231-63661231 on Build GRCh38
rs370532222 Chr.18: 63661254-63661254 on Build GRCh38
rs780447173 Chr.18: 63661178-63661178 on Build GRCh38
rs778646037 Chr.18: 63661170-63661170 on Build GRCh38
rs200936767 Chr.18: 63661048-63661048 on Build GRCh38
rs141517348 Chr.18: 63660998-63660998 on Build GRCh38
rs774587965 Chr.18: 63661131-63661131 on Build GRCh38
rs761528409 Chr.18: 63661136-63661136 on Build GRCh38
rs144341650 Chr.18: 63661380-63661380 on Build GRCh38
rs139092423 Chr.18: 63661217-63661217 on Build GRCh38
rs374199626 Chr.18: 63661229-63661229 on Build GRCh38
rs1006070461 Chr.18: 63661023-63661023 on Build GRCh38
rs1012811659 Chr.18: 63661379-63661379 on Build GRCh38
rs778833698 Chr.18: 63661011-63661012 on Build GRCh38
rs182802060 Chr.18: 63661032-63661032 on Build GRCh38
rs760631782 Chr.18: 63661151-63661151 on Build GRCh38
rs779614140 Chr.18: 63661082-63661082 on Build GRCh38
rs553279329 Chr.18: 63661334-63661334 on Build GRCh38
rs183882015 Chr.18: 63661332-63661332 on Build GRCh38
rs372246924 Chr.18: 63661038-63661038 on Build GRCh38
rs753340443 Chr.18: 63661159-63661159 on Build GRCh38
rs756065094 Chr.18: 63661075-63661075 on Build GRCh38
rs998746632 Chr.18: 63661440-63661440 on Build GRCh38
rs772322065 Chr.18: 63661203-63661203 on Build GRCh38
rs904231865 Chr.18: 63661396-63661396 on Build GRCh38
rs868056621 Chr.18: 63661086-63661086 on Build GRCh38
rs573526343 Chr.18: 63661338-63661338 on Build GRCh38
rs777612933 Chr.18: 63661237-63661237 on Build GRCh38
rs146226385 Chr.18: 63661026-63661026 on Build GRCh38
rs142095760 Chr.18: 63661215-63661215 on Build GRCh38
rs769892482 Chr.18: 63661397-63661397 on Build GRCh38
rs1031841376 Chr.18: 63661419-63661419 on Build GRCh38
rs372277533 Chr.18: 63661143-63661143 on Build GRCh38
rs557509967 Chr.18: 63661311-63661311 on Build GRCh38
rs148760425 Chr.18: 63661420-63661420 on Build GRCh38
rs35298803 Chr.18: 63661366-63661367 on Build GRCh38
rs750364027 Chr.18: 63661064-63661064 on Build GRCh38
rs752413507 Chr.18: 63661172-63661172 on Build GRCh38
rs187745639 Chr.18: 63661051-63661051 on Build GRCh38

View Full Product Details