Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 63897648-63898162 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SERPINB2
Entrez Gene ID
5055
Gene Name
serpin family B member 2
Gene Aliases
HsT1201, PAI, PAI-2, PAI2, PLANH2
UniGene
Hs.594481
Gene Chromosome Location
Chr.18: 63887705-63903890 on Build GRCh38
Transcripts
NM_001143818.1 NM_002575.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3422280 c.507T>G p.I169M Chr.18: 63897816-63897816 on Build GRCh38
1750569 c.427C>T p.R143* Chr.18: 63897736-63897736 on Build GRCh38
989527 c.428G>A p.R143Q Chr.18: 63897737-63897737 on Build GRCh38
284902 c.512C>T p.S171F Chr.18: 63897821-63897821 on Build GRCh38
1680402 c.478G>T p.E160* Chr.18: 63897787-63897787 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1035709564 Chr.18: 63898011-63898011 on Build GRCh38
rs752724853 Chr.18: 63897857-63897857 on Build GRCh38
rs17850949 Chr.18: 63897817-63897817 on Build GRCh38
rs769183693 Chr.18: 63897816-63897816 on Build GRCh38
rs778617491 Chr.18: 63897698-63897698 on Build GRCh38
rs533337669 Chr.18: 63897952-63897952 on Build GRCh38
rs950755945 Chr.18: 63898013-63898013 on Build GRCh38
rs141710617 Chr.18: 63898028-63898028 on Build GRCh38
rs766631504 Chr.18: 63897711-63897711 on Build GRCh38
rs1032162897 Chr.18: 63897729-63897729 on Build GRCh38
rs200662224 Chr.18: 63897758-63897758 on Build GRCh38
rs758858514 Chr.18: 63898088-63898088 on Build GRCh38
rs6101 Chr.18: 63897813-63897813 on Build GRCh38
rs139439211 Chr.18: 63897765-63897765 on Build GRCh38
rs746586973 Chr.18: 63897877-63897877 on Build GRCh38
rs185872665 Chr.18: 63897801-63897801 on Build GRCh38
rs775941879 Chr.18: 63897736-63897736 on Build GRCh38
rs368768832 Chr.18: 63897881-63897881 on Build GRCh38
rs374709649 Chr.18: 63897800-63897800 on Build GRCh38
rs770044796 Chr.18: 63897799-63897799 on Build GRCh38
rs536161962 Chr.18: 63898006-63898006 on Build GRCh38
rs763497538 Chr.18: 63897737-63897737 on Build GRCh38
rs765756137 Chr.18: 63897973-63897973 on Build GRCh38
rs1916659 Chr.18: 63897974-63897974 on Build GRCh38
rs372579942 Chr.18: 63897865-63897865 on Build GRCh38
rs750604420 Chr.18: 63897832-63897832 on Build GRCh38
rs751668373 Chr.18: 63897751-63897751 on Build GRCh38
rs6105 Chr.18: 63897828-63897828 on Build GRCh38
rs950819396 Chr.18: 63897847-63897847 on Build GRCh38
rs770256617 Chr.18: 63897733-63897733 on Build GRCh38
rs771657947 Chr.18: 63897715-63897715 on Build GRCh38
rs150561049 Chr.18: 63898109-63898109 on Build GRCh38
rs893375819 Chr.18: 63897833-63897833 on Build GRCh38
rs768251879 Chr.18: 63897697-63897697 on Build GRCh38
rs747757888 Chr.18: 63897709-63897709 on Build GRCh38
rs1023468854 Chr.18: 63897770-63897770 on Build GRCh38
rs988098998 Chr.18: 63898112-63898112 on Build GRCh38
rs903205843 Chr.18: 63898105-63898105 on Build GRCh38
rs780037278 Chr.18: 63897760-63897760 on Build GRCh38
rs138135796 Chr.18: 63898003-63898003 on Build GRCh38
rs1028951960 Chr.18: 63897922-63897922 on Build GRCh38
rs767647157 Chr.18: 63897754-63897754 on Build GRCh38
rs766217243 Chr.18: 63897841-63897841 on Build GRCh38
rs368842388 Chr.18: 63897850-63897850 on Build GRCh38
rs922200994 Chr.18: 63898092-63898092 on Build GRCh38
rs952050093 Chr.18: 63897936-63897936 on Build GRCh38
rs771666547 Chr.18: 63897774-63897774 on Build GRCh38
rs763385499 Chr.18: 63897814-63897814 on Build GRCh38
rs777403086 Chr.18: 63897875-63897875 on Build GRCh38
rs149666377 Chr.18: 63897767-63897767 on Build GRCh38
rs914859234 Chr.18: 63898068-63898068 on Build GRCh38
rs759802409 Chr.18: 63897725-63897725 on Build GRCh38
rs1012028233 Chr.18: 63897996-63897996 on Build GRCh38
rs893642301 Chr.18: 63897988-63897988 on Build GRCh38
rs1041949734 Chr.18: 63898095-63898095 on Build GRCh38
rs145098902 Chr.18: 63897757-63897757 on Build GRCh38
rs746745519 Chr.18: 63897783-63897783 on Build GRCh38
rs903300835 Chr.18: 63897883-63897883 on Build GRCh38
rs189979351 Chr.18: 63897999-63897999 on Build GRCh38
rs148477234 Chr.18: 63897820-63897820 on Build GRCh38
rs933018967 Chr.18: 63897918-63897918 on Build GRCh38
rs756248213 Chr.18: 63897681-63897681 on Build GRCh38
rs983914248 Chr.18: 63897873-63897873 on Build GRCh38
rs747639390 Chr.18: 63897771-63897771 on Build GRCh38
rs761013904 Chr.18: 63897840-63897840 on Build GRCh38
rs367882625 Chr.18: 63897752-63897752 on Build GRCh38
rs776991929 Chr.18: 63897718-63897718 on Build GRCh38
rs879691869 Chr.18: 63897898-63897898 on Build GRCh38
rs974997241 Chr.18: 63897908-63897908 on Build GRCh38
rs947609068 Chr.18: 63898090-63898090 on Build GRCh38
rs754046467 Chr.18: 63897806-63897807 on Build GRCh38
rs778529592 Chr.18: 63897768-63897768 on Build GRCh38
rs532811143 Chr.18: 63897919-63897922 on Build GRCh38
rs147169778 Chr.18: 63897761-63897761 on Build GRCh38
rs1051418604 Chr.18: 63897966-63897966 on Build GRCh38
rs757844942 Chr.18: 63897874-63897874 on Build GRCh38
rs1016325991 Chr.18: 63898030-63898030 on Build GRCh38
rs922168278 Chr.18: 63897914-63897914 on Build GRCh38
rs374280711 Chr.18: 63897727-63897727 on Build GRCh38
rs923713907 Chr.18: 63897970-63897970 on Build GRCh38
rs145523377 Chr.18: 63897680-63897680 on Build GRCh38
rs963570990 Chr.18: 63898048-63898048 on Build GRCh38
rs1045001030 Chr.18: 63897998-63897998 on Build GRCh38
rs923647716 Chr.18: 63898124-63898124 on Build GRCh38
rs17072020 Chr.18: 63897904-63897904 on Build GRCh38
rs753687273 Chr.18: 63897844-63897844 on Build GRCh38
rs6106 Chr.18: 63897780-63897780 on Build GRCh38
rs779011859 Chr.18: 63897851-63897851 on Build GRCh38
rs974902433 Chr.18: 63898080-63898080 on Build GRCh38

View Full Product Details