Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 65750174-65750684 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH7
Entrez Gene ID
1005
Gene Name
cadherin 7
Gene Aliases
CDH7L1
UniGene
Hs.541003
Gene Chromosome Location
Chr.18: 65750252-65885136 on Build GRCh38
Transcripts
NM_001317214.1 NM_004361.3 NM_033646.2

SNP Detail

SNP ID Chromosome Location
rs1050656976 Chr.18: 65750266-65750266 on Build GRCh38
rs958762775 Chr.18: 65750459-65750459 on Build GRCh38
rs998023930 Chr.18: 65750466-65750466 on Build GRCh38
rs759061863 Chr.18: 65750306-65750306 on Build GRCh38
rs764692003 Chr.18: 65750387-65750387 on Build GRCh38
rs1052749449 Chr.18: 65750627-65750627 on Build GRCh38
rs962481535 Chr.18: 65750383-65750383 on Build GRCh38
rs933890207 Chr.18: 65750642-65750642 on Build GRCh38
rs996902640 Chr.18: 65750260-65750260 on Build GRCh38
rs968638147 Chr.18: 65750472-65750472 on Build GRCh38
rs918821526 Chr.18: 65750301-65750301 on Build GRCh38
rs1007374431 Chr.18: 65750517-65750517 on Build GRCh38
rs973376525 Chr.18: 65750245-65750245 on Build GRCh38
rs948466101 Chr.18: 65750345-65750345 on Build GRCh38
rs397940490 Chr.18: 65750245-65750246 on Build GRCh38
rs899529317 Chr.18: 65750214-65750214 on Build GRCh38
rs1006400492 Chr.18: 65750344-65750344 on Build GRCh38
rs185955249 Chr.18: 65750305-65750305 on Build GRCh38
rs972731152 Chr.18: 65750543-65750543 on Build GRCh38
rs183862550 Chr.18: 65750467-65750467 on Build GRCh38
rs191999213 Chr.18: 65750399-65750399 on Build GRCh38
rs563619161 Chr.18: 65750541-65750541 on Build GRCh38
rs1050944912 Chr.18: 65750654-65750654 on Build GRCh38
rs542889526 Chr.18: 65750590-65750590 on Build GRCh38
rs914555751 Chr.18: 65750470-65750470 on Build GRCh38
rs992319911 Chr.18: 65750557-65750557 on Build GRCh38
rs751085145 Chr.18: 65750569-65750569 on Build GRCh38
rs572579506 Chr.18: 65750331-65750331 on Build GRCh38
rs559310809 Chr.18: 65750597-65750597 on Build GRCh38
rs925695404 Chr.18: 65750603-65750603 on Build GRCh38
rs891680673 Chr.18: 65750452-65750452 on Build GRCh38
rs1026324831 Chr.18: 65750548-65750548 on Build GRCh38
rs529138907 Chr.18: 65750554-65750554 on Build GRCh38
rs150143527 Chr.18: 65750246-65750246 on Build GRCh38
rs578076975 Chr.18: 65750465-65750465 on Build GRCh38
rs888047144 Chr.18: 65750307-65750307 on Build GRCh38
rs574475788 Chr.18: 65750369-65750369 on Build GRCh38
rs977304527 Chr.18: 65750502-65750502 on Build GRCh38
rs935671631 Chr.18: 65750614-65750614 on Build GRCh38
rs935659392 Chr.18: 65750413-65750413 on Build GRCh38
rs1036569345 Chr.18: 65750580-65750581 on Build GRCh38
rs533924855 Chr.18: 65750224-65750225 on Build GRCh38
rs920992627 Chr.18: 65750550-65750550 on Build GRCh38
rs891758777 Chr.18: 65750639-65750639 on Build GRCh38
rs1016960617 Chr.18: 65750524-65750524 on Build GRCh38
rs1053461315 Chr.18: 65750434-65750434 on Build GRCh38
rs898029299 Chr.18: 65750581-65750581 on Build GRCh38
rs889606291 Chr.18: 65750664-65750664 on Build GRCh38
rs888079656 Chr.18: 65750559-65750559 on Build GRCh38
rs917035823 Chr.18: 65750338-65750338 on Build GRCh38
rs375426583 Chr.18: 65750379-65750379 on Build GRCh38
rs1042724261 Chr.18: 65750216-65750216 on Build GRCh38
rs925678990 Chr.18: 65750222-65750222 on Build GRCh38
rs992247787 Chr.18: 65750329-65750329 on Build GRCh38
rs867664716 Chr.18: 65750615-65750615 on Build GRCh38
rs1012979331 Chr.18: 65750638-65750638 on Build GRCh38
rs945677355 Chr.18: 65750212-65750212 on Build GRCh38
rs948495524 Chr.18: 65750596-65750596 on Build GRCh38
rs1002999034 Chr.18: 65750443-65750443 on Build GRCh38
rs889910092 Chr.18: 65750495-65750495 on Build GRCh38
rs1006430090 Chr.18: 65750568-65750568 on Build GRCh38

View Full Product Details