Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 68675161-68675662 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMX3
Entrez Gene ID
54495
Gene Name
thioredoxin related transmembrane protein 3
Gene Aliases
PDIA13, TXNDC10
UniGene
Hs.440534
Gene Chromosome Location
Chr.18: 68673688-68715116 on Build GRCh38
Transcripts
NM_019022.3 XM_005266714.2 XM_011526033.2

SNP Detail

SNP ID Chromosome Location
rs559802487 Chr.18: 68675544-68675544 on Build GRCh38
rs773049092 Chr.18: 68675387-68675387 on Build GRCh38
rs964052718 Chr.18: 68675195-68675195 on Build GRCh38
rs920155434 Chr.18: 68675217-68675217 on Build GRCh38
rs573874795 Chr.18: 68675469-68675469 on Build GRCh38
rs956405613 Chr.18: 68675225-68675225 on Build GRCh38
rs1056127482 Chr.18: 68675374-68675374 on Build GRCh38
rs968946754 Chr.18: 68675575-68675576 on Build GRCh38
rs762654104 Chr.18: 68675623-68675623 on Build GRCh38
rs113263300 Chr.18: 68675443-68675443 on Build GRCh38
rs942921437 Chr.18: 68675615-68675615 on Build GRCh38
rs187140851 Chr.18: 68675582-68675582 on Build GRCh38
rs901252202 Chr.18: 68675457-68675457 on Build GRCh38
rs936996093 Chr.18: 68675350-68675350 on Build GRCh38
rs879451048 Chr.18: 68675559-68675559 on Build GRCh38
rs184511218 Chr.18: 68675524-68675524 on Build GRCh38
rs771905456 Chr.18: 68675337-68675337 on Build GRCh38
rs144939224 Chr.18: 68675617-68675617 on Build GRCh38
rs565534057 Chr.18: 68675388-68675388 on Build GRCh38
rs903004438 Chr.18: 68675409-68675409 on Build GRCh38
rs544822795 Chr.18: 68675536-68675536 on Build GRCh38
rs774215225 Chr.18: 68675234-68675236 on Build GRCh38
rs111798343 Chr.18: 68675240-68675240 on Build GRCh38
rs1005448699 Chr.18: 68675485-68675485 on Build GRCh38
rs551320255 Chr.18: 68675341-68675342 on Build GRCh38
rs144059283 Chr.18: 68675496-68675496 on Build GRCh38
rs527415624 Chr.18: 68675554-68675554 on Build GRCh38
rs914886901 Chr.18: 68675563-68675563 on Build GRCh38
rs922739465 Chr.18: 68675624-68675624 on Build GRCh38
rs893016964 Chr.18: 68675397-68675397 on Build GRCh38
rs976082663 Chr.18: 68675207-68675207 on Build GRCh38
rs911456628 Chr.18: 68675586-68675589 on Build GRCh38
rs745359305 Chr.18: 68675465-68675468 on Build GRCh38
rs983019652 Chr.18: 68675622-68675622 on Build GRCh38
rs964398294 Chr.18: 68675526-68675526 on Build GRCh38
rs969214210 Chr.18: 68675508-68675508 on Build GRCh38
rs1045467869 Chr.18: 68675325-68675325 on Build GRCh38
rs979003370 Chr.18: 68675575-68675575 on Build GRCh38
rs1011447689 Chr.18: 68675398-68675398 on Build GRCh38
rs1023891284 Chr.18: 68675503-68675503 on Build GRCh38
rs940450342 Chr.18: 68675316-68675316 on Build GRCh38
rs554692010 Chr.18: 68675408-68675408 on Build GRCh38
rs1020054428 Chr.18: 68675187-68675187 on Build GRCh38
rs753506161 Chr.18: 68675572-68675572 on Build GRCh38
rs1047081055 Chr.18: 68675482-68675482 on Build GRCh38
rs554279215 Chr.18: 68675223-68675223 on Build GRCh38
rs111665238 Chr.18: 68675591-68675591 on Build GRCh38
rs536631137 Chr.18: 68675334-68675334 on Build GRCh38
rs930016071 Chr.18: 68675626-68675626 on Build GRCh38
rs1047111937 Chr.18: 68675634-68675634 on Build GRCh38
rs982987006 Chr.18: 68675273-68675273 on Build GRCh38
rs218286 Chr.18: 68675484-68675484 on Build GRCh38
rs994193893 Chr.18: 68675461-68675461 on Build GRCh38
rs35374224 Chr.18: 68675378-68675379 on Build GRCh38
rs572586395 Chr.18: 68675275-68675275 on Build GRCh38
rs761677719 Chr.18: 68675329-68675329 on Build GRCh38
rs760438817 Chr.18: 68675473-68675473 on Build GRCh38
rs952859590 Chr.18: 68675246-68675246 on Build GRCh38

View Full Product Details