Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 74631653-74632163 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF407
Entrez Gene ID
55628
Gene Name
zinc finger protein 407
Gene Aliases
-
UniGene
Hs.536490
Gene Chromosome Location
Chr.18: 74597735-75065672 on Build GRCh38
Transcripts
NM_001146189.1 NM_001146190.1 NM_017757.2 XM_005266726.4 XM_006722500.3 XM_011526068.2 XM_011526069.2 XM_011526070.1 XM_017025838.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4743709 c.962G>A p.R321Q Chr.18: 74631981-74631981 on Build GRCh38
3422323 c.955G>T p.G319* Chr.18: 74631974-74631974 on Build GRCh38
1303854 c.971G>A p.G324E Chr.18: 74631990-74631990 on Build GRCh38
6149038 c.978G>T p.T326T Chr.18: 74631997-74631997 on Build GRCh38
5470841 c.792T>C p.L264L Chr.18: 74631811-74631811 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778710668 Chr.18: 74631830-74631830 on Build GRCh38
rs759765873 Chr.18: 74631965-74631965 on Build GRCh38
rs770394467 Chr.18: 74631900-74631900 on Build GRCh38
rs772295581 Chr.18: 74632053-74632053 on Build GRCh38
rs752581430 Chr.18: 74631839-74631839 on Build GRCh38
rs768971490 Chr.18: 74631730-74631730 on Build GRCh38
rs966604763 Chr.18: 74631828-74631828 on Build GRCh38
rs750584810 Chr.18: 74631772-74631772 on Build GRCh38
rs201043970 Chr.18: 74632018-74632018 on Build GRCh38
rs781180214 Chr.18: 74631801-74631801 on Build GRCh38
rs746567804 Chr.18: 74631899-74631899 on Build GRCh38
rs762835052 Chr.18: 74632101-74632101 on Build GRCh38
rs777269843 Chr.18: 74631715-74631715 on Build GRCh38
rs769026348 Chr.18: 74631922-74631922 on Build GRCh38
rs115368653 Chr.18: 74632046-74632046 on Build GRCh38
rs758102217 Chr.18: 74631985-74631985 on Build GRCh38
rs778262377 Chr.18: 74631723-74631723 on Build GRCh38
rs745863286 Chr.18: 74631811-74631811 on Build GRCh38
rs760824473 Chr.18: 74631776-74631776 on Build GRCh38
rs776241248 Chr.18: 74631718-74631718 on Build GRCh38
rs752592082 Chr.18: 74631981-74631981 on Build GRCh38
rs548121332 Chr.18: 74632103-74632103 on Build GRCh38
rs766516797 Chr.18: 74631959-74631959 on Build GRCh38
rs893001848 Chr.18: 74631722-74631722 on Build GRCh38
rs762327804 Chr.18: 74631743-74631743 on Build GRCh38
rs141778977 Chr.18: 74632124-74632124 on Build GRCh38
rs764631895 Chr.18: 74631678-74631678 on Build GRCh38
rs763745169 Chr.18: 74632104-74632104 on Build GRCh38
rs369415246 Chr.18: 74631905-74631905 on Build GRCh38
rs751514308 Chr.18: 74632113-74632113 on Build GRCh38
rs773888680 Chr.18: 74632056-74632056 on Build GRCh38
rs759675097 Chr.18: 74632092-74632092 on Build GRCh38
rs147229491 Chr.18: 74632135-74632135 on Build GRCh38
rs760879417 Chr.18: 74631958-74631958 on Build GRCh38
rs372639119 Chr.18: 74631681-74631681 on Build GRCh38
rs751455269 Chr.18: 74632004-74632004 on Build GRCh38
rs762079638 Chr.18: 74631932-74631932 on Build GRCh38
rs373035128 Chr.18: 74631933-74631933 on Build GRCh38
rs771137840 Chr.18: 74632066-74632066 on Build GRCh38
rs547606522 Chr.18: 74631697-74631697 on Build GRCh38
rs754124154 Chr.18: 74631962-74631962 on Build GRCh38
rs745528057 Chr.18: 74632035-74632035 on Build GRCh38
rs368173554 Chr.18: 74631848-74631848 on Build GRCh38
rs1041132254 Chr.18: 74632128-74632128 on Build GRCh38
rs780851065 Chr.18: 74631904-74631904 on Build GRCh38
rs757314870 Chr.18: 74631682-74631682 on Build GRCh38
rs779798640 Chr.18: 74632139-74632139 on Build GRCh38
rs1020246651 Chr.18: 74631855-74631855 on Build GRCh38
rs376455038 Chr.18: 74632133-74632133 on Build GRCh38
rs766758339 Chr.18: 74631813-74631813 on Build GRCh38
rs750627056 Chr.18: 74631689-74631689 on Build GRCh38
rs749993749 Chr.18: 74632130-74632130 on Build GRCh38
rs535689232 Chr.18: 74631711-74631711 on Build GRCh38
rs746492857 Chr.18: 74631716-74631716 on Build GRCh38
rs754083341 Chr.18: 74631820-74631820 on Build GRCh38
rs774744706 Chr.18: 74631739-74631739 on Build GRCh38
rs893685208 Chr.18: 74631696-74631696 on Build GRCh38
rs77148611 Chr.18: 74632096-74632096 on Build GRCh38
rs959135749 Chr.18: 74631918-74631918 on Build GRCh38
rs774713580 Chr.18: 74631923-74631923 on Build GRCh38
rs756785043 Chr.18: 74632016-74632016 on Build GRCh38
rs748595292 Chr.18: 74632048-74632048 on Build GRCh38
rs763344710 Chr.18: 74631721-74631721 on Build GRCh38
rs767443149 Chr.18: 74632126-74632126 on Build GRCh38
rs769736079 Chr.18: 74632029-74632029 on Build GRCh38
rs567735887 Chr.18: 74631707-74631707 on Build GRCh38
rs780625964 Chr.18: 74632017-74632017 on Build GRCh38
rs558001922 Chr.18: 74632064-74632064 on Build GRCh38
rs776915979 Chr.18: 74632080-74632080 on Build GRCh38
rs780420064 Chr.18: 74631699-74631699 on Build GRCh38
rs376114695 Chr.18: 74631849-74631849 on Build GRCh38
rs754841482 Chr.18: 74631824-74631824 on Build GRCh38
rs770623421 Chr.18: 74631717-74631717 on Build GRCh38
rs767502329 Chr.18: 74631759-74631759 on Build GRCh38
rs866226111 Chr.18: 74631882-74631882 on Build GRCh38
rs555599822 Chr.18: 74631713-74631713 on Build GRCh38
rs897424536 Chr.18: 74631908-74631908 on Build GRCh38
rs369016101 Chr.18: 74632094-74632094 on Build GRCh38
rs377258780 Chr.18: 74631698-74631698 on Build GRCh38
rs575463875 Chr.18: 74631827-74631827 on Build GRCh38
rs200077967 Chr.18: 74632136-74632136 on Build GRCh38
rs773572221 Chr.18: 74631953-74631953 on Build GRCh38
rs970096780 Chr.18: 74631997-74631997 on Build GRCh38
rs765373037 Chr.18: 74631979-74631979 on Build GRCh38
rs745476120 Chr.18: 74631920-74631920 on Build GRCh38

View Full Product Details