Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 39841325-39841832 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIT1
Entrez Gene ID
54802
Gene Name
tRNA isopentenyltransferase 1
Gene Aliases
GRO1, IPPT, IPT, IPTase, MOD5, hGRO1
UniGene
Hs.356554
Gene Chromosome Location
Chr.1: 39841031-39883511 on Build GRCh38
Transcripts
NM_001312691.1 NM_001312692.1 NM_017646.5 NR_132401.1 NR_132402.1 NR_132403.1 NR_132404.1 NR_132405.1 NR_132406.1 NR_132407.1 NR_132408.1 NR_132409.1 NR_132410.1 NR_132412.1 NR_132413.1 NR_132414.1 NR_132415.1 XM_005270954.2 XM_006710706.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7089419 c.1330C>A p.P444T Chr.1: 39841818-39841818 on Build GRCh38
4772455 c.1402_1403insTT p.*468fs? Chr.1: 39841745-39841746 on Build GRCh38
4736359 c.1347A>C p.E449D Chr.1: 39841801-39841801 on Build GRCh38
4736360 c.1331delC p.P444fs*20 Chr.1: 39841817-39841817 on Build GRCh38
1473971 c.1381C>G p.Q461E Chr.1: 39841767-39841767 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1035490091 Chr.1: 39841763-39841763 on Build GRCh38
rs768127290 Chr.1: 39841783-39841783 on Build GRCh38
rs748353166 Chr.1: 39841707-39841708 on Build GRCh38
rs935864491 Chr.1: 39841680-39841680 on Build GRCh38
rs764987272 Chr.1: 39841779-39841779 on Build GRCh38
rs889112032 Chr.1: 39841355-39841355 on Build GRCh38
rs934568539 Chr.1: 39841546-39841546 on Build GRCh38
rs1007592491 Chr.1: 39841377-39841377 on Build GRCh38
rs560729900 Chr.1: 39841708-39841708 on Build GRCh38
rs1029892060 Chr.1: 39841396-39841396 on Build GRCh38
rs915992314 Chr.1: 39841482-39841482 on Build GRCh38
rs775461671 Chr.1: 39841749-39841749 on Build GRCh38
rs368883851 Chr.1: 39841576-39841576 on Build GRCh38
rs1000287360 Chr.1: 39841352-39841352 on Build GRCh38
rs1004427831 Chr.1: 39841671-39841671 on Build GRCh38
rs3215539 Chr.1: 39841494-39841495 on Build GRCh38
rs981287489 Chr.1: 39841753-39841753 on Build GRCh38
rs745713927 Chr.1: 39841737-39841737 on Build GRCh38
rs917134011 Chr.1: 39841624-39841624 on Build GRCh38
rs749480271 Chr.1: 39841718-39841718 on Build GRCh38
rs749999525 Chr.1: 39841476-39841476 on Build GRCh38
rs955596748 Chr.1: 39841409-39841409 on Build GRCh38
rs969910123 Chr.1: 39841487-39841487 on Build GRCh38
rs557546197 Chr.1: 39841442-39841442 on Build GRCh38
rs958353221 Chr.1: 39841602-39841602 on Build GRCh38
rs199579505 Chr.1: 39841750-39841750 on Build GRCh38
rs781773936 Chr.1: 39841700-39841700 on Build GRCh38
rs951166027 Chr.1: 39841513-39841513 on Build GRCh38
rs963119592 Chr.1: 39841448-39841448 on Build GRCh38
rs777746600 Chr.1: 39841795-39841795 on Build GRCh38
rs949978380 Chr.1: 39841636-39841636 on Build GRCh38
rs200337239 Chr.1: 39841706-39841706 on Build GRCh38
rs1051175606 Chr.1: 39841655-39841655 on Build GRCh38
rs914203959 Chr.1: 39841587-39841587 on Build GRCh38
rs368380890 Chr.1: 39841807-39841807 on Build GRCh38
rs753273813 Chr.1: 39841784-39841784 on Build GRCh38
rs575120115 Chr.1: 39841402-39841402 on Build GRCh38
rs928556795 Chr.1: 39841495-39841495 on Build GRCh38
rs753015621 Chr.1: 39841782-39841782 on Build GRCh38
rs909658376 Chr.1: 39841586-39841586 on Build GRCh38
rs771968271 Chr.1: 39841739-39841739 on Build GRCh38
rs996987612 Chr.1: 39841648-39841648 on Build GRCh38
rs891384840 Chr.1: 39841667-39841667 on Build GRCh38
rs565017816 Chr.1: 39841621-39841621 on Build GRCh38
rs770195357 Chr.1: 39841723-39841723 on Build GRCh38
rs537752574 Chr.1: 39841427-39841427 on Build GRCh38
rs966303710 Chr.1: 39841417-39841417 on Build GRCh38
rs1015559345 Chr.1: 39841441-39841441 on Build GRCh38
rs761390528 Chr.1: 39841768-39841768 on Build GRCh38
rs571940493 Chr.1: 39841630-39841630 on Build GRCh38
rs554236571 Chr.1: 39841706-39841706 on Build GRCh38
rs1040412358 Chr.1: 39841734-39841734 on Build GRCh38
rs1041492013 Chr.1: 39841647-39841647 on Build GRCh38
rs974222079 Chr.1: 39841464-39841464 on Build GRCh38
rs3180539 Chr.1: 39841494-39841494 on Build GRCh38
rs74069777 Chr.1: 39841605-39841605 on Build GRCh38
rs1033548301 Chr.1: 39841354-39841354 on Build GRCh38
rs767890 Chr.1: 39841376-39841376 on Build GRCh38
rs397722768 Chr.1: 39841503-39841504 on Build GRCh38
rs773596178 Chr.1: 39841809-39841813 on Build GRCh38
rs766681378 Chr.1: 39841696-39841696 on Build GRCh38
rs748236801 Chr.1: 39841705-39841705 on Build GRCh38
rs755516143 Chr.1: 39841698-39841698 on Build GRCh38
rs183734442 Chr.1: 39841535-39841535 on Build GRCh38
rs375032892 Chr.1: 39841713-39841713 on Build GRCh38
rs770720261 Chr.1: 39841733-39841733 on Build GRCh38
rs397840109 Chr.1: 39841504-39841505 on Build GRCh38
rs187949560 Chr.1: 39841702-39841702 on Build GRCh38
rs768054657 Chr.1: 39841767-39841767 on Build GRCh38
rs755544912 Chr.1: 39841479-39841479 on Build GRCh38
rs983885012 Chr.1: 39841561-39841561 on Build GRCh38
rs924464634 Chr.1: 39841652-39841652 on Build GRCh38
rs768103035 Chr.1: 39841657-39841657 on Build GRCh38

View Full Product Details