Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 40624782-40625288 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RIMS3
Entrez Gene ID
9783
Gene Name
regulating synaptic membrane exocytosis 3
Gene Aliases
NIM3, RIM3
UniGene
Hs.654808
Gene Chromosome Location
Chr.1: 40620680-40692104 on Build GRCh38
Transcripts
NM_014747.2 XM_011542479.2

SNP Detail

SNP ID Chromosome Location
rs550007036 Chr.1: 40625103-40625103 on Build GRCh38
rs750514293 Chr.1: 40624854-40624854 on Build GRCh38
rs569848079 Chr.1: 40624850-40624850 on Build GRCh38
rs999946654 Chr.1: 40624817-40624817 on Build GRCh38
rs1032910980 Chr.1: 40624818-40624818 on Build GRCh38
rs187141336 Chr.1: 40625019-40625019 on Build GRCh38
rs945415884 Chr.1: 40625247-40625247 on Build GRCh38
rs962873013 Chr.1: 40624923-40624924 on Build GRCh38
rs953267825 Chr.1: 40624835-40624835 on Build GRCh38
rs938223564 Chr.1: 40624976-40624976 on Build GRCh38
rs938299660 Chr.1: 40625180-40625180 on Build GRCh38
rs1017472595 Chr.1: 40625151-40625151 on Build GRCh38
rs911567997 Chr.1: 40625006-40625006 on Build GRCh38
rs1039224257 Chr.1: 40625016-40625016 on Build GRCh38
rs150920491 Chr.1: 40624864-40624864 on Build GRCh38
rs895164161 Chr.1: 40625120-40625120 on Build GRCh38
rs984978750 Chr.1: 40625138-40625138 on Build GRCh38
rs140723827 Chr.1: 40624865-40624865 on Build GRCh38
rs114243295 Chr.1: 40625143-40625143 on Build GRCh38
rs535235899 Chr.1: 40624997-40624997 on Build GRCh38
rs190805337 Chr.1: 40625020-40625020 on Build GRCh38
rs926889576 Chr.1: 40624953-40624953 on Build GRCh38
rs903925159 Chr.1: 40625030-40625030 on Build GRCh38
rs181721891 Chr.1: 40625119-40625119 on Build GRCh38
rs528606420 Chr.1: 40624951-40624951 on Build GRCh38
rs186114730 Chr.1: 40625235-40625235 on Build GRCh38
rs532334137 Chr.1: 40624840-40624840 on Build GRCh38
rs955631052 Chr.1: 40624986-40624986 on Build GRCh38
rs1017086707 Chr.1: 40624894-40624894 on Build GRCh38
rs773522849 Chr.1: 40624935-40624935 on Build GRCh38
rs991323440 Chr.1: 40624882-40624882 on Build GRCh38
rs971846832 Chr.1: 40624886-40624886 on Build GRCh38
rs953319881 Chr.1: 40625062-40625062 on Build GRCh38
rs569968139 Chr.1: 40625104-40625104 on Build GRCh38
rs930815951 Chr.1: 40624918-40624918 on Build GRCh38
rs767465065 Chr.1: 40625239-40625239 on Build GRCh38
rs918969793 Chr.1: 40624901-40624901 on Build GRCh38
rs992432410 Chr.1: 40625206-40625206 on Build GRCh38
rs959461505 Chr.1: 40624874-40624874 on Build GRCh38
rs896480643 Chr.1: 40625012-40625012 on Build GRCh38
rs1007468741 Chr.1: 40625089-40625089 on Build GRCh38
rs1019244313 Chr.1: 40625093-40625093 on Build GRCh38
rs962843917 Chr.1: 40625159-40625159 on Build GRCh38
rs1032794858 Chr.1: 40625052-40625052 on Build GRCh38
rs994263570 Chr.1: 40625177-40625177 on Build GRCh38
rs955600074 Chr.1: 40625242-40625242 on Build GRCh38
rs774482757 Chr.1: 40625106-40625106 on Build GRCh38
rs1020908506 Chr.1: 40624832-40624832 on Build GRCh38
rs1000000440 Chr.1: 40625031-40625031 on Build GRCh38

View Full Product Details