Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 77015409-77015904 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MBP
Entrez Gene ID
4155
Gene Name
myelin basic protein
Gene Aliases
-
UniGene
Hs.551713
Gene Chromosome Location
Chr.18: 76978833-77133708 on Build GRCh38
Transcripts
NM_001025081.1 NM_001025090.1 NM_001025092.1 NM_001025100.1 NM_001025101.1 NM_002385.2 XM_011526009.2 XM_017025778.1 XM_017025780.1

SNP Detail

SNP ID Chromosome Location
rs530584412 Chr.18: 77015583-77015583 on Build GRCh38
rs1044394315 Chr.18: 77015652-77015652 on Build GRCh38
rs569496406 Chr.18: 77015844-77015844 on Build GRCh38
rs770523788 Chr.18: 77015856-77015856 on Build GRCh38
rs576640710 Chr.18: 77015452-77015452 on Build GRCh38
rs1005866452 Chr.18: 77015744-77015744 on Build GRCh38
rs986676494 Chr.18: 77015684-77015684 on Build GRCh38
rs7232502 Chr.18: 77015662-77015662 on Build GRCh38
rs907781333 Chr.18: 77015447-77015447 on Build GRCh38
rs62105705 Chr.18: 77015743-77015743 on Build GRCh38
rs551874335 Chr.18: 77015593-77015593 on Build GRCh38
rs911294018 Chr.18: 77015855-77015855 on Build GRCh38
rs118032229 Chr.18: 77015795-77015795 on Build GRCh38
rs536166384 Chr.18: 77015764-77015764 on Build GRCh38
rs894297710 Chr.18: 77015485-77015485 on Build GRCh38
rs142844021 Chr.18: 77015625-77015625 on Build GRCh38
rs192162300 Chr.18: 77015676-77015676 on Build GRCh38
rs552795703 Chr.18: 77015872-77015872 on Build GRCh38
rs1056575271 Chr.18: 77015696-77015696 on Build GRCh38
rs545664087 Chr.18: 77015466-77015466 on Build GRCh38
rs140658405 Chr.18: 77015562-77015562 on Build GRCh38
rs369125527 Chr.18: 77015853-77015853 on Build GRCh38
rs539232882 Chr.18: 77015862-77015862 on Build GRCh38
rs147406960 Chr.18: 77015751-77015751 on Build GRCh38
rs998595550 Chr.18: 77015828-77015828 on Build GRCh38
rs1031337288 Chr.18: 77015594-77015594 on Build GRCh38
rs556836957 Chr.18: 77015451-77015451 on Build GRCh38
rs12959006 Chr.18: 77015675-77015675 on Build GRCh38
rs183485580 Chr.18: 77015705-77015705 on Build GRCh38
rs7236968 Chr.18: 77015547-77015547 on Build GRCh38
rs946737766 Chr.18: 77015639-77015639 on Build GRCh38
rs867433691 Chr.18: 77015740-77015740 on Build GRCh38
rs930561371 Chr.18: 77015467-77015467 on Build GRCh38
rs944753966 Chr.18: 77015715-77015715 on Build GRCh38
rs183398239 Chr.18: 77015533-77015533 on Build GRCh38
rs745349296 Chr.18: 77015863-77015863 on Build GRCh38
rs1049403697 Chr.18: 77015484-77015484 on Build GRCh38
rs897750381 Chr.18: 77015460-77015460 on Build GRCh38
rs541458492 Chr.18: 77015512-77015512 on Build GRCh38
rs187100049 Chr.18: 77015549-77015549 on Build GRCh38
rs781703731 Chr.18: 77015829-77015829 on Build GRCh38
rs972078930 Chr.18: 77015726-77015726 on Build GRCh38
rs906988243 Chr.18: 77015822-77015822 on Build GRCh38
rs1015283322 Chr.18: 77015746-77015746 on Build GRCh38
rs572836342 Chr.18: 77015509-77015509 on Build GRCh38
rs552826249 Chr.18: 77015506-77015506 on Build GRCh38
rs117335468 Chr.18: 77015727-77015727 on Build GRCh38
rs551516330 Chr.18: 77015850-77015850 on Build GRCh38
rs989735742 Chr.18: 77015630-77015630 on Build GRCh38

View Full Product Details