Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 79715206-79715711 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTDP1
Entrez Gene ID
9150
Gene Name
CTD phosphatase subunit 1
Gene Aliases
CCFDN, FCP1
UniGene
Hs.465490
Gene Chromosome Location
Chr.18: 79679801-79754510 on Build GRCh38
Transcripts
NM_001202504.1 NM_001318511.1 NM_004715.4 NM_048368.3 XM_011526261.1 XM_017026078.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
306954 c.2028C>T p.D676D Chr.18: 79715488-79715488 on Build GRCh38
361929 c.1823A>T p.Y608F Chr.18: 79715283-79715283 on Build GRCh38
3712716 c.1780G>A p.E594K Chr.18: 79715240-79715240 on Build GRCh38
708816 c.1962G>C p.T654T Chr.18: 79715422-79715422 on Build GRCh38
310385 c.1880C>T p.P627L Chr.18: 79715340-79715340 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749653694 Chr.18: 79715518-79715518 on Build GRCh38
rs765183908 Chr.18: 79715398-79715398 on Build GRCh38
rs563093692 Chr.18: 79715669-79715669 on Build GRCh38
rs753719249 Chr.18: 79715297-79715297 on Build GRCh38
rs753615274 Chr.18: 79715397-79715397 on Build GRCh38
rs977934934 Chr.18: 79715661-79715661 on Build GRCh38
rs201937277 Chr.18: 79715261-79715261 on Build GRCh38
rs577434989 Chr.18: 79715402-79715402 on Build GRCh38
rs762993214 Chr.18: 79715570-79715570 on Build GRCh38
rs145290303 Chr.18: 79715640-79715640 on Build GRCh38
rs746652553 Chr.18: 79715422-79715422 on Build GRCh38
rs764076790 Chr.18: 79715267-79715267 on Build GRCh38
rs761182450 Chr.18: 79715618-79715619 on Build GRCh38
rs377483381 Chr.18: 79715503-79715503 on Build GRCh38
rs79944224 Chr.18: 79715375-79715375 on Build GRCh38
rs149848787 Chr.18: 79715256-79715256 on Build GRCh38
rs747075037 Chr.18: 79715536-79715536 on Build GRCh38
rs200244999 Chr.18: 79715568-79715568 on Build GRCh38
rs754882827 Chr.18: 79715298-79715298 on Build GRCh38
rs1034218084 Chr.18: 79715399-79715399 on Build GRCh38
rs762136157 Chr.18: 79715280-79715280 on Build GRCh38
rs780295853 Chr.18: 79715424-79715424 on Build GRCh38
rs1011277105 Chr.18: 79715531-79715531 on Build GRCh38
rs11549119 Chr.18: 79715455-79715455 on Build GRCh38
rs769147755 Chr.18: 79715328-79715328 on Build GRCh38
rs1019341209 Chr.18: 79715574-79715574 on Build GRCh38
rs756863087 Chr.18: 79715423-79715423 on Build GRCh38
rs369463770 Chr.18: 79715488-79715488 on Build GRCh38
rs752298646 Chr.18: 79715410-79715410 on Build GRCh38
rs771102094 Chr.18: 79715540-79715540 on Build GRCh38
rs755171841 Chr.18: 79715645-79715645 on Build GRCh38
rs756710537 Chr.18: 79715300-79715302 on Build GRCh38
rs761839272 Chr.18: 79715336-79715336 on Build GRCh38
rs755884007 Chr.18: 79715288-79715288 on Build GRCh38
rs748659205 Chr.18: 79715523-79715523 on Build GRCh38
rs147092934 Chr.18: 79715446-79715446 on Build GRCh38
rs145888904 Chr.18: 79715309-79715309 on Build GRCh38
rs542907664 Chr.18: 79715534-79715534 on Build GRCh38
rs763374239 Chr.18: 79715320-79715320 on Build GRCh38
rs375108513 Chr.18: 79715317-79715317 on Build GRCh38
rs760531133 Chr.18: 79715372-79715372 on Build GRCh38
rs772510890 Chr.18: 79715526-79715526 on Build GRCh38
rs746280862 Chr.18: 79715311-79715311 on Build GRCh38
rs928756841 Chr.18: 79715674-79715674 on Build GRCh38
rs914758233 Chr.18: 79715652-79715652 on Build GRCh38
rs372238734 Chr.18: 79715289-79715289 on Build GRCh38
rs775354745 Chr.18: 79715569-79715569 on Build GRCh38
rs980615603 Chr.18: 79715400-79715400 on Build GRCh38
rs1054633467 Chr.18: 79715486-79715486 on Build GRCh38
rs975088687 Chr.18: 79715618-79715618 on Build GRCh38
rs775836672 Chr.18: 79715263-79715263 on Build GRCh38
rs747051978 Chr.18: 79715470-79715470 on Build GRCh38
rs374774711 Chr.18: 79715431-79715431 on Build GRCh38
rs893733140 Chr.18: 79715520-79715520 on Build GRCh38
rs763697720 Chr.18: 79715489-79715489 on Build GRCh38
rs763149665 Chr.18: 79715264-79715264 on Build GRCh38
rs374209226 Chr.18: 79715502-79715502 on Build GRCh38
rs776891959 Chr.18: 79715542-79715542 on Build GRCh38
rs137986892 Chr.18: 79715447-79715447 on Build GRCh38
rs965476971 Chr.18: 79715612-79715612 on Build GRCh38
rs564610662 Chr.18: 79715585-79715585 on Build GRCh38
rs868166103 Chr.18: 79715510-79715510 on Build GRCh38
rs756882272 Chr.18: 79715506-79715506 on Build GRCh38
rs566825980 Chr.18: 79715308-79715308 on Build GRCh38
rs1027956462 Chr.18: 79715639-79715639 on Build GRCh38
rs766523914 Chr.18: 79715519-79715519 on Build GRCh38
rs777623040 Chr.18: 79715421-79715421 on Build GRCh38
rs141527633 Chr.18: 79715558-79715558 on Build GRCh38
rs776128110 Chr.18: 79715318-79715318 on Build GRCh38
rs143177926 Chr.18: 79715368-79715368 on Build GRCh38
rs79257976 Chr.18: 79715573-79715573 on Build GRCh38
rs776325532 Chr.18: 79715458-79715458 on Build GRCh38
rs990397085 Chr.18: 79715648-79715648 on Build GRCh38
rs907197199 Chr.18: 79715386-79715386 on Build GRCh38
rs374004081 Chr.18: 79715442-79715442 on Build GRCh38
rs76553883 Chr.18: 79715659-79715659 on Build GRCh38
rs757956731 Chr.18: 79715413-79715413 on Build GRCh38
rs779464326 Chr.18: 79715521-79715521 on Build GRCh38
rs767674446 Chr.18: 79715284-79715284 on Build GRCh38
rs377634005 Chr.18: 79715443-79715443 on Build GRCh38
rs932192170 Chr.18: 79715478-79715478 on Build GRCh38
rs867198820 Chr.18: 79715505-79715505 on Build GRCh38
rs758577959 Chr.18: 79715468-79715468 on Build GRCh38
rs767778473 Chr.18: 79715341-79715341 on Build GRCh38
rs762033206 Chr.18: 79715278-79715278 on Build GRCh38
rs749615075 Chr.18: 79715428-79715428 on Build GRCh38
rs751586923 Chr.18: 79715271-79715271 on Build GRCh38
rs931327104 Chr.18: 79715624-79715624 on Build GRCh38
rs774316208 Chr.18: 79715335-79715335 on Build GRCh38
rs750247555 Chr.18: 79715287-79715287 on Build GRCh38
rs752664929 Chr.18: 79715469-79715469 on Build GRCh38
rs1054604568 Chr.18: 79715319-79715319 on Build GRCh38
rs769072618 Chr.18: 79715437-79715437 on Build GRCh38
rs918841277 Chr.18: 79715438-79715438 on Build GRCh38
rs938737792 Chr.18: 79715679-79715679 on Build GRCh38

View Full Product Details