Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 759821-760326 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MISP
Entrez Gene ID
126353
Gene Name
mitotic spindle positioning
Gene Aliases
C19orf21, MISP1
UniGene
Hs.439180
Gene Chromosome Location
Chr.19: 748413-764319 on Build GRCh38
Transcripts
NM_173481.3 NR_135168.1 XM_011527685.2 XM_011527686.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4983573 c.1871T>A p.V624E Chr.19: 759999-759999 on Build GRCh38
4843242 c.1851G>A p.A617A Chr.19: 759979-759979 on Build GRCh38
6306607 c.1890G>T p.G630G Chr.19: 760018-760018 on Build GRCh38
4829647 c.1787C>A p.T596K Chr.19: 759915-759915 on Build GRCh38
3541286 c.1882C>T p.P628S Chr.19: 760010-760010 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs557537362 Chr.19: 760296-760296 on Build GRCh38
rs747102485 Chr.19: 759951-759951 on Build GRCh38
rs1010133549 Chr.19: 760161-760161 on Build GRCh38
rs769680799 Chr.19: 759960-759960 on Build GRCh38
rs545631552 Chr.19: 760272-760272 on Build GRCh38
rs753065377 Chr.19: 760072-760072 on Build GRCh38
rs918132967 Chr.19: 760192-760192 on Build GRCh38
rs918408714 Chr.19: 760258-760258 on Build GRCh38
rs371491651 Chr.19: 759973-759973 on Build GRCh38
rs369571645 Chr.19: 760055-760055 on Build GRCh38
rs75424782 Chr.19: 759864-759864 on Build GRCh38
rs373266238 Chr.19: 760087-760087 on Build GRCh38
rs1031226188 Chr.19: 760159-760159 on Build GRCh38
rs190322853 Chr.19: 759985-759985 on Build GRCh38
rs752388310 Chr.19: 759891-759891 on Build GRCh38
rs534693105 Chr.19: 760212-760212 on Build GRCh38
rs969871572 Chr.19: 760195-760195 on Build GRCh38
rs960141426 Chr.19: 760247-760247 on Build GRCh38
rs202184785 Chr.19: 759979-759979 on Build GRCh38
rs199558284 Chr.19: 760298-760299 on Build GRCh38
rs755222104 Chr.19: 760025-760027 on Build GRCh38
rs930233207 Chr.19: 760003-760003 on Build GRCh38
rs73921311 Chr.19: 759890-759890 on Build GRCh38
rs756519229 Chr.19: 759923-759923 on Build GRCh38
rs773363192 Chr.19: 759948-759948 on Build GRCh38
rs527351392 Chr.19: 759959-759959 on Build GRCh38
rs375797268 Chr.19: 760162-760162 on Build GRCh38
rs567667163 Chr.19: 760208-760208 on Build GRCh38
rs891479009 Chr.19: 760157-760157 on Build GRCh38
rs942030297 Chr.19: 760241-760241 on Build GRCh38
rs745876662 Chr.19: 760035-760035 on Build GRCh38
rs778165912 Chr.19: 760008-760008 on Build GRCh38
rs1006673863 Chr.19: 760042-760042 on Build GRCh38
rs780631710 Chr.19: 759925-759925 on Build GRCh38
rs144706404 Chr.19: 760041-760041 on Build GRCh38
rs554061987 Chr.19: 760245-760245 on Build GRCh38
rs375065737 Chr.19: 760049-760049 on Build GRCh38
rs112194249 Chr.19: 760036-760036 on Build GRCh38
rs369969065 Chr.19: 760004-760004 on Build GRCh38
rs748435161 Chr.19: 759928-759928 on Build GRCh38
rs376144112 Chr.19: 760066-760066 on Build GRCh38
rs767109142 Chr.19: 760060-760060 on Build GRCh38
rs900253004 Chr.19: 760069-760069 on Build GRCh38
rs757882556 Chr.19: 759906-759906 on Build GRCh38
rs62131316 Chr.19: 760302-760302 on Build GRCh38
rs543106836 Chr.19: 759859-759859 on Build GRCh38
rs770727929 Chr.19: 759953-759953 on Build GRCh38
rs772278393 Chr.19: 759942-759942 on Build GRCh38
rs759399873 Chr.19: 759882-759882 on Build GRCh38
rs548087869 Chr.19: 759905-759905 on Build GRCh38
rs367612583 Chr.19: 759927-759927 on Build GRCh38
rs185794418 Chr.19: 759978-759978 on Build GRCh38
rs763652188 Chr.19: 759911-759911 on Build GRCh38
rs980909750 Chr.19: 760205-760205 on Build GRCh38
rs62131317 Chr.19: 760304-760304 on Build GRCh38
rs780970700 Chr.19: 760032-760032 on Build GRCh38
rs773912296 Chr.19: 760218-760218 on Build GRCh38
rs774030536 Chr.19: 760054-760054 on Build GRCh38
rs960272011 Chr.19: 759965-759965 on Build GRCh38
rs552002916 Chr.19: 759995-759995 on Build GRCh38
rs201079131 Chr.19: 759947-759947 on Build GRCh38
rs758867759 Chr.19: 760082-760082 on Build GRCh38
rs768255410 Chr.19: 760053-760053 on Build GRCh38
rs896960254 Chr.19: 760016-760016 on Build GRCh38
rs566362835 Chr.19: 759956-759956 on Build GRCh38
rs373129325 Chr.19: 760015-760015 on Build GRCh38
rs779323000 Chr.19: 760060-760061 on Build GRCh38
rs765856233 Chr.19: 760071-760071 on Build GRCh38
rs192745978 Chr.19: 760068-760068 on Build GRCh38
rs763748517 Chr.19: 759974-759974 on Build GRCh38
rs1001049191 Chr.19: 760121-760121 on Build GRCh38
rs779483029 Chr.19: 759990-759990 on Build GRCh38
rs866642002 Chr.19: 760190-760190 on Build GRCh38
rs575782486 Chr.19: 760299-760299 on Build GRCh38
rs386805671 Chr.19: 760302-760304 on Build GRCh38
rs968558132 Chr.19: 759915-759915 on Build GRCh38
rs935520017 Chr.19: 759872-759872 on Build GRCh38
rs778112481 Chr.19: 760084-760084 on Build GRCh38
rs747022398 Chr.19: 760014-760014 on Build GRCh38
rs138014814 Chr.19: 759916-759916 on Build GRCh38
rs201970290 Chr.19: 760303-760303 on Build GRCh38
rs113603391 Chr.19: 760251-760251 on Build GRCh38
rs775111216 Chr.19: 759961-759961 on Build GRCh38
rs773265063 Chr.19: 759863-759863 on Build GRCh38
rs199785759 Chr.19: 760304-760304 on Build GRCh38
rs776233756 Chr.19: 759875-759875 on Build GRCh38
rs1018344814 Chr.19: 760057-760057 on Build GRCh38

View Full Product Details