Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1074204-1074720 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP45
Entrez Gene ID
23526
Gene Name
Rho GTPase activating protein 45
Gene Aliases
HA-1, HLA-HA1, HMHA1
UniGene
Hs.465521
Gene Chromosome Location
Chr.19: 1065923-1087831 on Build GRCh38
Transcripts
NM_001258328.2 NM_001282335.1 NM_001321232.1 NM_012292.4 XM_006722713.1 XM_011527858.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5165659 c.514-1G>A p.? Chr.19: 1074613-1074613 on Build GRCh38
4073907 c.999C>T p.H333H Chr.19: 1074619-1074619 on Build GRCh38
474134 c.1048T>A p.F350I Chr.19: 1074668-1074668 on Build GRCh38
4884080 c.511G>T p.E171* Chr.19: 1074405-1074405 on Build GRCh38
6414561 c.1018T>C p.Y340H Chr.19: 1074638-1074638 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145275771 Chr.19: 1074313-1074314 on Build GRCh38
rs1016301591 Chr.19: 1074328-1074328 on Build GRCh38
rs760660422 Chr.19: 1074309-1074309 on Build GRCh38
rs773831902 Chr.19: 1074677-1074677 on Build GRCh38
rs544484826 Chr.19: 1074444-1074444 on Build GRCh38
rs367566451 Chr.19: 1074336-1074336 on Build GRCh38
rs984085053 Chr.19: 1074516-1074516 on Build GRCh38
rs546916938 Chr.19: 1074648-1074648 on Build GRCh38
rs375278742 Chr.19: 1074307-1074307 on Build GRCh38
rs772113900 Chr.19: 1074393-1074393 on Build GRCh38
rs780363408 Chr.19: 1074350-1074350 on Build GRCh38
rs11084881 Chr.19: 1074253-1074253 on Build GRCh38
rs758030000 Chr.19: 1074325-1074325 on Build GRCh38
rs376489330 Chr.19: 1074466-1074466 on Build GRCh38
rs374937382 Chr.19: 1074459-1074459 on Build GRCh38
rs766167132 Chr.19: 1074313-1074313 on Build GRCh38
rs964962548 Chr.19: 1074449-1074449 on Build GRCh38
rs575124517 Chr.19: 1074375-1074375 on Build GRCh38
rs766187310 Chr.19: 1074254-1074254 on Build GRCh38
rs371078841 Chr.19: 1074487-1074487 on Build GRCh38
rs369822039 Chr.19: 1074564-1074564 on Build GRCh38
rs779143760 Chr.19: 1074373-1074373 on Build GRCh38
rs769688537 Chr.19: 1074429-1074429 on Build GRCh38
rs775430041 Chr.19: 1074630-1074630 on Build GRCh38
rs746672216 Chr.19: 1074278-1074278 on Build GRCh38
rs746362953 Chr.19: 1074277-1074277 on Build GRCh38
rs775375720 Chr.19: 1074454-1074455 on Build GRCh38
rs200436029 Chr.19: 1074231-1074231 on Build GRCh38
rs933757344 Chr.19: 1074587-1074587 on Build GRCh38
rs765084467 Chr.19: 1074450-1074451 on Build GRCh38
rs753505391 Chr.19: 1074256-1074256 on Build GRCh38
rs772119525 Chr.19: 1074303-1074303 on Build GRCh38
rs942226123 Chr.19: 1074437-1074437 on Build GRCh38
rs747144536 Chr.19: 1074599-1074599 on Build GRCh38
rs776394067 Chr.19: 1074411-1074411 on Build GRCh38
rs1009469323 Chr.19: 1074673-1074673 on Build GRCh38
rs372343317 Chr.19: 1074281-1074281 on Build GRCh38
rs749545384 Chr.19: 1074283-1074283 on Build GRCh38
rs755395589 Chr.19: 1074580-1074580 on Build GRCh38
rs776360887 Chr.19: 1074325-1074325 on Build GRCh38
rs578092303 Chr.19: 1074536-1074536 on Build GRCh38
rs199745412 Chr.19: 1074595-1074595 on Build GRCh38
rs769777568 Chr.19: 1074628-1074628 on Build GRCh38
rs759240698 Chr.19: 1074444-1074445 on Build GRCh38
rs757944288 Chr.19: 1074267-1074267 on Build GRCh38
rs774504449 Chr.19: 1074291-1074291 on Build GRCh38
rs781588672 Chr.19: 1074268-1074268 on Build GRCh38
rs781329831 Chr.19: 1074617-1074617 on Build GRCh38
rs12974537 Chr.19: 1074316-1074316 on Build GRCh38
rs759446297 Chr.19: 1074414-1074414 on Build GRCh38
rs773173588 Chr.19: 1074250-1074250 on Build GRCh38
rs202170443 Chr.19: 1074319-1074319 on Build GRCh38
rs371129613 Chr.19: 1074447-1074448 on Build GRCh38
rs755293165 Chr.19: 1074360-1074360 on Build GRCh38
rs113175128 Chr.19: 1074341-1074341 on Build GRCh38
rs1044025613 Chr.19: 1074640-1074640 on Build GRCh38
rs534098093 Chr.19: 1074501-1074501 on Build GRCh38
rs371367357 Chr.19: 1074410-1074410 on Build GRCh38
rs748360399 Chr.19: 1074382-1074382 on Build GRCh38
rs747003749 Chr.19: 1074402-1074402 on Build GRCh38
rs569021641 Chr.19: 1074359-1074359 on Build GRCh38
rs761820565 Chr.19: 1074237-1074237 on Build GRCh38
rs761422984 Chr.19: 1074691-1074691 on Build GRCh38
rs766937574 Chr.19: 1074450-1074450 on Build GRCh38
rs111831177 Chr.19: 1074598-1074598 on Build GRCh38
rs895642507 Chr.19: 1074301-1074301 on Build GRCh38
rs528751431 Chr.19: 1074647-1074647 on Build GRCh38
rs762930661 Chr.19: 1074571-1074571 on Build GRCh38
rs776499654 Chr.19: 1074314-1074314 on Build GRCh38
rs748256203 Chr.19: 1074297-1074297 on Build GRCh38
rs752278644 Chr.19: 1074263-1074263 on Build GRCh38
rs1017795745 Chr.19: 1074448-1074448 on Build GRCh38
rs113245134 Chr.19: 1074340-1074340 on Build GRCh38
rs988974785 Chr.19: 1074502-1074502 on Build GRCh38
rs187324975 Chr.19: 1074540-1074540 on Build GRCh38
rs763645536 Chr.19: 1074327-1074327 on Build GRCh38
rs942440025 Chr.19: 1074311-1074311 on Build GRCh38
rs758664112 Chr.19: 1074596-1074596 on Build GRCh38
rs764044863 Chr.19: 1074607-1074607 on Build GRCh38
rs890569928 Chr.19: 1074623-1074623 on Build GRCh38
rs374747409 Chr.19: 1074324-1074324 on Build GRCh38
rs978392249 Chr.19: 1074489-1074489 on Build GRCh38
rs770885669 Chr.19: 1074605-1074605 on Build GRCh38
rs909536007 Chr.19: 1074522-1074522 on Build GRCh38
rs570931229 Chr.19: 1074261-1074261 on Build GRCh38
rs754682303 Chr.19: 1074259-1074259 on Build GRCh38
rs575792246 Chr.19: 1074593-1074593 on Build GRCh38
rs745785243 Chr.19: 1074625-1074625 on Build GRCh38
rs74422982 Chr.19: 1074511-1074511 on Build GRCh38
rs761175961 Chr.19: 1074447-1074447 on Build GRCh38
rs768875377 Chr.19: 1074287-1074287 on Build GRCh38
rs1017764456 Chr.19: 1074683-1074683 on Build GRCh38
rs199502722 Chr.19: 1074436-1074436 on Build GRCh38
rs200347805 Chr.19: 1074431-1074431 on Build GRCh38
rs751089939 Chr.19: 1074446-1074446 on Build GRCh38
rs925661664 Chr.19: 1074490-1074490 on Build GRCh38

View Full Product Details