Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1110928-1111422 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SBNO2
Entrez Gene ID
22904
Gene Name
strawberry notch homolog 2
Gene Aliases
KIAA0963, SNO, STNO
UniGene
Hs.408708
Gene Chromosome Location
Chr.19: 1107634-1174283 on Build GRCh38
Transcripts
NM_001100122.1 NM_014963.2 XM_011527804.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
710129 c.2879G>A p.R960Q Chr.19: 1111045-1111045 on Build GRCh38
1390180 c.2863G>A p.G955S Chr.19: 1111061-1111061 on Build GRCh38
1390179 c.2842G>A p.G948S Chr.19: 1111061-1111061 on Build GRCh38
4600572 c.2857C>T p.R953W Chr.19: 1111046-1111046 on Build GRCh38
6411483 c.2819A>G p.Q940R Chr.19: 1111084-1111084 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs530504329 Chr.19: 1111224-1111224 on Build GRCh38
rs994778770 Chr.19: 1111295-1111295 on Build GRCh38
rs1022628304 Chr.19: 1111184-1111184 on Build GRCh38
rs752762613 Chr.19: 1110983-1110983 on Build GRCh38
rs781295095 Chr.19: 1110981-1110981 on Build GRCh38
rs774145770 Chr.19: 1111109-1111109 on Build GRCh38
rs767490708 Chr.19: 1111048-1111048 on Build GRCh38
rs183596629 Chr.19: 1111235-1111235 on Build GRCh38
rs1005569542 Chr.19: 1111234-1111234 on Build GRCh38
rs985114457 Chr.19: 1111346-1111346 on Build GRCh38
rs774232176 Chr.19: 1111044-1111044 on Build GRCh38
rs960387623 Chr.19: 1111202-1111202 on Build GRCh38
rs779110376 Chr.19: 1111076-1111076 on Build GRCh38
rs752672743 Chr.19: 1111049-1111049 on Build GRCh38
rs766999204 Chr.19: 1111141-1111141 on Build GRCh38
rs377254047 Chr.19: 1111074-1111074 on Build GRCh38
rs772546100 Chr.19: 1111103-1111103 on Build GRCh38
rs947994444 Chr.19: 1111382-1111382 on Build GRCh38
rs373100652 Chr.19: 1111016-1111016 on Build GRCh38
rs775173408 Chr.19: 1111194-1111194 on Build GRCh38
rs770427346 Chr.19: 1111108-1111108 on Build GRCh38
rs889096272 Chr.19: 1110966-1110966 on Build GRCh38
rs766053622 Chr.19: 1110972-1110972 on Build GRCh38
rs146515706 Chr.19: 1110977-1110978 on Build GRCh38
rs1014251901 Chr.19: 1111187-1111187 on Build GRCh38
rs1027936814 Chr.19: 1110964-1110964 on Build GRCh38
rs749449592 Chr.19: 1110989-1110989 on Build GRCh38
rs746242934 Chr.19: 1111000-1111000 on Build GRCh38
rs376907651 Chr.19: 1111114-1111114 on Build GRCh38
rs576378029 Chr.19: 1111026-1111026 on Build GRCh38
rs1056347614 Chr.19: 1111161-1111161 on Build GRCh38
rs369211627 Chr.19: 1111041-1111041 on Build GRCh38
rs1022103707 Chr.19: 1111374-1111374 on Build GRCh38
rs905833242 Chr.19: 1111395-1111395 on Build GRCh38
rs557208951 Chr.19: 1111375-1111375 on Build GRCh38
rs935161893 Chr.19: 1111152-1111152 on Build GRCh38
rs1044973896 Chr.19: 1111385-1111385 on Build GRCh38
rs961862873 Chr.19: 1111105-1111105 on Build GRCh38
rs910956911 Chr.19: 1111126-1111126 on Build GRCh38
rs377201548 Chr.19: 1111029-1111029 on Build GRCh38
rs780708208 Chr.19: 1111005-1111006 on Build GRCh38
rs115495838 Chr.19: 1111120-1111120 on Build GRCh38
rs927620847 Chr.19: 1111195-1111195 on Build GRCh38
rs1029913695 Chr.19: 1111329-1111329 on Build GRCh38
rs541193469 Chr.19: 1111099-1111099 on Build GRCh38
rs981112717 Chr.19: 1111190-1111190 on Build GRCh38
rs763762719 Chr.19: 1111343-1111343 on Build GRCh38
rs756644958 Chr.19: 1110981-1110981 on Build GRCh38
rs866574071 Chr.19: 1111345-1111345 on Build GRCh38
rs967722281 Chr.19: 1111351-1111351 on Build GRCh38
rs923692235 Chr.19: 1111357-1111357 on Build GRCh38
rs1017927523 Chr.19: 1111350-1111350 on Build GRCh38
rs777967079 Chr.19: 1110988-1110988 on Build GRCh38
rs1056800556 Chr.19: 1111059-1111059 on Build GRCh38
rs961628908 Chr.19: 1111009-1111009 on Build GRCh38
rs191437815 Chr.19: 1110979-1110979 on Build GRCh38
rs370262573 Chr.19: 1111097-1111097 on Build GRCh38
rs762469840 Chr.19: 1111035-1111035 on Build GRCh38
rs748449429 Chr.19: 1110956-1110956 on Build GRCh38
rs539825120 Chr.19: 1110959-1110959 on Build GRCh38
rs573918859 Chr.19: 1111085-1111085 on Build GRCh38
rs772500510 Chr.19: 1111001-1111001 on Build GRCh38
rs567332623 Chr.19: 1111237-1111237 on Build GRCh38
rs897289164 Chr.19: 1111247-1111247 on Build GRCh38
rs943611678 Chr.19: 1111312-1111312 on Build GRCh38
rs758748563 Chr.19: 1111079-1111079 on Build GRCh38
rs61738770 Chr.19: 1111065-1111065 on Build GRCh38
rs956522746 Chr.19: 1111147-1111147 on Build GRCh38
rs769160417 Chr.19: 1111028-1111028 on Build GRCh38
rs367574811 Chr.19: 1111146-1111146 on Build GRCh38
rs545673330 Chr.19: 1111186-1111186 on Build GRCh38
rs528302781 Chr.19: 1111292-1111292 on Build GRCh38
rs896426810 Chr.19: 1111173-1111173 on Build GRCh38
rs879081981 Chr.19: 1111145-1111145 on Build GRCh38
rs1008234110 Chr.19: 1111127-1111127 on Build GRCh38
rs756148979 Chr.19: 1110987-1110987 on Build GRCh38
rs747601166 Chr.19: 1111125-1111126 on Build GRCh38
rs775466941 Chr.19: 1111129-1111129 on Build GRCh38
rs746154940 Chr.19: 1111077-1111077 on Build GRCh38
rs563170189 Chr.19: 1111221-1111221 on Build GRCh38
rs373256042 Chr.19: 1111055-1111055 on Build GRCh38
rs779109010 Chr.19: 1110992-1110992 on Build GRCh38
rs538598985 Chr.19: 1111373-1111373 on Build GRCh38
rs780446082 Chr.19: 1111092-1111092 on Build GRCh38
rs910714239 Chr.19: 1111290-1111290 on Build GRCh38
rs1030769059 Chr.19: 1110998-1110998 on Build GRCh38
rs376340506 Chr.19: 1111107-1111107 on Build GRCh38
rs756126247 Chr.19: 1111054-1111054 on Build GRCh38
rs919048035 Chr.19: 1111219-1111219 on Build GRCh38
rs998867957 Chr.19: 1111139-1111139 on Build GRCh38
rs147283234 Chr.19: 1111356-1111356 on Build GRCh38
rs1043506068 Chr.19: 1111133-1111133 on Build GRCh38
rs1044398770 Chr.19: 1111197-1111197 on Build GRCh38
rs571418875 Chr.19: 1111372-1111372 on Build GRCh38
rs941373319 Chr.19: 1111401-1111401 on Build GRCh38
rs1049458363 Chr.19: 1111034-1111034 on Build GRCh38
rs1053132161 Chr.19: 1111371-1111371 on Build GRCh38
rs901859351 Chr.19: 1111130-1111130 on Build GRCh38
rs369878903 Chr.19: 1110990-1110990 on Build GRCh38
rs759411667 Chr.19: 1111045-1111045 on Build GRCh38
rs28612115 Chr.19: 1111128-1111128 on Build GRCh38
rs905943550 Chr.19: 1111204-1111204 on Build GRCh38
rs569981925 Chr.19: 1111396-1111396 on Build GRCh38

View Full Product Details