Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 2427712-2428211 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LMNB2
Entrez Gene ID
84823
Gene Name
lamin B2
Gene Aliases
EPM9, LAMB2, LMN2
UniGene
Hs.538286
Gene Chromosome Location
Chr.19: 2428165-2456968 on Build GRCh38
Transcripts
NM_032737.3 NM_012458.3

Gene Symbol
TIMM13
Entrez Gene ID
26517
Gene Name
translocase of inner mitochondrial membrane 13
Gene Aliases
TIM13, TIM13B, TIMM13A, TIMM13B, ppv1
UniGene
Hs.75056
Gene Chromosome Location
Chr.19: 2425624-2427916 on Build GRCh38
Transcripts
NM_032737.3 NM_012458.3

SNP Detail

SNP ID Chromosome Location
rs186599801 Chr.19: 2428120-2428120 on Build GRCh38
rs1003521548 Chr.19: 2427997-2427998 on Build GRCh38
rs576621852 Chr.19: 2427931-2427931 on Build GRCh38
rs991251749 Chr.19: 2427829-2427829 on Build GRCh38
rs577509806 Chr.19: 2427816-2427816 on Build GRCh38
rs935548999 Chr.19: 2428170-2428170 on Build GRCh38
rs920451685 Chr.19: 2427917-2427917 on Build GRCh38
rs888848205 Chr.19: 2428002-2428002 on Build GRCh38
rs867356024 Chr.19: 2427858-2427858 on Build GRCh38
rs916604739 Chr.19: 2427830-2427830 on Build GRCh38
rs999798375 Chr.19: 2427967-2427967 on Build GRCh38
rs961833023 Chr.19: 2427915-2427915 on Build GRCh38
rs548397156 Chr.19: 2427759-2427759 on Build GRCh38
rs971481558 Chr.19: 2428158-2428158 on Build GRCh38
rs530064906 Chr.19: 2427857-2427857 on Build GRCh38
rs116048643 Chr.19: 2427981-2427981 on Build GRCh38
rs528481865 Chr.19: 2427913-2427913 on Build GRCh38
rs1022947177 Chr.19: 2428142-2428142 on Build GRCh38
rs1033342062 Chr.19: 2428096-2428096 on Build GRCh38
rs78272496 Chr.19: 2427943-2427943 on Build GRCh38
rs761786072 Chr.19: 2427847-2427847 on Build GRCh38
rs116680227 Chr.19: 2427892-2427892 on Build GRCh38
rs184169315 Chr.19: 2427811-2427811 on Build GRCh38
rs764984429 Chr.19: 2427853-2427853 on Build GRCh38
rs560098803 Chr.19: 2428113-2428113 on Build GRCh38
rs920360361 Chr.19: 2427861-2427861 on Build GRCh38
rs890010475 Chr.19: 2427784-2427784 on Build GRCh38
rs772842441 Chr.19: 2427867-2427867 on Build GRCh38
rs540799740 Chr.19: 2427801-2427801 on Build GRCh38
rs562744226 Chr.19: 2427839-2427839 on Build GRCh38
rs541849221 Chr.19: 2428105-2428105 on Build GRCh38
rs977430486 Chr.19: 2427810-2427810 on Build GRCh38
rs994599009 Chr.19: 2427898-2427898 on Build GRCh38
rs940570201 Chr.19: 2427855-2427855 on Build GRCh38
rs754369309 Chr.19: 2428087-2428087 on Build GRCh38
rs1024025506 Chr.19: 2427904-2427904 on Build GRCh38
rs920312513 Chr.19: 2427976-2427976 on Build GRCh38
rs957565023 Chr.19: 2427951-2427951 on Build GRCh38
rs531553313 Chr.19: 2427903-2427903 on Build GRCh38
rs981487739 Chr.19: 2428177-2428177 on Build GRCh38
rs114211152 Chr.19: 2427971-2427971 on Build GRCh38
rs367776733 Chr.19: 2427764-2427764 on Build GRCh38
rs961906105 Chr.19: 2428005-2428005 on Build GRCh38
rs4806847 Chr.19: 2427817-2427817 on Build GRCh38
rs386805856 Chr.19: 2427817-2427819 on Build GRCh38
rs977062436 Chr.19: 2428107-2428107 on Build GRCh38
rs147811020 Chr.19: 2428028-2428028 on Build GRCh38
rs527758462 Chr.19: 2427877-2427877 on Build GRCh38
rs893983498 Chr.19: 2427972-2427972 on Build GRCh38
rs11542678 Chr.19: 2427851-2427851 on Build GRCh38
rs182320329 Chr.19: 2427962-2427962 on Build GRCh38
rs908679422 Chr.19: 2427966-2427966 on Build GRCh38
rs957429658 Chr.19: 2428104-2428104 on Build GRCh38
rs898792584 Chr.19: 2427894-2427894 on Build GRCh38
rs966869427 Chr.19: 2427804-2427804 on Build GRCh38
rs374086519 Chr.19: 2427922-2427922 on Build GRCh38
rs1037398626 Chr.19: 2427859-2427859 on Build GRCh38
rs1008444297 Chr.19: 2427872-2427872 on Build GRCh38
rs967381929 Chr.19: 2427806-2427806 on Build GRCh38
rs986275411 Chr.19: 2428072-2428072 on Build GRCh38
rs1040196805 Chr.19: 2428034-2428034 on Build GRCh38
rs888982362 Chr.19: 2427909-2427909 on Build GRCh38
rs991629138 Chr.19: 2427954-2427954 on Build GRCh38
rs573488831 Chr.19: 2427766-2427766 on Build GRCh38
rs973350257 Chr.19: 2428016-2428016 on Build GRCh38
rs188753454 Chr.19: 2427914-2427914 on Build GRCh38
rs1014894596 Chr.19: 2427998-2427998 on Build GRCh38
rs1008495270 Chr.19: 2427797-2427797 on Build GRCh38
rs931688983 Chr.19: 2427927-2427927 on Build GRCh38
rs577323525 Chr.19: 2427968-2427968 on Build GRCh38
rs994966787 Chr.19: 2427742-2427742 on Build GRCh38
rs902674968 Chr.19: 2427876-2427876 on Build GRCh38
rs191529223 Chr.19: 2427798-2427798 on Build GRCh38
rs1054046512 Chr.19: 2428179-2428179 on Build GRCh38
rs931767103 Chr.19: 2427862-2427862 on Build GRCh38
rs367985895 Chr.19: 2427957-2427957 on Build GRCh38
rs535391246 Chr.19: 2427921-2427921 on Build GRCh38
rs531223091 Chr.19: 2427871-2427871 on Build GRCh38
rs1008672617 Chr.19: 2427910-2427910 on Build GRCh38
rs568043476 Chr.19: 2427731-2427731 on Build GRCh38
rs752964320 Chr.19: 2427987-2427987 on Build GRCh38
rs907677221 Chr.19: 2427850-2427850 on Build GRCh38
rs981994321 Chr.19: 2427912-2427912 on Build GRCh38
rs191577372 Chr.19: 2427960-2427960 on Build GRCh38
rs570303316 Chr.19: 2427901-2427901 on Build GRCh38
rs898930679 Chr.19: 2427741-2427741 on Build GRCh38
rs552637202 Chr.19: 2427874-2427874 on Build GRCh38
rs1028834115 Chr.19: 2427749-2427749 on Build GRCh38
rs568587950 Chr.19: 2428046-2428046 on Build GRCh38
rs567971729 Chr.19: 2427916-2427916 on Build GRCh38
rs541936745 Chr.19: 2427865-2427865 on Build GRCh38
rs944388704 Chr.19: 2427958-2427958 on Build GRCh38
rs991951012 Chr.19: 2427833-2427833 on Build GRCh38
rs999724091 Chr.19: 2427880-2427882 on Build GRCh38
rs144299347 Chr.19: 2427739-2427739 on Build GRCh38
rs756254732 Chr.19: 2428037-2428037 on Build GRCh38

View Full Product Details