Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3118817-3119319 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNA11
Entrez Gene ID
2767
Gene Name
G protein subunit alpha 11
Gene Aliases
FBH, FBH2, FHH2, GNA-11, HHC2, HYPOC2
UniGene
Hs.650575
Gene Chromosome Location
Chr.19: 3094410-3124002 on Build GRCh38
Transcripts
NM_002067.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6971905 c.767G>A p.R256Q Chr.19: 3119237-3119237 on Build GRCh38
338587 c.693C>A p.A231A Chr.19: 3119011-3119011 on Build GRCh38
6845094 c.625_626CA>GC p.Q209A Chr.19: 3118943-3118944 on Build GRCh38
52972 c.626_627AG>TT p.Q209L Chr.19: 3118944-3118945 on Build GRCh38
52970 c.626A>C p.Q209P Chr.19: 3118944-3118944 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754566491 Chr.19: 3119053-3119054 on Build GRCh38
rs757393272 Chr.19: 3118981-3118981 on Build GRCh38
rs191674809 Chr.19: 3118888-3118888 on Build GRCh38
rs587777022 Chr.19: 3118950-3118950 on Build GRCh38
rs775356005 Chr.19: 3119173-3119173 on Build GRCh38
rs562714666 Chr.19: 3118862-3118862 on Build GRCh38
rs747291840 Chr.19: 3119008-3119008 on Build GRCh38
rs749372899 Chr.19: 3118960-3118960 on Build GRCh38
rs778948783 Chr.19: 3118985-3118985 on Build GRCh38
rs771537617 Chr.19: 3119232-3119232 on Build GRCh38
rs149637752 Chr.19: 3119044-3119044 on Build GRCh38
rs780843711 Chr.19: 3118880-3118880 on Build GRCh38
rs529942770 Chr.19: 3119237-3119237 on Build GRCh38
rs751628153 Chr.19: 3119283-3119283 on Build GRCh38
rs770720811 Chr.19: 3119201-3119201 on Build GRCh38
rs746432688 Chr.19: 3118993-3118993 on Build GRCh38
rs878908474 Chr.19: 3118839-3118839 on Build GRCh38
rs752549242 Chr.19: 3119050-3119050 on Build GRCh38
rs369692834 Chr.19: 3119199-3119199 on Build GRCh38
rs376322895 Chr.19: 3118939-3118939 on Build GRCh38
rs779001946 Chr.19: 3119080-3119080 on Build GRCh38
rs761542744 Chr.19: 3119248-3119248 on Build GRCh38
rs367580054 Chr.19: 3119183-3119183 on Build GRCh38
rs780180140 Chr.19: 3119188-3119188 on Build GRCh38
rs776428312 Chr.19: 3119244-3119244 on Build GRCh38
rs767395295 Chr.19: 3118946-3118946 on Build GRCh38
rs966854111 Chr.19: 3119062-3119062 on Build GRCh38
rs748378708 Chr.19: 3118887-3118887 on Build GRCh38
rs11667202 Chr.19: 3119113-3119113 on Build GRCh38
rs144580097 Chr.19: 3118857-3118857 on Build GRCh38
rs983553501 Chr.19: 3119190-3119190 on Build GRCh38
rs558544590 Chr.19: 3119185-3119185 on Build GRCh38
rs747407950 Chr.19: 3119094-3119094 on Build GRCh38
rs371149978 Chr.19: 3118996-3118996 on Build GRCh38
rs769503200 Chr.19: 3119009-3119009 on Build GRCh38
rs763870839 Chr.19: 3119068-3119068 on Build GRCh38
rs773942927 Chr.19: 3119155-3119155 on Build GRCh38
rs187043784 Chr.19: 3119166-3119166 on Build GRCh38
rs138168314 Chr.19: 3119023-3119023 on Build GRCh38
rs751267772 Chr.19: 3118912-3118912 on Build GRCh38
rs926383500 Chr.19: 3119128-3119128 on Build GRCh38
rs536442833 Chr.19: 3119126-3119126 on Build GRCh38
rs539354791 Chr.19: 3119099-3119099 on Build GRCh38
rs374086915 Chr.19: 3119076-3119076 on Build GRCh38
rs763923944 Chr.19: 3119175-3119175 on Build GRCh38
rs144829732 Chr.19: 3119275-3119275 on Build GRCh38
rs555086739 Chr.19: 3119159-3119159 on Build GRCh38
rs4900 Chr.19: 3119241-3119241 on Build GRCh38
rs766426049 Chr.19: 3118925-3118925 on Build GRCh38
rs755520418 Chr.19: 3119195-3119195 on Build GRCh38
rs569436825 Chr.19: 3119104-3119104 on Build GRCh38
rs760030169 Chr.19: 3118908-3118908 on Build GRCh38
rs917913405 Chr.19: 3119138-3119138 on Build GRCh38
rs751616481 Chr.19: 3118936-3118936 on Build GRCh38
rs766046727 Chr.19: 3119036-3119036 on Build GRCh38
rs892390765 Chr.19: 3118901-3118901 on Build GRCh38
rs765454337 Chr.19: 3119259-3119259 on Build GRCh38
rs308046 Chr.19: 3119186-3119186 on Build GRCh38
rs920981485 Chr.19: 3118850-3118850 on Build GRCh38
rs775040372 Chr.19: 3119236-3119236 on Build GRCh38
rs367937642 Chr.19: 3119081-3119081 on Build GRCh38
rs758970195 Chr.19: 3119084-3119084 on Build GRCh38
rs771890434 Chr.19: 3119054-3119055 on Build GRCh38
rs377219320 Chr.19: 3119198-3119198 on Build GRCh38
rs200532051 Chr.19: 3118909-3118909 on Build GRCh38
rs745528917 Chr.19: 3119220-3119220 on Build GRCh38
rs1007967128 Chr.19: 3119042-3119042 on Build GRCh38
rs997103699 Chr.19: 3119230-3119230 on Build GRCh38
rs755796821 Chr.19: 3119063-3119063 on Build GRCh38
rs762692548 Chr.19: 3119032-3119032 on Build GRCh38
rs754114633 Chr.19: 3119069-3119069 on Build GRCh38
rs758912691 Chr.19: 3119162-3119162 on Build GRCh38
rs774771956 Chr.19: 3118906-3118906 on Build GRCh38
rs371052478 Chr.19: 3118918-3118918 on Build GRCh38
rs748460076 Chr.19: 3118883-3118883 on Build GRCh38
rs777740843 Chr.19: 3119093-3119110 on Build GRCh38
rs199749164 Chr.19: 3119005-3119005 on Build GRCh38
rs116848044 Chr.19: 3119200-3119200 on Build GRCh38
rs777912441 Chr.19: 3118955-3118955 on Build GRCh38
rs371624161 Chr.19: 3119090-3119090 on Build GRCh38
rs1000957441 Chr.19: 3119064-3119064 on Build GRCh38
rs374918092 Chr.19: 3119102-3119102 on Build GRCh38
rs544105544 Chr.19: 3119194-3119194 on Build GRCh38
rs375617124 Chr.19: 3118889-3118889 on Build GRCh38
rs544068043 Chr.19: 3118858-3118858 on Build GRCh38
rs778362669 Chr.19: 3119209-3119209 on Build GRCh38
rs753595497 Chr.19: 3119179-3119179 on Build GRCh38
rs750681091 Chr.19: 3119274-3119274 on Build GRCh38
rs773976709 Chr.19: 3119043-3119043 on Build GRCh38
rs374438586 Chr.19: 3118921-3118921 on Build GRCh38
rs756381542 Chr.19: 3118951-3118951 on Build GRCh38
rs767308128 Chr.19: 3119046-3119046 on Build GRCh38
rs757446459 Chr.19: 3119073-3119073 on Build GRCh38
rs1045239691 Chr.19: 3118882-3118883 on Build GRCh38
rs755573572 Chr.19: 3119286-3119286 on Build GRCh38
rs754620313 Chr.19: 3118984-3118984 on Build GRCh38
rs1046005669 Chr.19: 3118860-3118860 on Build GRCh38
rs764761442 Chr.19: 3118916-3118916 on Build GRCh38
rs771440706 Chr.19: 3118898-3118898 on Build GRCh38

View Full Product Details