Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3643107-3643598 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIP5K1C
Entrez Gene ID
23396
Gene Name
phosphatidylinositol-4-phosphate 5-kinase type 1 gamma
Gene Aliases
LCCS3, PIP5K-GAMMA, PIP5K1-gamma, PIP5Kgamma
UniGene
Hs.282177
Gene Chromosome Location
Chr.19: 3630181-3700492 on Build GRCh38
Transcripts
NM_001195733.1 NM_001300849.1 NM_012398.2 XM_011527845.1 XM_011527846.1 XM_011527848.1 XM_011527849.1 XM_011527850.2 XM_011527851.2 XM_011527853.2 XM_017026540.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3199286 c.1594T>C p.S532P Chr.19: 3643298-3643298 on Build GRCh38
6338508 c.1631C>T p.S544L Chr.19: 3643261-3643261 on Build GRCh38
7156225 c.1622C>T p.S541L Chr.19: 3643270-3643270 on Build GRCh38
1129886 c.1647C>T p.Y549Y Chr.19: 3643245-3643245 on Build GRCh38
7156224 c.1622C>T p.S541L Chr.19: 3643270-3643270 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758816584 Chr.19: 3643432-3643432 on Build GRCh38
rs538492995 Chr.19: 3643185-3643185 on Build GRCh38
rs940577822 Chr.19: 3643293-3643293 on Build GRCh38
rs548427661 Chr.19: 3643537-3643537 on Build GRCh38
rs764109368 Chr.19: 3643398-3643398 on Build GRCh38
rs1019794662 Chr.19: 3643455-3643455 on Build GRCh38
rs1004843759 Chr.19: 3643135-3643135 on Build GRCh38
rs35766010 Chr.19: 3643285-3643285 on Build GRCh38
rs1056911209 Chr.19: 3643473-3643473 on Build GRCh38
rs377110165 Chr.19: 3643225-3643225 on Build GRCh38
rs112567031 Chr.19: 3643385-3643385 on Build GRCh38
rs369900954 Chr.19: 3643204-3643204 on Build GRCh38
rs754320017 Chr.19: 3643340-3643340 on Build GRCh38
rs529683530 Chr.19: 3643551-3643551 on Build GRCh38
rs780859590 Chr.19: 3643270-3643270 on Build GRCh38
rs768300936 Chr.19: 3643233-3643233 on Build GRCh38
rs905546264 Chr.19: 3643413-3643413 on Build GRCh38
rs975569686 Chr.19: 3643483-3643483 on Build GRCh38
rs202235514 Chr.19: 3643212-3643212 on Build GRCh38
rs766300538 Chr.19: 3643202-3643225 on Build GRCh38
rs974872310 Chr.19: 3643379-3643379 on Build GRCh38
rs759703366 Chr.19: 3643225-3643226 on Build GRCh38
rs145403606 Chr.19: 3643291-3643291 on Build GRCh38
rs753927194 Chr.19: 3643201-3643202 on Build GRCh38
rs1030288285 Chr.19: 3643545-3643545 on Build GRCh38
rs757538262 Chr.19: 3643263-3643263 on Build GRCh38
rs752717133 Chr.19: 3643250-3643250 on Build GRCh38
rs566222648 Chr.19: 3643183-3643183 on Build GRCh38
rs114428049 Chr.19: 3643165-3643165 on Build GRCh38
rs1026129469 Chr.19: 3643182-3643205 on Build GRCh38
rs762902576 Chr.19: 3643279-3643279 on Build GRCh38
rs550176292 Chr.19: 3643140-3643140 on Build GRCh38
rs1018582785 Chr.19: 3643527-3643527 on Build GRCh38
rs867200093 Chr.19: 3643521-3643521 on Build GRCh38
rs772319622 Chr.19: 3643394-3643394 on Build GRCh38
rs1016292731 Chr.19: 3643362-3643362 on Build GRCh38
rs993914283 Chr.19: 3643488-3643488 on Build GRCh38
rs765473697 Chr.19: 3643249-3643250 on Build GRCh38
rs984233963 Chr.19: 3643512-3643512 on Build GRCh38
rs759715712 Chr.19: 3643248-3643248 on Build GRCh38
rs766509839 Chr.19: 3643218-3643218 on Build GRCh38
rs372407268 Chr.19: 3643411-3643411 on Build GRCh38
rs947694643 Chr.19: 3643410-3643410 on Build GRCh38
rs370703895 Chr.19: 3643222-3643222 on Build GRCh38
rs139195886 Chr.19: 3643244-3643244 on Build GRCh38
rs753028086 Chr.19: 3643259-3643260 on Build GRCh38
rs373904192 Chr.19: 3643237-3643237 on Build GRCh38
rs760640808 Chr.19: 3643318-3643318 on Build GRCh38
rs892229015 Chr.19: 3643564-3643564 on Build GRCh38
rs185851048 Chr.19: 3643228-3643228 on Build GRCh38
rs767588604 Chr.19: 3643249-3643249 on Build GRCh38
rs773003605 Chr.19: 3643274-3643274 on Build GRCh38
rs1033326322 Chr.19: 3643135-3643158 on Build GRCh38
rs917810183 Chr.19: 3643523-3643523 on Build GRCh38
rs80032856 Chr.19: 3643163-3643163 on Build GRCh38
rs781531508 Chr.19: 3643265-3643265 on Build GRCh38
rs913385946 Chr.19: 3643401-3643401 on Build GRCh38
rs750861310 Chr.19: 3643429-3643429 on Build GRCh38
rs538100028 Chr.19: 3643211-3643211 on Build GRCh38
rs189826987 Chr.19: 3643380-3643380 on Build GRCh38
rs770318100 Chr.19: 3643197-3643197 on Build GRCh38
rs376029185 Chr.19: 3643389-3643389 on Build GRCh38
rs1048435947 Chr.19: 3643134-3643134 on Build GRCh38
rs758748230 Chr.19: 3643422-3643423 on Build GRCh38
rs774067623 Chr.19: 3643280-3643280 on Build GRCh38
rs566882145 Chr.19: 3643159-3643159 on Build GRCh38
rs755031701 Chr.19: 3643221-3643221 on Build GRCh38
rs984426151 Chr.19: 3643343-3643343 on Build GRCh38
rs778214208 Chr.19: 3643227-3643227 on Build GRCh38
rs562043000 Chr.19: 3643544-3643544 on Build GRCh38
rs936964650 Chr.19: 3643459-3643459 on Build GRCh38
rs751680689 Chr.19: 3643220-3643220 on Build GRCh38
rs776702962 Chr.19: 3643404-3643404 on Build GRCh38
rs369536238 Chr.19: 3643230-3643230 on Build GRCh38
rs892082357 Chr.19: 3643198-3643198 on Build GRCh38
rs754747586 Chr.19: 3643374-3643374 on Build GRCh38
rs111450009 Chr.19: 3643533-3643533 on Build GRCh38
rs1053444736 Chr.19: 3643562-3643562 on Build GRCh38
rs200558261 Chr.19: 3643289-3643289 on Build GRCh38
rs964742701 Chr.19: 3643542-3643542 on Build GRCh38
rs367612171 Chr.19: 3643378-3643378 on Build GRCh38
rs950701930 Chr.19: 3643150-3643150 on Build GRCh38
rs750571696 Chr.19: 3643208-3643208 on Build GRCh38
rs759768552 Chr.19: 3643284-3643284 on Build GRCh38
rs79423827 Chr.19: 3643277-3643277 on Build GRCh38
rs771822119 Chr.19: 3643229-3643229 on Build GRCh38
rs776684308 Chr.19: 3643205-3643205 on Build GRCh38
rs201670126 Chr.19: 3643264-3643264 on Build GRCh38
rs768352126 Chr.19: 3643269-3643269 on Build GRCh38
rs773506250 Chr.19: 3643203-3643203 on Build GRCh38
rs956942611 Chr.19: 3643189-3643189 on Build GRCh38
rs773077124 Chr.19: 3643192-3643193 on Build GRCh38
rs1036306281 Chr.19: 3643353-3643353 on Build GRCh38
rs966021138 Chr.19: 3643471-3643471 on Build GRCh38
rs764709446 Chr.19: 3643322-3643322 on Build GRCh38
rs760802308 Chr.19: 3643196-3643197 on Build GRCh38
rs963071637 Chr.19: 3643144-3643167 on Build GRCh38
rs1005478665 Chr.19: 3643522-3643522 on Build GRCh38
rs769814850 Chr.19: 3643273-3643273 on Build GRCh38
rs752888493 Chr.19: 3643309-3643309 on Build GRCh38
rs771566116 Chr.19: 3643199-3643199 on Build GRCh38
rs371054884 Chr.19: 3643534-3643534 on Build GRCh38
rs1000550499 Chr.19: 3643556-3643557 on Build GRCh38
rs1042755132 Chr.19: 3643210-3643210 on Build GRCh38
rs988845125 Chr.19: 3643566-3643566 on Build GRCh38
rs781201102 Chr.19: 3643223-3643223 on Build GRCh38
rs973155570 Chr.19: 3643160-3643181 on Build GRCh38
rs750756928 Chr.19: 3643368-3643368 on Build GRCh38
rs757796989 Chr.19: 3643350-3643350 on Build GRCh38
rs761603377 Chr.19: 3643206-3643206 on Build GRCh38
rs377164357 Chr.19: 3643240-3643240 on Build GRCh38
rs117112524 Chr.19: 3643196-3643196 on Build GRCh38
rs370385321 Chr.19: 3643252-3643252 on Build GRCh38
rs895600622 Chr.19: 3643476-3643476 on Build GRCh38
rs776236359 Chr.19: 3643236-3643236 on Build GRCh38
rs921472203 Chr.19: 3643492-3643492 on Build GRCh38
rs143219762 Chr.19: 3643377-3643377 on Build GRCh38
rs748687530 Chr.19: 3643226-3643226 on Build GRCh38
rs896351050 Chr.19: 3643147-3643147 on Build GRCh38
rs754268073 Chr.19: 3643260-3643260 on Build GRCh38
rs779419757 Chr.19: 3643356-3643356 on Build GRCh38
rs111959359 Chr.19: 3643493-3643493 on Build GRCh38
rs765823385 Chr.19: 3643420-3643420 on Build GRCh38
rs756701195 Chr.19: 3643251-3643251 on Build GRCh38
rs115336798 Chr.19: 3643395-3643395 on Build GRCh38
rs558698273 Chr.19: 3643193-3643193 on Build GRCh38

View Full Product Details