Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3769406-3769935 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAX2
Entrez Gene ID
84839
Gene Name
retina and anterior neural fold homeobox 2
Gene Aliases
ARMD6, CORD11, QRX, RAXL1
UniGene
Hs.532691
Gene Chromosome Location
Chr.19: 3769089-3772221 on Build GRCh38
Transcripts
NM_001319074.1 NM_032753.3

SNP Detail

SNP ID Chromosome Location
rs1009656822 Chr.19: 3769802-3769802 on Build GRCh38
rs1017319851 Chr.19: 3769704-3769704 on Build GRCh38
rs887917665 Chr.19: 3769845-3769845 on Build GRCh38
rs1054691513 Chr.19: 3769460-3769460 on Build GRCh38
rs1007241070 Chr.19: 3769693-3769693 on Build GRCh38
rs1017267736 Chr.19: 3769851-3769851 on Build GRCh38
rs926699833 Chr.19: 3769806-3769806 on Build GRCh38
rs181573768 Chr.19: 3769712-3769712 on Build GRCh38
rs1020082058 Chr.19: 3769809-3769809 on Build GRCh38
rs979570262 Chr.19: 3769615-3769615 on Build GRCh38
rs1860322 Chr.19: 3769472-3769472 on Build GRCh38
rs781672239 Chr.19: 3769477-3769477 on Build GRCh38
rs917547 Chr.19: 3769901-3769901 on Build GRCh38
rs901586216 Chr.19: 3769870-3769870 on Build GRCh38
rs1045194428 Chr.19: 3769483-3769483 on Build GRCh38
rs143816706 Chr.19: 3769529-3769539 on Build GRCh38
rs945501026 Chr.19: 3769528-3769538 on Build GRCh38
rs1032491752 Chr.19: 3769626-3769626 on Build GRCh38
rs1056680876 Chr.19: 3769517-3769517 on Build GRCh38
rs886054367 Chr.19: 3769672-3769672 on Build GRCh38
rs988214081 Chr.19: 3769650-3769650 on Build GRCh38
rs956854433 Chr.19: 3769791-3769791 on Build GRCh38
rs929531306 Chr.19: 3769837-3769837 on Build GRCh38
rs896134547 Chr.19: 3769583-3769583 on Build GRCh38
rs940480224 Chr.19: 3769727-3769727 on Build GRCh38
rs1014572912 Chr.19: 3769568-3769568 on Build GRCh38
rs372452808 Chr.19: 3769711-3769711 on Build GRCh38
rs10405125 Chr.19: 3769719-3769719 on Build GRCh38
rs148990863 Chr.19: 3769748-3769748 on Build GRCh38
rs934470760 Chr.19: 3769453-3769453 on Build GRCh38
rs990041035 Chr.19: 3769903-3769903 on Build GRCh38
rs905167387 Chr.19: 3769491-3769491 on Build GRCh38
rs1032544180 Chr.19: 3769785-3769785 on Build GRCh38
rs965420416 Chr.19: 3769828-3769828 on Build GRCh38
rs773142284 Chr.19: 3769733-3769733 on Build GRCh38
rs898071508 Chr.19: 3769544-3769544 on Build GRCh38
rs917546 Chr.19: 3769836-3769836 on Build GRCh38
rs551864984 Chr.19: 3769844-3769844 on Build GRCh38
rs748630795 Chr.19: 3769703-3769703 on Build GRCh38
rs886054365 Chr.19: 3769476-3769476 on Build GRCh38
rs865777598 Chr.19: 3769507-3769507 on Build GRCh38
rs956803181 Chr.19: 3769644-3769644 on Build GRCh38
rs912603253 Chr.19: 3769662-3769662 on Build GRCh38
rs917545 Chr.19: 3769755-3769755 on Build GRCh38
rs948551826 Chr.19: 3769754-3769754 on Build GRCh38
rs571601214 Chr.19: 3769871-3769871 on Build GRCh38
rs924339003 Chr.19: 3769735-3769735 on Build GRCh38
rs939491559 Chr.19: 3769814-3769814 on Build GRCh38
rs1031739378 Chr.19: 3769899-3769899 on Build GRCh38
rs554924169 Chr.19: 3769452-3769452 on Build GRCh38
rs1003963778 Chr.19: 3769508-3769508 on Build GRCh38
rs1056861754 Chr.19: 3769816-3769816 on Build GRCh38
rs969593840 Chr.19: 3769604-3769604 on Build GRCh38
rs368356816 Chr.19: 3769567-3769567 on Build GRCh38
rs887600199 Chr.19: 3769726-3769726 on Build GRCh38
rs138512126 Chr.19: 3769897-3769900 on Build GRCh38
rs533160062 Chr.19: 3769586-3769586 on Build GRCh38
rs886054366 Chr.19: 3769649-3769649 on Build GRCh38
rs866962886 Chr.19: 3769647-3769647 on Build GRCh38
rs914387817 Chr.19: 3769751-3769751 on Build GRCh38
rs921814565 Chr.19: 3769717-3769717 on Build GRCh38
rs541141934 Chr.19: 3769475-3769475 on Build GRCh38
rs1007689446 Chr.19: 3769850-3769850 on Build GRCh38
rs956141720 Chr.19: 3769739-3769739 on Build GRCh38
rs143706741 Chr.19: 3769798-3769798 on Build GRCh38
rs543639717 Chr.19: 3769602-3769602 on Build GRCh38
rs778752074 Chr.19: 3769439-3769439 on Build GRCh38

View Full Product Details