Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3778851-3779397 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MATK
Entrez Gene ID
4145
Gene Name
megakaryocyte-associated tyrosine kinase
Gene Aliases
CHK, CTK, HHYLTK, HYL, HYLTK, Lsk
UniGene
Hs.631845
Gene Chromosome Location
Chr.19: 3777969-3801812 on Build GRCh38
Transcripts
NM_002378.3 NM_139354.2 NM_139355.2 XM_011528018.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1392571 c.1151G>A p.S384N Chr.19: 3779038-3779038 on Build GRCh38
5365499 c.1107C>T p.S369S Chr.19: 3779082-3779082 on Build GRCh38
995687 c.1165A>G p.K389E Chr.19: 3779024-3779024 on Build GRCh38
995691 c.995T>G p.F332C Chr.19: 3779384-3779384 on Build GRCh38
6298345 c.985C>A p.L329I Chr.19: 3779394-3779394 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs888869138 Chr.19: 3779273-3779273 on Build GRCh38
rs867618261 Chr.19: 3779283-3779283 on Build GRCh38
rs769576742 Chr.19: 3779138-3779138 on Build GRCh38
rs893992900 Chr.19: 3779058-3779058 on Build GRCh38
rs373232103 Chr.19: 3778945-3778945 on Build GRCh38
rs368995190 Chr.19: 3778995-3778995 on Build GRCh38
rs781009151 Chr.19: 3779234-3779234 on Build GRCh38
rs576393660 Chr.19: 3778887-3778887 on Build GRCh38
rs915372604 Chr.19: 3779262-3779262 on Build GRCh38
rs747902835 Chr.19: 3779135-3779135 on Build GRCh38
rs566235480 Chr.19: 3779313-3779313 on Build GRCh38
rs771130003 Chr.19: 3779333-3779333 on Build GRCh38
rs550252601 Chr.19: 3779151-3779151 on Build GRCh38
rs762659930 Chr.19: 3779201-3779201 on Build GRCh38
rs1041475245 Chr.19: 3779035-3779035 on Build GRCh38
rs769891104 Chr.19: 3779022-3779022 on Build GRCh38
rs1053755040 Chr.19: 3779194-3779194 on Build GRCh38
rs201794530 Chr.19: 3779329-3779329 on Build GRCh38
rs866581341 Chr.19: 3779225-3779225 on Build GRCh38
rs993788610 Chr.19: 3779314-3779314 on Build GRCh38
rs969037057 Chr.19: 3778933-3778933 on Build GRCh38
rs976759052 Chr.19: 3779224-3779224 on Build GRCh38
rs758003714 Chr.19: 3779107-3779107 on Build GRCh38
rs549288354 Chr.19: 3779287-3779287 on Build GRCh38
rs763036889 Chr.19: 3779050-3779050 on Build GRCh38
rs920442148 Chr.19: 3778983-3778983 on Build GRCh38
rs764503769 Chr.19: 3779191-3779191 on Build GRCh38
rs150394159 Chr.19: 3779127-3779127 on Build GRCh38
rs778545174 Chr.19: 3778991-3778991 on Build GRCh38
rs775883499 Chr.19: 3779341-3779341 on Build GRCh38
rs951861871 Chr.19: 3779186-3779186 on Build GRCh38
rs757000023 Chr.19: 3779085-3779085 on Build GRCh38
rs767064322 Chr.19: 3779175-3779175 on Build GRCh38
rs765479697 Chr.19: 3779091-3779091 on Build GRCh38
rs766400646 Chr.19: 3779053-3779053 on Build GRCh38
rs1010631824 Chr.19: 3778894-3778894 on Build GRCh38
rs1010579422 Chr.19: 3779060-3779060 on Build GRCh38
rs140717053 Chr.19: 3779013-3779013 on Build GRCh38
rs750131543 Chr.19: 3779193-3779193 on Build GRCh38
rs761203915 Chr.19: 3779344-3779344 on Build GRCh38
rs1023229897 Chr.19: 3778924-3778924 on Build GRCh38
rs946884714 Chr.19: 3779307-3779307 on Build GRCh38
rs764168366 Chr.19: 3778948-3778948 on Build GRCh38
rs556261757 Chr.19: 3779354-3779354 on Build GRCh38
rs1042010098 Chr.19: 3779244-3779244 on Build GRCh38
rs376170806 Chr.19: 3779350-3779350 on Build GRCh38
rs949529668 Chr.19: 3778927-3778927 on Build GRCh38
rs77048445 Chr.19: 3779162-3779162 on Build GRCh38
rs1029176359 Chr.19: 3779087-3779087 on Build GRCh38
rs779667508 Chr.19: 3779114-3779114 on Build GRCh38
rs200953484 Chr.19: 3779061-3779061 on Build GRCh38
rs562052367 Chr.19: 3779010-3779010 on Build GRCh38
rs766879117 Chr.19: 3778952-3778952 on Build GRCh38
rs760149262 Chr.19: 3779177-3779177 on Build GRCh38
rs755396835 Chr.19: 3778971-3778971 on Build GRCh38
rs1005970819 Chr.19: 3779295-3779295 on Build GRCh38
rs774392424 Chr.19: 3779156-3779156 on Build GRCh38
rs962326982 Chr.19: 3779141-3779141 on Build GRCh38
rs751551743 Chr.19: 3779125-3779125 on Build GRCh38
rs781095789 Chr.19: 3779128-3779128 on Build GRCh38
rs200915721 Chr.19: 3779363-3779363 on Build GRCh38
rs369916435 Chr.19: 3778942-3778942 on Build GRCh38
rs767974303 Chr.19: 3779057-3779057 on Build GRCh38
rs369331552 Chr.19: 3779352-3779352 on Build GRCh38
rs1003204076 Chr.19: 3779117-3779117 on Build GRCh38
rs748448154 Chr.19: 3779017-3779017 on Build GRCh38
rs774874162 Chr.19: 3779055-3779055 on Build GRCh38
rs775997521 Chr.19: 3779183-3779183 on Build GRCh38
rs988809378 Chr.19: 3779257-3779257 on Build GRCh38
rs112889670 Chr.19: 3779353-3779353 on Build GRCh38
rs751018172 Chr.19: 3779228-3779228 on Build GRCh38
rs756024860 Chr.19: 3779373-3779373 on Build GRCh38
rs541647241 Chr.19: 3778994-3778994 on Build GRCh38
rs980934276 Chr.19: 3779190-3779190 on Build GRCh38
rs200899340 Chr.19: 3779338-3779338 on Build GRCh38
rs371447841 Chr.19: 3779269-3779269 on Build GRCh38
rs767017298 Chr.19: 3779367-3779367 on Build GRCh38
rs373719206 Chr.19: 3779218-3779218 on Build GRCh38
rs935571040 Chr.19: 3779256-3779256 on Build GRCh38
rs918120089 Chr.19: 3779167-3779167 on Build GRCh38
rs749562797 Chr.19: 3779332-3779332 on Build GRCh38
rs751638171 Chr.19: 3778953-3778954 on Build GRCh38
rs962305370 Chr.19: 3778962-3778962 on Build GRCh38
rs772454720 Chr.19: 3779178-3779178 on Build GRCh38
rs201235893 Chr.19: 3779337-3779337 on Build GRCh38
rs1034941592 Chr.19: 3778953-3778953 on Build GRCh38
rs752009663 Chr.19: 3778967-3778967 on Build GRCh38
rs201270076 Chr.19: 3778982-3778982 on Build GRCh38
rs570048768 Chr.19: 3779236-3779236 on Build GRCh38
rs750037878 Chr.19: 3779100-3779100 on Build GRCh38
rs374812538 Chr.19: 3779184-3779184 on Build GRCh38
rs751169376 Chr.19: 3779362-3779362 on Build GRCh38
rs757341443 Chr.19: 3779207-3779208 on Build GRCh38
rs757913493 Chr.19: 3779008-3779008 on Build GRCh38
rs35660675 Chr.19: 3778961-3778962 on Build GRCh38
rs539042051 Chr.19: 3779340-3779340 on Build GRCh38
rs140632879 Chr.19: 3779147-3779147 on Build GRCh38
rs545500806 Chr.19: 3778907-3778907 on Build GRCh38
rs536087136 Chr.19: 3779255-3779255 on Build GRCh38
rs756839620 Chr.19: 3778986-3778986 on Build GRCh38
rs755006770 Chr.19: 3779229-3779229 on Build GRCh38
rs984212533 Chr.19: 3779001-3779001 on Build GRCh38
rs901738052 Chr.19: 3779254-3779254 on Build GRCh38
rs201459758 Chr.19: 3779056-3779056 on Build GRCh38
rs761968866 Chr.19: 3779029-3779029 on Build GRCh38
rs576230717 Chr.19: 3778985-3778985 on Build GRCh38
rs773358810 Chr.19: 3779043-3779043 on Build GRCh38
rs36072143 Chr.19: 3779016-3779016 on Build GRCh38
rs777517062 Chr.19: 3779331-3779331 on Build GRCh38

View Full Product Details