Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 4354914-4355336 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MPND
Entrez Gene ID
84954
Gene Name
MPN domain containing
Gene Aliases
-
UniGene
Hs.321689
Gene Chromosome Location
Chr.19: 4343527-4360086 on Build GRCh38
Transcripts
NM_001159846.2 NM_001300862.1 NM_032868.5 XM_006722926.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5474178 c.898C>A p.R300S Chr.19: 4355000-4355000 on Build GRCh38
7283124 c.967G>A p.A323T Chr.19: 4355144-4355144 on Build GRCh38
6714743 c.950G>A p.R317Q Chr.19: 4355127-4355127 on Build GRCh38
5643744 c.951G>A p.R317R Chr.19: 4355128-4355128 on Build GRCh38
7323184 c.960C>T p.L320L Chr.19: 4355137-4355137 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746692889 Chr.19: 4355095-4355095 on Build GRCh38
rs193099915 Chr.19: 4355046-4355046 on Build GRCh38
rs767315796 Chr.19: 4355062-4355069 on Build GRCh38
rs764642079 Chr.19: 4355002-4355002 on Build GRCh38
rs771022819 Chr.19: 4355157-4355157 on Build GRCh38
rs201982212 Chr.19: 4355137-4355137 on Build GRCh38
rs765205633 Chr.19: 4355055-4355055 on Build GRCh38
rs201494111 Chr.19: 4355164-4355164 on Build GRCh38
rs552048325 Chr.19: 4354971-4354971 on Build GRCh38
rs751211217 Chr.19: 4354992-4354993 on Build GRCh38
rs2041774 Chr.19: 4355206-4355206 on Build GRCh38
rs1056654621 Chr.19: 4355014-4355014 on Build GRCh38
rs779451087 Chr.19: 4355135-4355135 on Build GRCh38
rs369865886 Chr.19: 4355045-4355045 on Build GRCh38
rs771494799 Chr.19: 4355091-4355091 on Build GRCh38
rs750324415 Chr.19: 4355174-4355174 on Build GRCh38
rs961003552 Chr.19: 4355146-4355146 on Build GRCh38
rs764444037 Chr.19: 4355050-4355050 on Build GRCh38
rs768235781 Chr.19: 4355141-4355141 on Build GRCh38
rs368705731 Chr.19: 4355170-4355170 on Build GRCh38
rs770395380 Chr.19: 4355112-4355112 on Build GRCh38
rs574037276 Chr.19: 4355236-4355236 on Build GRCh38
rs376039143 Chr.19: 4355071-4355071 on Build GRCh38
rs781059163 Chr.19: 4354946-4354946 on Build GRCh38
rs560027818 Chr.19: 4355220-4355220 on Build GRCh38
rs777735361 Chr.19: 4355065-4355065 on Build GRCh38
rs751132283 Chr.19: 4355208-4355208 on Build GRCh38
rs892444729 Chr.19: 4355302-4355302 on Build GRCh38
rs779688687 Chr.19: 4355093-4355093 on Build GRCh38
rs2041773 Chr.19: 4355057-4355057 on Build GRCh38
rs998100120 Chr.19: 4355090-4355090 on Build GRCh38
rs2041772 Chr.19: 4355038-4355038 on Build GRCh38
rs770061864 Chr.19: 4355143-4355143 on Build GRCh38
rs762273208 Chr.19: 4355105-4355105 on Build GRCh38
rs998172913 Chr.19: 4355292-4355292 on Build GRCh38
rs373394568 Chr.19: 4355008-4355008 on Build GRCh38
rs370388121 Chr.19: 4355116-4355116 on Build GRCh38
rs773732214 Chr.19: 4355144-4355144 on Build GRCh38
rs761166529 Chr.19: 4355001-4355001 on Build GRCh38
rs758540139 Chr.19: 4355061-4355061 on Build GRCh38
rs376145951 Chr.19: 4355138-4355138 on Build GRCh38
rs756070455 Chr.19: 4355168-4355170 on Build GRCh38
rs766569307 Chr.19: 4355062-4355062 on Build GRCh38
rs760970461 Chr.19: 4355049-4355049 on Build GRCh38
rs750239081 Chr.19: 4355110-4355111 on Build GRCh38
rs759256590 Chr.19: 4354993-4354993 on Build GRCh38
rs745441665 Chr.19: 4355074-4355074 on Build GRCh38
rs764971420 Chr.19: 4355172-4355172 on Build GRCh38
rs555051709 Chr.19: 4355103-4355103 on Build GRCh38
rs899690474 Chr.19: 4355131-4355131 on Build GRCh38
rs370186418 Chr.19: 4355005-4355005 on Build GRCh38
rs755989874 Chr.19: 4355037-4355037 on Build GRCh38
rs748037243 Chr.19: 4355035-4355035 on Build GRCh38
rs751738605 Chr.19: 4355063-4355063 on Build GRCh38
rs377426285 Chr.19: 4355025-4355025 on Build GRCh38
rs773837814 Chr.19: 4354982-4354982 on Build GRCh38
rs200906141 Chr.19: 4355070-4355070 on Build GRCh38
rs766404757 Chr.19: 4355119-4355119 on Build GRCh38
rs757883930 Chr.19: 4355133-4355133 on Build GRCh38
rs572035450 Chr.19: 4355104-4355104 on Build GRCh38
rs748752148 Chr.19: 4354975-4354975 on Build GRCh38
rs376470885 Chr.19: 4354981-4354981 on Build GRCh38
rs534597197 Chr.19: 4355128-4355128 on Build GRCh38
rs751784350 Chr.19: 4355120-4355120 on Build GRCh38
rs528775556 Chr.19: 4354970-4354970 on Build GRCh38
rs980692136 Chr.19: 4354977-4354977 on Build GRCh38
rs900089881 Chr.19: 4355288-4355288 on Build GRCh38
rs2041775 Chr.19: 4355303-4355303 on Build GRCh38
rs763394875 Chr.19: 4355151-4355151 on Build GRCh38
rs778851855 Chr.19: 4355129-4355129 on Build GRCh38
rs370837318 Chr.19: 4355042-4355042 on Build GRCh38
rs939038782 Chr.19: 4355203-4355203 on Build GRCh38
rs374599119 Chr.19: 4355165-4355165 on Build GRCh38
rs774616006 Chr.19: 4355161-4355161 on Build GRCh38
rs757259049 Chr.19: 4355018-4355018 on Build GRCh38
rs771824110 Chr.19: 4355048-4355048 on Build GRCh38
rs779786883 Chr.19: 4355202-4355211 on Build GRCh38
rs1040359995 Chr.19: 4355280-4355280 on Build GRCh38
rs745478748 Chr.19: 4355047-4355047 on Build GRCh38
rs767751541 Chr.19: 4354997-4354997 on Build GRCh38
rs748009479 Chr.19: 4354947-4354947 on Build GRCh38
rs373830032 Chr.19: 4355132-4355132 on Build GRCh38
rs758692442 Chr.19: 4355028-4355028 on Build GRCh38
rs759888114 Chr.19: 4355123-4355123 on Build GRCh38
rs765329731 Chr.19: 4355021-4355021 on Build GRCh38
rs756894894 Chr.19: 4354996-4354997 on Build GRCh38
rs201096146 Chr.19: 4355000-4355000 on Build GRCh38
rs753752752 Chr.19: 4355127-4355127 on Build GRCh38
rs563771474 Chr.19: 4355234-4355234 on Build GRCh38
rs941504584 Chr.19: 4355274-4355274 on Build GRCh38
rs755936457 Chr.19: 4355064-4355064 on Build GRCh38
rs929020893 Chr.19: 4355232-4355232 on Build GRCh38
rs749234250 Chr.19: 4355218-4355218 on Build GRCh38
rs758312935 Chr.19: 4355175-4355175 on Build GRCh38
rs867283545 Chr.19: 4355297-4355297 on Build GRCh38
rs763951560 Chr.19: 4355126-4355126 on Build GRCh38
rs780815063 Chr.19: 4355032-4355032 on Build GRCh38
rs763713986 Chr.19: 4354991-4355001 on Build GRCh38
rs897577344 Chr.19: 4355011-4355011 on Build GRCh38
rs754607641 Chr.19: 4355211-4355211 on Build GRCh38
rs768626204 Chr.19: 4355237-4355237 on Build GRCh38

View Full Product Details