Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 4529693-4530194 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLIN5
Entrez Gene ID
440503
Gene Name
perilipin 5
Gene Aliases
LSDA5, LSDP5, MLDP, OXPAT
UniGene
Hs.131034
Gene Chromosome Location
Chr.19: 4522531-4535196 on Build GRCh38
Transcripts
NM_001013706.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6455478 c.282C>A p.C94* Chr.19: 4529841-4529841 on Build GRCh38
7078909 c.306G>A p.E102E Chr.19: 4529817-4529817 on Build GRCh38
5936510 c.339+7C>T p.? Chr.19: 4529777-4529777 on Build GRCh38
439763 c.276C>T p.L92L Chr.19: 4529847-4529847 on Build GRCh38
1393657 c.257-1G>A p.? Chr.19: 4529867-4529867 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1049936262 Chr.19: 4529876-4529876 on Build GRCh38
rs971007283 Chr.19: 4530145-4530145 on Build GRCh38
rs376776152 Chr.19: 4529778-4529778 on Build GRCh38
rs769058461 Chr.19: 4529916-4529916 on Build GRCh38
rs371042131 Chr.19: 4529772-4529772 on Build GRCh38
rs10411813 Chr.19: 4530056-4530056 on Build GRCh38
rs759106996 Chr.19: 4529886-4529886 on Build GRCh38
rs1044726017 Chr.19: 4529820-4529820 on Build GRCh38
rs532302541 Chr.19: 4529953-4529956 on Build GRCh38
rs984527589 Chr.19: 4529982-4529982 on Build GRCh38
rs991193031 Chr.19: 4529720-4529720 on Build GRCh38
rs552215933 Chr.19: 4529939-4529939 on Build GRCh38
rs1015576444 Chr.19: 4530160-4530160 on Build GRCh38
rs770513934 Chr.19: 4529871-4529872 on Build GRCh38
rs374858315 Chr.19: 4529804-4529804 on Build GRCh38
rs371073816 Chr.19: 4529784-4529784 on Build GRCh38
rs1053727113 Chr.19: 4530068-4530068 on Build GRCh38
rs1024781465 Chr.19: 4529944-4529944 on Build GRCh38
rs866773277 Chr.19: 4529981-4529981 on Build GRCh38
rs367964761 Chr.19: 4529790-4529790 on Build GRCh38
rs777943544 Chr.19: 4530121-4530122 on Build GRCh38
rs941715520 Chr.19: 4529991-4529991 on Build GRCh38
rs35866605 Chr.19: 4530027-4530027 on Build GRCh38
rs773084796 Chr.19: 4529879-4529879 on Build GRCh38
rs746313247 Chr.19: 4529905-4529905 on Build GRCh38
rs144265522 Chr.19: 4530002-4530002 on Build GRCh38
rs779392936 Chr.19: 4529846-4529846 on Build GRCh38
rs797007753 Chr.19: 4530147-4530147 on Build GRCh38
rs369680654 Chr.19: 4529883-4529883 on Build GRCh38
rs779951782 Chr.19: 4529733-4529734 on Build GRCh38
rs200836591 Chr.19: 4529795-4529795 on Build GRCh38
rs778272511 Chr.19: 4529776-4529776 on Build GRCh38
rs763719467 Chr.19: 4529897-4529897 on Build GRCh38
rs554502233 Chr.19: 4529715-4529715 on Build GRCh38
rs912745272 Chr.19: 4530020-4530020 on Build GRCh38
rs201340592 Chr.19: 4529785-4529785 on Build GRCh38
rs562411864 Chr.19: 4530164-4530164 on Build GRCh38
rs576365794 Chr.19: 4530099-4530099 on Build GRCh38
rs779932326 Chr.19: 4529911-4529911 on Build GRCh38
rs532215790 Chr.19: 4529922-4529922 on Build GRCh38
rs772026049 Chr.19: 4529857-4529857 on Build GRCh38
rs181454955 Chr.19: 4529777-4529777 on Build GRCh38
rs769716536 Chr.19: 4529878-4529878 on Build GRCh38
rs747269189 Chr.19: 4529914-4529914 on Build GRCh38
rs752077883 Chr.19: 4529874-4529875 on Build GRCh38
rs369228137 Chr.19: 4529927-4529927 on Build GRCh38
rs747161287 Chr.19: 4529867-4529867 on Build GRCh38
rs200136383 Chr.19: 4529791-4529791 on Build GRCh38
rs752876915 Chr.19: 4529992-4529993 on Build GRCh38
rs375375547 Chr.19: 4529904-4529904 on Build GRCh38
rs754898958 Chr.19: 4529910-4529910 on Build GRCh38
rs576886863 Chr.19: 4529773-4529773 on Build GRCh38
rs761512979 Chr.19: 4529727-4529727 on Build GRCh38
rs545860870 Chr.19: 4530091-4530091 on Build GRCh38
rs916789004 Chr.19: 4529887-4529887 on Build GRCh38
rs1012121639 Chr.19: 4530115-4530115 on Build GRCh38
rs534002384 Chr.19: 4530026-4530026 on Build GRCh38
rs965566270 Chr.19: 4529745-4529745 on Build GRCh38
rs540417796 Chr.19: 4529731-4529731 on Build GRCh38
rs200153211 Chr.19: 4529842-4529842 on Build GRCh38
rs574553019 Chr.19: 4529728-4529728 on Build GRCh38
rs953025967 Chr.19: 4529953-4529954 on Build GRCh38
rs983641017 Chr.19: 4529958-4529958 on Build GRCh38
rs190555079 Chr.19: 4529984-4529984 on Build GRCh38
rs373225131 Chr.19: 4530128-4530128 on Build GRCh38
rs917066393 Chr.19: 4530012-4530012 on Build GRCh38
rs746004625 Chr.19: 4529847-4529847 on Build GRCh38
rs902492174 Chr.19: 4530041-4530041 on Build GRCh38
rs866063434 Chr.19: 4530102-4530102 on Build GRCh38
rs542196515 Chr.19: 4530134-4530134 on Build GRCh38
rs752240334 Chr.19: 4529829-4529829 on Build GRCh38
rs1008202703 Chr.19: 4529764-4529764 on Build GRCh38
rs758137364 Chr.19: 4530126-4530126 on Build GRCh38
rs776962537 Chr.19: 4529918-4529918 on Build GRCh38
rs766458874 Chr.19: 4529882-4529882 on Build GRCh38
rs775805572 Chr.19: 4529866-4529866 on Build GRCh38
rs752395924 Chr.19: 4529889-4529889 on Build GRCh38
rs185753001 Chr.19: 4529821-4529821 on Build GRCh38
rs116267473 Chr.19: 4530018-4530018 on Build GRCh38
rs367567613 Chr.19: 4529817-4529817 on Build GRCh38
rs757590276 Chr.19: 4529898-4529898 on Build GRCh38
rs992927264 Chr.19: 4530008-4530008 on Build GRCh38
rs949151228 Chr.19: 4529786-4529786 on Build GRCh38
rs779671791 Chr.19: 4529737-4529737 on Build GRCh38
rs908547410 Chr.19: 4529986-4529986 on Build GRCh38
rs201818128 Chr.19: 4529844-4529844 on Build GRCh38
rs562517824 Chr.19: 4529779-4529779 on Build GRCh38
rs961390199 Chr.19: 4530167-4530167 on Build GRCh38
rs1005810987 Chr.19: 4530143-4530143 on Build GRCh38
rs1008577544 Chr.19: 4530079-4530079 on Build GRCh38
rs758005899 Chr.19: 4529729-4529729 on Build GRCh38
rs372793168 Chr.19: 4529845-4529845 on Build GRCh38
rs891474662 Chr.19: 4530059-4530059 on Build GRCh38
rs753785620 Chr.19: 4529748-4529748 on Build GRCh38
rs895041712 Chr.19: 4530088-4530088 on Build GRCh38
rs958480058 Chr.19: 4529716-4529716 on Build GRCh38
rs990584522 Chr.19: 4529936-4529936 on Build GRCh38
rs1000784970 Chr.19: 4529926-4529926 on Build GRCh38
rs202046404 Chr.19: 4529757-4529757 on Build GRCh38
rs920148342 Chr.19: 4530007-4530007 on Build GRCh38
rs753728593 Chr.19: 4529721-4529721 on Build GRCh38
rs566572915 Chr.19: 4529888-4529888 on Build GRCh38
rs1039066379 Chr.19: 4529809-4529809 on Build GRCh38
rs970464013 Chr.19: 4529955-4529955 on Build GRCh38
rs376661482 Chr.19: 4529881-4529881 on Build GRCh38
rs1024728828 Chr.19: 4530123-4530123 on Build GRCh38
rs62115195 Chr.19: 4529863-4529863 on Build GRCh38
rs10403749 Chr.19: 4530013-4530013 on Build GRCh38
rs372388117 Chr.19: 4529896-4529896 on Build GRCh38
rs751262473 Chr.19: 4529746-4529746 on Build GRCh38
rs959185901 Chr.19: 4529933-4529933 on Build GRCh38
rs758538661 Chr.19: 4529906-4529906 on Build GRCh38
rs556175122 Chr.19: 4530094-4530094 on Build GRCh38

View Full Product Details