Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 4816638-4817139 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TICAM1
Entrez Gene ID
148022
Gene Name
toll like receptor adaptor molecule 1
Gene Aliases
IIAE6, MyD88-3, PRVTIRB, TICAM-1, TRIF
UniGene
Hs.29344
Gene Chromosome Location
Chr.19: 4815924-4831742 on Build GRCh38
Transcripts
NM_182919.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6230119 c.1591C>T p.H531Y Chr.19: 4816787-4816787 on Build GRCh38
140821 c.1582T>C p.F528L Chr.19: 4816796-4816796 on Build GRCh38
1641199 c.1350T>C p.A450A Chr.19: 4817028-4817028 on Build GRCh38
439874 c.1381G>C p.D461H Chr.19: 4816997-4816997 on Build GRCh38
5128621 c.1590C>A p.P530P Chr.19: 4816788-4816788 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377509189 Chr.19: 4816762-4816762 on Build GRCh38
rs777207773 Chr.19: 4816866-4816866 on Build GRCh38
rs142171170 Chr.19: 4816778-4816778 on Build GRCh38
rs912548534 Chr.19: 4816896-4816896 on Build GRCh38
rs777151236 Chr.19: 4817010-4817010 on Build GRCh38
rs763408253 Chr.19: 4816716-4816716 on Build GRCh38
rs758048210 Chr.19: 4817111-4817111 on Build GRCh38
rs753309282 Chr.19: 4816947-4816947 on Build GRCh38
rs770034965 Chr.19: 4817023-4817023 on Build GRCh38
rs139939816 Chr.19: 4816700-4816700 on Build GRCh38
rs779864319 Chr.19: 4816686-4816686 on Build GRCh38
rs763237315 Chr.19: 4816937-4816937 on Build GRCh38
rs376286418 Chr.19: 4816763-4816763 on Build GRCh38
rs752723765 Chr.19: 4816854-4816854 on Build GRCh38
rs748211950 Chr.19: 4817021-4817021 on Build GRCh38
rs754776028 Chr.19: 4816850-4816850 on Build GRCh38
rs912607337 Chr.19: 4816998-4816998 on Build GRCh38
rs756715938 Chr.19: 4816950-4816950 on Build GRCh38
rs775159579 Chr.19: 4817073-4817073 on Build GRCh38
rs769503444 Chr.19: 4816712-4816712 on Build GRCh38
rs780460725 Chr.19: 4816968-4816968 on Build GRCh38
rs767884569 Chr.19: 4816995-4817001 on Build GRCh38
rs995707106 Chr.19: 4817102-4817102 on Build GRCh38
rs543684306 Chr.19: 4816838-4816838 on Build GRCh38
rs150740529 Chr.19: 4816817-4816817 on Build GRCh38
rs925405032 Chr.19: 4817058-4817058 on Build GRCh38
rs538901003 Chr.19: 4816869-4816869 on Build GRCh38
rs750634396 Chr.19: 4816936-4816936 on Build GRCh38
rs748943819 Chr.19: 4816812-4816812 on Build GRCh38
rs2292151 Chr.19: 4816707-4816707 on Build GRCh38
rs772590478 Chr.19: 4816822-4816822 on Build GRCh38
rs966777780 Chr.19: 4817016-4817016 on Build GRCh38
rs550630185 Chr.19: 4816713-4816713 on Build GRCh38
rs768703212 Chr.19: 4816706-4816706 on Build GRCh38
rs772749430 Chr.19: 4816797-4816797 on Build GRCh38
rs373671679 Chr.19: 4816750-4816750 on Build GRCh38
rs199751361 Chr.19: 4816924-4816924 on Build GRCh38
rs756098119 Chr.19: 4816741-4816741 on Build GRCh38
rs766695629 Chr.19: 4816728-4816728 on Build GRCh38
rs774037288 Chr.19: 4816739-4816739 on Build GRCh38
rs781384413 Chr.19: 4817115-4817115 on Build GRCh38
rs753854196 Chr.19: 4817091-4817091 on Build GRCh38
rs770119168 Chr.19: 4817071-4817071 on Build GRCh38
rs781222011 Chr.19: 4816779-4816779 on Build GRCh38
rs772462289 Chr.19: 4816890-4816890 on Build GRCh38
rs143679494 Chr.19: 4816676-4816676 on Build GRCh38
rs756058204 Chr.19: 4816860-4816860 on Build GRCh38
rs754043860 Chr.19: 4816761-4816761 on Build GRCh38
rs143873813 Chr.19: 4817093-4817093 on Build GRCh38
rs1052560323 Chr.19: 4816892-4816892 on Build GRCh38
rs777706412 Chr.19: 4816944-4816944 on Build GRCh38
rs776581496 Chr.19: 4816928-4816928 on Build GRCh38
rs146110283 Chr.19: 4816730-4816730 on Build GRCh38
rs747684301 Chr.19: 4816782-4816782 on Build GRCh38
rs763968688 Chr.19: 4817081-4817081 on Build GRCh38
rs766706840 Chr.19: 4817069-4817069 on Build GRCh38
rs765491561 Chr.19: 4816844-4816844 on Build GRCh38
rs773404292 Chr.19: 4817046-4817046 on Build GRCh38
rs779700744 Chr.19: 4817112-4817112 on Build GRCh38
rs765583204 Chr.19: 4816932-4816932 on Build GRCh38
rs370630160 Chr.19: 4816818-4816818 on Build GRCh38
rs139114216 Chr.19: 4816872-4816872 on Build GRCh38
rs762011679 Chr.19: 4816843-4816843 on Build GRCh38
rs761898820 Chr.19: 4816931-4816931 on Build GRCh38
rs769147928 Chr.19: 4816791-4816791 on Build GRCh38
rs775806785 Chr.19: 4816893-4816893 on Build GRCh38
rs201878837 Chr.19: 4816959-4816959 on Build GRCh38
rs1012551976 Chr.19: 4816781-4816781 on Build GRCh38
rs374044969 Chr.19: 4816956-4816956 on Build GRCh38
rs143066432 Chr.19: 4816709-4816709 on Build GRCh38
rs757160857 Chr.19: 4817094-4817094 on Build GRCh38
rs986785530 Chr.19: 4816973-4816973 on Build GRCh38
rs764782405 Chr.19: 4817103-4817103 on Build GRCh38
rs771696411 Chr.19: 4816964-4816964 on Build GRCh38
rs767470486 Chr.19: 4816941-4816941 on Build GRCh38
rs746799783 Chr.19: 4816773-4816773 on Build GRCh38
rs140568066 Chr.19: 4817114-4817114 on Build GRCh38
rs769060173 Chr.19: 4817002-4817002 on Build GRCh38
rs747334545 Chr.19: 4816977-4816977 on Build GRCh38
rs779772992 Chr.19: 4816771-4816771 on Build GRCh38
rs749928559 Chr.19: 4817108-4817108 on Build GRCh38
rs775931722 Chr.19: 4816830-4816830 on Build GRCh38
rs752524876 Chr.19: 4816942-4816942 on Build GRCh38
rs544182648 Chr.19: 4817066-4817066 on Build GRCh38
rs746263278 Chr.19: 4816874-4816874 on Build GRCh38
rs769204651 Chr.19: 4816927-4816927 on Build GRCh38
rs753938233 Chr.19: 4816842-4816842 on Build GRCh38
rs567288271 Chr.19: 4816738-4816738 on Build GRCh38
rs144531955 Chr.19: 4817118-4817118 on Build GRCh38

View Full Product Details