Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 5039535-5040022 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDM4B
Entrez Gene ID
23030
Gene Name
lysine demethylase 4B
Gene Aliases
JMJD2B, TDRD14B
UniGene
Hs.654816
Gene Chromosome Location
Chr.19: 4969112-5153603 on Build GRCh38
Transcripts
NM_015015.2 XM_005259521.3 XM_011527814.2 XM_011527818.2 XM_011527819.1 XM_011527820.1 XM_011527821.1 XM_011527822.1 XM_017026503.1 XM_017026504.1 XM_017026505.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
999500 c.153G>A p.P51P Chr.19: 5039847-5039847 on Build GRCh38
4694961 c.177G>A p.T59T Chr.19: 5039871-5039871 on Build GRCh38
6585358 c.246C>A p.L82L Chr.19: 5039940-5039940 on Build GRCh38
5162544 c.150C>T p.P50P Chr.19: 5039844-5039844 on Build GRCh38
6245581 c.223G>T p.V75L Chr.19: 5039917-5039917 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs948156411 Chr.19: 5039610-5039610 on Build GRCh38
rs376710315 Chr.19: 5039798-5039798 on Build GRCh38
rs532861979 Chr.19: 5039660-5039660 on Build GRCh38
rs753456068 Chr.19: 5039886-5039886 on Build GRCh38
rs1046882882 Chr.19: 5039739-5039739 on Build GRCh38
rs773015575 Chr.19: 5039940-5039940 on Build GRCh38
rs1048109936 Chr.19: 5039710-5039710 on Build GRCh38
rs572673151 Chr.19: 5039887-5039887 on Build GRCh38
rs746110307 Chr.19: 5039903-5039903 on Build GRCh38
rs928172734 Chr.19: 5039633-5039633 on Build GRCh38
rs141838567 Chr.19: 5039889-5039889 on Build GRCh38
rs376663467 Chr.19: 5039946-5039946 on Build GRCh38
rs1055947075 Chr.19: 5039755-5039755 on Build GRCh38
rs973990819 Chr.19: 5039717-5039717 on Build GRCh38
rs1030945422 Chr.19: 5039797-5039797 on Build GRCh38
rs772790273 Chr.19: 5039833-5039833 on Build GRCh38
rs1012538153 Chr.19: 5039628-5039628 on Build GRCh38
rs768416962 Chr.19: 5039818-5039818 on Build GRCh38
rs765832707 Chr.19: 5039945-5039945 on Build GRCh38
rs373494253 Chr.19: 5040002-5040002 on Build GRCh38
rs906978916 Chr.19: 5039585-5039586 on Build GRCh38
rs757254378 Chr.19: 5039890-5039890 on Build GRCh38
rs911266005 Chr.19: 5039704-5039704 on Build GRCh38
rs552876774 Chr.19: 5039719-5039719 on Build GRCh38
rs751238075 Chr.19: 5039862-5039862 on Build GRCh38
rs139663878 Chr.19: 5039880-5039880 on Build GRCh38
rs1002997915 Chr.19: 5039586-5039586 on Build GRCh38
rs965279324 Chr.19: 5039742-5039742 on Build GRCh38
rs986928763 Chr.19: 5039991-5039991 on Build GRCh38
rs34116402 Chr.19: 5039697-5039698 on Build GRCh38
rs144724831 Chr.19: 5039871-5039871 on Build GRCh38
rs928022517 Chr.19: 5039893-5039893 on Build GRCh38
rs779243475 Chr.19: 5039901-5039901 on Build GRCh38
rs760502389 Chr.19: 5039806-5039806 on Build GRCh38
rs372914632 Chr.19: 5039925-5039925 on Build GRCh38
rs929871062 Chr.19: 5039738-5039738 on Build GRCh38
rs754632930 Chr.19: 5039869-5039869 on Build GRCh38
rs560442512 Chr.19: 5039651-5039651 on Build GRCh38
rs888074266 Chr.19: 5039714-5039714 on Build GRCh38
rs867966482 Chr.19: 5039562-5039562 on Build GRCh38
rs547214964 Chr.19: 5039638-5039638 on Build GRCh38
rs776413964 Chr.19: 5039910-5039910 on Build GRCh38
rs376802800 Chr.19: 5039760-5039760 on Build GRCh38
rs989426492 Chr.19: 5039563-5039563 on Build GRCh38
rs915211951 Chr.19: 5039600-5039600 on Build GRCh38
rs1046852551 Chr.19: 5039574-5039574 on Build GRCh38
rs968234660 Chr.19: 5039645-5039645 on Build GRCh38
rs529751180 Chr.19: 5039624-5039624 on Build GRCh38
rs955493207 Chr.19: 5039690-5039691 on Build GRCh38
rs907009994 Chr.19: 5039741-5039741 on Build GRCh38
rs761769525 Chr.19: 5039830-5039830 on Build GRCh38
rs369063729 Chr.19: 5039993-5039993 on Build GRCh38
rs919410120 Chr.19: 5039992-5039992 on Build GRCh38
rs373875176 Chr.19: 5039723-5039723 on Build GRCh38
rs368253544 Chr.19: 5039724-5039724 on Build GRCh38
rs759202602 Chr.19: 5040006-5040006 on Build GRCh38
rs995071728 Chr.19: 5039703-5039703 on Build GRCh38
rs561065507 Chr.19: 5039590-5039590 on Build GRCh38
rs766119212 Chr.19: 5039853-5039853 on Build GRCh38
rs368842866 Chr.19: 5039847-5039847 on Build GRCh38
rs768027092 Chr.19: 5039870-5039870 on Build GRCh38
rs915242955 Chr.19: 5039883-5039883 on Build GRCh38
rs538559552 Chr.19: 5039904-5039904 on Build GRCh38
rs898060115 Chr.19: 5039674-5039674 on Build GRCh38
rs987283903 Chr.19: 5039700-5039700 on Build GRCh38
rs762560855 Chr.19: 5039941-5039941 on Build GRCh38
rs111503637 Chr.19: 5039658-5039658 on Build GRCh38
rs765013799 Chr.19: 5039832-5039832 on Build GRCh38
rs546684476 Chr.19: 5039967-5039967 on Build GRCh38
rs776360072 Chr.19: 5039829-5039829 on Build GRCh38
rs747877649 Chr.19: 5039924-5039924 on Build GRCh38
rs1011643177 Chr.19: 5039811-5039811 on Build GRCh38
rs746026529 Chr.19: 5039557-5039557 on Build GRCh38
rs151023792 Chr.19: 5039753-5039753 on Build GRCh38

View Full Product Details