Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 5641660-5642158 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SAFB
Entrez Gene ID
6294
Gene Name
scaffold attachment factor B
Gene Aliases
HAP, HET, SAB-B1, SAF-B, SAF-B1, SAFB1
UniGene
Hs.728802
Gene Chromosome Location
Chr.19: 5623035-5668478 on Build GRCh38
Transcripts
NM_001201338.1 NM_001201339.1 NM_001201340.1 NM_001320571.1 NM_001320572.1 NM_002967.3 XM_017027114.1 XM_017027115.1 XM_017027116.1 XM_017027117.1 XM_017027118.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
212863 c.406G>A p.E136K Chr.19: 5641806-5641806 on Build GRCh38
1481558 c.483G>A p.S161S Chr.19: 5641883-5641883 on Build GRCh38
4723445 c.445G>A p.E149K Chr.19: 5641845-5641845 on Build GRCh38
352771 c.361G>T p.E121* Chr.19: 5641761-5641761 on Build GRCh38
1494429 c.458C>T p.A153V Chr.19: 5641858-5641858 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745917784 Chr.19: 5641769-5641769 on Build GRCh38
rs201829821 Chr.19: 5641705-5641705 on Build GRCh38
rs777726893 Chr.19: 5641955-5641955 on Build GRCh38
rs772227517 Chr.19: 5641874-5641874 on Build GRCh38
rs182749782 Chr.19: 5641851-5641851 on Build GRCh38
rs763539987 Chr.19: 5641993-5641993 on Build GRCh38
rs762714662 Chr.19: 5641790-5641790 on Build GRCh38
rs780293472 Chr.19: 5641876-5641876 on Build GRCh38
rs754338398 Chr.19: 5641912-5641912 on Build GRCh38
rs747097630 Chr.19: 5641936-5641936 on Build GRCh38
rs372814398 Chr.19: 5641868-5641868 on Build GRCh38
rs758890651 Chr.19: 5642031-5642031 on Build GRCh38
rs765718923 Chr.19: 5642124-5642124 on Build GRCh38
rs369929220 Chr.19: 5641712-5641712 on Build GRCh38
rs187821467 Chr.19: 5641990-5641990 on Build GRCh38
rs139125544 Chr.19: 5641686-5641686 on Build GRCh38
rs750812289 Chr.19: 5642030-5642030 on Build GRCh38
rs745473423 Chr.19: 5641844-5641844 on Build GRCh38
rs766678219 Chr.19: 5642066-5642066 on Build GRCh38
rs774139180 Chr.19: 5641962-5641962 on Build GRCh38
rs765629319 Chr.19: 5642024-5642024 on Build GRCh38
rs759396649 Chr.19: 5641970-5641970 on Build GRCh38
rs1002629130 Chr.19: 5642022-5642022 on Build GRCh38
rs764882015 Chr.19: 5641778-5641778 on Build GRCh38
rs899902072 Chr.19: 5642104-5642104 on Build GRCh38
rs1008305083 Chr.19: 5641731-5641731 on Build GRCh38
rs996829021 Chr.19: 5641788-5641788 on Build GRCh38
rs755694216 Chr.19: 5641856-5641856 on Build GRCh38
rs759202406 Chr.19: 5641904-5641904 on Build GRCh38
rs200741247 Chr.19: 5641765-5641765 on Build GRCh38
rs60436199 Chr.19: 5641836-5641836 on Build GRCh38
rs754213071 Chr.19: 5641845-5641845 on Build GRCh38
rs61760907 Chr.19: 5641758-5641758 on Build GRCh38
rs771702768 Chr.19: 5641985-5641985 on Build GRCh38
rs760164733 Chr.19: 5641906-5641906 on Build GRCh38
rs749406826 Chr.19: 5641750-5641750 on Build GRCh38
rs370566877 Chr.19: 5641816-5641816 on Build GRCh38
rs377193879 Chr.19: 5641701-5641701 on Build GRCh38
rs750613942 Chr.19: 5641924-5641924 on Build GRCh38
rs892300965 Chr.19: 5642071-5642071 on Build GRCh38
rs751056693 Chr.19: 5641708-5641708 on Build GRCh38
rs191428945 Chr.19: 5641707-5641707 on Build GRCh38
rs766033591 Chr.19: 5641799-5641799 on Build GRCh38
rs779440043 Chr.19: 5641920-5641920 on Build GRCh38
rs757607710 Chr.19: 5641850-5641850 on Build GRCh38
rs749207345 Chr.19: 5641957-5641957 on Build GRCh38
rs755037537 Chr.19: 5642076-5642076 on Build GRCh38
rs377234567 Chr.19: 5641961-5641961 on Build GRCh38
rs924926328 Chr.19: 5641976-5641976 on Build GRCh38
rs765110177 Chr.19: 5641700-5641700 on Build GRCh38
rs777145058 Chr.19: 5641997-5641997 on Build GRCh38
rs761831306 Chr.19: 5641776-5641776 on Build GRCh38
rs931049111 Chr.19: 5641784-5641784 on Build GRCh38
rs757598112 Chr.19: 5641913-5641913 on Build GRCh38
rs373038496 Chr.19: 5641721-5641721 on Build GRCh38
rs538698847 Chr.19: 5641773-5641773 on Build GRCh38
rs747968577 Chr.19: 5641883-5641883 on Build GRCh38
rs751761434 Chr.19: 5642068-5642068 on Build GRCh38
rs753148563 Chr.19: 5641830-5641830 on Build GRCh38
rs775032378 Chr.19: 5642082-5642082 on Build GRCh38
rs137931208 Chr.19: 5641829-5641829 on Build GRCh38
rs764636753 Chr.19: 5641911-5641911 on Build GRCh38
rs911417623 Chr.19: 5641882-5641882 on Build GRCh38
rs193134354 Chr.19: 5642122-5642122 on Build GRCh38
rs773063351 Chr.19: 5641779-5641779 on Build GRCh38
rs972333585 Chr.19: 5641728-5641728 on Build GRCh38
rs781146576 Chr.19: 5642079-5642079 on Build GRCh38
rs762779310 Chr.19: 5641901-5641901 on Build GRCh38
rs772315227 Chr.19: 5641770-5641770 on Build GRCh38
rs770745797 Chr.19: 5641902-5641902 on Build GRCh38
rs775049286 Chr.19: 5641987-5641987 on Build GRCh38
rs778216236 Chr.19: 5641747-5641747 on Build GRCh38
rs570059868 Chr.19: 5641697-5641697 on Build GRCh38
rs760244715 Chr.19: 5642096-5642096 on Build GRCh38
rs141699045 Chr.19: 5641886-5641886 on Build GRCh38
rs749186109 Chr.19: 5642084-5642084 on Build GRCh38
rs776755838 Chr.19: 5641693-5641693 on Build GRCh38
rs752202368 Chr.19: 5641905-5641905 on Build GRCh38
rs374187283 Chr.19: 5641689-5641689 on Build GRCh38
rs1045396938 Chr.19: 5641692-5641692 on Build GRCh38
rs537832211 Chr.19: 5642009-5642009 on Build GRCh38
rs770862829 Chr.19: 5641685-5641685 on Build GRCh38
rs905731143 Chr.19: 5641999-5641999 on Build GRCh38
rs748687499 Chr.19: 5641715-5641715 on Build GRCh38
rs1053760866 Chr.19: 5641934-5641934 on Build GRCh38
rs774767604 Chr.19: 5641762-5641762 on Build GRCh38
rs536870574 Chr.19: 5641706-5641706 on Build GRCh38
rs758696281 Chr.19: 5641925-5641925 on Build GRCh38
rs1026636004 Chr.19: 5642118-5642118 on Build GRCh38
rs780489708 Chr.19: 5641932-5641932 on Build GRCh38
rs757185559 Chr.19: 5642099-5642099 on Build GRCh38
rs768659441 Chr.19: 5641942-5641942 on Build GRCh38
rs1041352909 Chr.19: 5641887-5641887 on Build GRCh38
rs773072911 Chr.19: 5641895-5641895 on Build GRCh38

View Full Product Details