Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 11507258-11507762 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECSIT
Entrez Gene ID
51295
Gene Name
ECSIT signalling integrator
Gene Aliases
SITPEC
UniGene
Hs.515146
Gene Chromosome Location
Chr.19: 11505916-11529172 on Build GRCh38
Transcripts
NM_001142464.2 NM_001142465.2 NM_001243204.1 NM_016581.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3371148 c.945+1G>A p.? Chr.19: 11507701-11507701 on Build GRCh38
1250913 c.962C>T p.P321L Chr.19: 11507546-11507546 on Build GRCh38
74478 c.970T>C p.W324R Chr.19: 11507538-11507538 on Build GRCh38
4561034 c.987G>T p.P329P Chr.19: 11507521-11507521 on Build GRCh38
6651450 c.959C>T p.T320M Chr.19: 11507549-11507549 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146112007 Chr.19: 11507499-11507499 on Build GRCh38
rs751988626 Chr.19: 11507710-11507710 on Build GRCh38
rs1011868049 Chr.19: 11507566-11507566 on Build GRCh38
rs868352048 Chr.19: 11507712-11507712 on Build GRCh38
rs779422347 Chr.19: 11507514-11507514 on Build GRCh38
rs972092628 Chr.19: 11507352-11507352 on Build GRCh38
rs747238446 Chr.19: 11507501-11507502 on Build GRCh38
rs759251213 Chr.19: 11507557-11507557 on Build GRCh38
rs768857517 Chr.19: 11507522-11507522 on Build GRCh38
rs34894792 Chr.19: 11507729-11507730 on Build GRCh38
rs768821158 Chr.19: 11507436-11507436 on Build GRCh38
rs73512787 Chr.19: 11507606-11507606 on Build GRCh38
rs536609911 Chr.19: 11507308-11507308 on Build GRCh38
rs370740324 Chr.19: 11507519-11507519 on Build GRCh38
rs113749351 Chr.19: 11507456-11507456 on Build GRCh38
rs776565085 Chr.19: 11507410-11507410 on Build GRCh38
rs149646232 Chr.19: 11507437-11507437 on Build GRCh38
rs915520994 Chr.19: 11507425-11507425 on Build GRCh38
rs767767929 Chr.19: 11507714-11507714 on Build GRCh38
rs369095428 Chr.19: 11507587-11507587 on Build GRCh38
rs758484299 Chr.19: 11507412-11507412 on Build GRCh38
rs371970463 Chr.19: 11507417-11507417 on Build GRCh38
rs751883069 Chr.19: 11507595-11507595 on Build GRCh38
rs776919087 Chr.19: 11507401-11507401 on Build GRCh38
rs112991639 Chr.19: 11507455-11507455 on Build GRCh38
rs1031188090 Chr.19: 11507612-11507612 on Build GRCh38
rs780546343 Chr.19: 11507588-11507588 on Build GRCh38
rs371377965 Chr.19: 11507602-11507602 on Build GRCh38
rs200789689 Chr.19: 11507652-11507652 on Build GRCh38
rs771263631 Chr.19: 11507511-11507511 on Build GRCh38
rs781302551 Chr.19: 11507526-11507526 on Build GRCh38
rs758975335 Chr.19: 11507515-11507515 on Build GRCh38
rs902914462 Chr.19: 11507549-11507549 on Build GRCh38
rs776054391 Chr.19: 11507670-11507670 on Build GRCh38
rs532786856 Chr.19: 11507676-11507676 on Build GRCh38
rs761805525 Chr.19: 11507438-11507438 on Build GRCh38
rs767087208 Chr.19: 11507483-11507483 on Build GRCh38
rs771737772 Chr.19: 11507613-11507613 on Build GRCh38
rs376828070 Chr.19: 11507612-11507612 on Build GRCh38
rs747389355 Chr.19: 11507521-11507521 on Build GRCh38
rs1056150695 Chr.19: 11507489-11507489 on Build GRCh38
rs748210830 Chr.19: 11507531-11507531 on Build GRCh38
rs777986679 Chr.19: 11507609-11507609 on Build GRCh38
rs148594999 Chr.19: 11507703-11507703 on Build GRCh38
rs577799667 Chr.19: 11507439-11507439 on Build GRCh38
rs780386970 Chr.19: 11507520-11507520 on Build GRCh38
rs963680756 Chr.19: 11507649-11507649 on Build GRCh38
rs796265812 Chr.19: 11507511-11507511 on Build GRCh38
rs769739912 Chr.19: 11507533-11507533 on Build GRCh38
rs1011894402 Chr.19: 11507283-11507286 on Build GRCh38
rs369260261 Chr.19: 11507416-11507416 on Build GRCh38
rs775876527 Chr.19: 11507422-11507422 on Build GRCh38
rs141979045 Chr.19: 11507462-11507462 on Build GRCh38
rs747231598 Chr.19: 11507424-11507424 on Build GRCh38
rs12610434 Chr.19: 11507323-11507323 on Build GRCh38
rs760234305 Chr.19: 11507666-11507666 on Build GRCh38
rs755607091 Chr.19: 11507503-11507503 on Build GRCh38
rs901498666 Chr.19: 11507623-11507623 on Build GRCh38
rs780748201 Chr.19: 11507409-11507409 on Build GRCh38
rs199827839 Chr.19: 11507457-11507457 on Build GRCh38
rs540194701 Chr.19: 11507283-11507283 on Build GRCh38
rs970544535 Chr.19: 11507318-11507318 on Build GRCh38
rs990085570 Chr.19: 11507280-11507280 on Build GRCh38
rs60161140 Chr.19: 11507300-11507301 on Build GRCh38
rs544534253 Chr.19: 11507398-11507398 on Build GRCh38
rs763594826 Chr.19: 11507572-11507572 on Build GRCh38
rs201950682 Chr.19: 11507563-11507563 on Build GRCh38
rs772444027 Chr.19: 11507420-11507420 on Build GRCh38
rs555007668 Chr.19: 11507309-11507309 on Build GRCh38
rs774228011 Chr.19: 11507548-11507548 on Build GRCh38
rs560236685 Chr.19: 11507545-11507545 on Build GRCh38
rs763033896 Chr.19: 11507444-11507444 on Build GRCh38
rs199926928 Chr.19: 11507659-11507659 on Build GRCh38
rs560570155 Chr.19: 11507598-11507598 on Build GRCh38
rs781585683 Chr.19: 11507562-11507563 on Build GRCh38
rs548522099 Chr.19: 11507565-11507565 on Build GRCh38
rs773305938 Chr.19: 11507442-11507442 on Build GRCh38
rs574344159 Chr.19: 11507681-11507685 on Build GRCh38
rs377089751 Chr.19: 11507419-11507419 on Build GRCh38
rs759248886 Chr.19: 11507476-11507476 on Build GRCh38
rs763752425 Chr.19: 11507667-11507667 on Build GRCh38
rs777869199 Chr.19: 11507412-11507413 on Build GRCh38
rs374063655 Chr.19: 11507449-11507449 on Build GRCh38
rs376516130 Chr.19: 11507393-11507393 on Build GRCh38
rs112010013 Chr.19: 11507711-11507711 on Build GRCh38
rs749390864 Chr.19: 11507611-11507611 on Build GRCh38
rs970783807 Chr.19: 11507706-11507706 on Build GRCh38
rs1031136894 Chr.19: 11507394-11507394 on Build GRCh38
rs753408500 Chr.19: 11507585-11507585 on Build GRCh38
rs781658682 Chr.19: 11507605-11507605 on Build GRCh38
rs7254233 Chr.19: 11507317-11507317 on Build GRCh38
rs940540626 Chr.19: 11507523-11507523 on Build GRCh38
rs376676777 Chr.19: 11507536-11507536 on Build GRCh38
rs975449655 Chr.19: 11507448-11507448 on Build GRCh38

View Full Product Details