Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12696084-12696588 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXW9
Entrez Gene ID
84261
Gene Name
F-box and WD repeat domain containing 9
Gene Aliases
Fbw9
UniGene
Hs.515154
Gene Chromosome Location
Chr.19: 12688916-12696900 on Build GRCh38
Transcripts
NM_032301.2 XM_005260096.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6046520 c.174G>A p.A58A Chr.19: 12696408-12696408 on Build GRCh38
6662631 c.126A>C p.K42N Chr.19: 12696456-12696456 on Build GRCh38
5643128 c.341T>C p.V114A Chr.19: 12696241-12696241 on Build GRCh38
3756338 c.209G>A p.R70K Chr.19: 12696373-12696373 on Build GRCh38
3796644 c.331C>G p.R111G Chr.19: 12696251-12696251 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1028121237 Chr.19: 12696434-12696434 on Build GRCh38
rs769543984 Chr.19: 12696548-12696548 on Build GRCh38
rs928234753 Chr.19: 12696139-12696139 on Build GRCh38
rs950749623 Chr.19: 12696349-12696349 on Build GRCh38
rs753136581 Chr.19: 12696475-12696475 on Build GRCh38
rs746891816 Chr.19: 12696529-12696529 on Build GRCh38
rs768431805 Chr.19: 12696362-12696362 on Build GRCh38
rs935160765 Chr.19: 12696143-12696143 on Build GRCh38
rs979627663 Chr.19: 12696567-12696567 on Build GRCh38
rs1012376323 Chr.19: 12696562-12696562 on Build GRCh38
rs200089523 Chr.19: 12696422-12696422 on Build GRCh38
rs868844099 Chr.19: 12696463-12696463 on Build GRCh38
rs891038910 Chr.19: 12696405-12696405 on Build GRCh38
rs548297496 Chr.19: 12696296-12696296 on Build GRCh38
rs964809762 Chr.19: 12696312-12696312 on Build GRCh38
rs753453083 Chr.19: 12696271-12696271 on Build GRCh38
rs376213981 Chr.19: 12696291-12696291 on Build GRCh38
rs775938029 Chr.19: 12696149-12696150 on Build GRCh38
rs950833948 Chr.19: 12696451-12696451 on Build GRCh38
rs749877697 Chr.19: 12696374-12696374 on Build GRCh38
rs530002089 Chr.19: 12696295-12696295 on Build GRCh38
rs775469170 Chr.19: 12696353-12696353 on Build GRCh38
rs868025901 Chr.19: 12696248-12696248 on Build GRCh38
rs769299171 Chr.19: 12696450-12696450 on Build GRCh38
rs777977524 Chr.19: 12696385-12696385 on Build GRCh38
rs553598870 Chr.19: 12696389-12696389 on Build GRCh38
rs553892898 Chr.19: 12696136-12696149 on Build GRCh38
rs368929567 Chr.19: 12696194-12696194 on Build GRCh38
rs763810884 Chr.19: 12696256-12696256 on Build GRCh38
rs747890380 Chr.19: 12696547-12696547 on Build GRCh38
rs756413442 Chr.19: 12696384-12696384 on Build GRCh38
rs1045139100 Chr.19: 12696134-12696154 on Build GRCh38
rs758872905 Chr.19: 12696193-12696193 on Build GRCh38
rs748480913 Chr.19: 12696307-12696307 on Build GRCh38
rs747343374 Chr.19: 12696206-12696206 on Build GRCh38
rs770978013 Chr.19: 12696335-12696335 on Build GRCh38
rs775307628 Chr.19: 12696250-12696250 on Build GRCh38
rs139535728 Chr.19: 12696156-12696156 on Build GRCh38
rs772845492 Chr.19: 12696461-12696461 on Build GRCh38
rs774187734 Chr.19: 12696342-12696342 on Build GRCh38
rs745908660 Chr.19: 12696394-12696394 on Build GRCh38
rs568421602 Chr.19: 12696212-12696212 on Build GRCh38
rs776277398 Chr.19: 12696367-12696367 on Build GRCh38
rs886671917 Chr.19: 12696480-12696480 on Build GRCh38
rs749368874 Chr.19: 12696566-12696566 on Build GRCh38
rs767091197 Chr.19: 12696127-12696127 on Build GRCh38
rs369109827 Chr.19: 12696231-12696231 on Build GRCh38
rs764394969 Chr.19: 12696487-12696487 on Build GRCh38
rs766797950 Chr.19: 12696279-12696279 on Build GRCh38
rs762436615 Chr.19: 12696462-12696462 on Build GRCh38
rs947434069 Chr.19: 12696403-12696403 on Build GRCh38
rs774288718 Chr.19: 12696235-12696235 on Build GRCh38
rs769950410 Chr.19: 12696287-12696287 on Build GRCh38
rs1035019454 Chr.19: 12696497-12696497 on Build GRCh38
rs763650958 Chr.19: 12696153-12696153 on Build GRCh38
rs1037850635 Chr.19: 12696523-12696523 on Build GRCh38
rs374984061 Chr.19: 12696211-12696211 on Build GRCh38
rs749341678 Chr.19: 12696334-12696334 on Build GRCh38
rs983426197 Chr.19: 12696356-12696356 on Build GRCh38
rs777959073 Chr.19: 12696322-12696322 on Build GRCh38
rs780074445 Chr.19: 12696417-12696417 on Build GRCh38
rs984087728 Chr.19: 12696134-12696134 on Build GRCh38
rs756730283 Chr.19: 12696275-12696275 on Build GRCh38
rs151118454 Chr.19: 12696501-12696501 on Build GRCh38
rs866858302 Chr.19: 12696555-12696555 on Build GRCh38
rs745772249 Chr.19: 12696350-12696350 on Build GRCh38
rs1050786415 Chr.19: 12696398-12696398 on Build GRCh38
rs781079332 Chr.19: 12696536-12696536 on Build GRCh38
rs761573951 Chr.19: 12696439-12696439 on Build GRCh38
rs894897654 Chr.19: 12696192-12696192 on Build GRCh38
rs781202836 Chr.19: 12696134-12696140 on Build GRCh38
rs928181146 Chr.19: 12696363-12696363 on Build GRCh38
rs761060479 Chr.19: 12696478-12696478 on Build GRCh38
rs373375055 Chr.19: 12696421-12696421 on Build GRCh38
rs772683000 Chr.19: 12696556-12696556 on Build GRCh38
rs773320018 Chr.19: 12696225-12696225 on Build GRCh38
rs755318458 Chr.19: 12696288-12696288 on Build GRCh38
rs988897546 Chr.19: 12696147-12696147 on Build GRCh38
rs753118110 Chr.19: 12696382-12696382 on Build GRCh38
rs868516820 Chr.19: 12696292-12696292 on Build GRCh38
rs989261905 Chr.19: 12696379-12696379 on Build GRCh38
rs776167680 Chr.19: 12696423-12696423 on Build GRCh38
rs868168653 Chr.19: 12696257-12696257 on Build GRCh38
rs370265857 Chr.19: 12696499-12696499 on Build GRCh38
rs6511833 Chr.19: 12696373-12696373 on Build GRCh38
rs772206895 Chr.19: 12696412-12696412 on Build GRCh38
rs769980960 Chr.19: 12696221-12696221 on Build GRCh38
rs760128835 Chr.19: 12696375-12696375 on Build GRCh38
rs1006838229 Chr.19: 12696414-12696414 on Build GRCh38
rs755719320 Chr.19: 12696132-12696132 on Build GRCh38
rs752062927 Chr.19: 12696282-12696282 on Build GRCh38
rs756394955 Chr.19: 12696317-12696317 on Build GRCh38
rs571151401 Chr.19: 12696387-12696387 on Build GRCh38
rs973418747 Chr.19: 12696106-12696106 on Build GRCh38
rs929558780 Chr.19: 12696133-12696133 on Build GRCh38
rs749383428 Chr.19: 12696230-12696230 on Build GRCh38
rs933328443 Chr.19: 12696442-12696442 on Build GRCh38
rs569766529 Chr.19: 12696354-12696354 on Build GRCh38
rs759256928 Chr.19: 12696242-12696242 on Build GRCh38
rs779002622 Chr.19: 12696500-12696500 on Build GRCh38
rs772116742 Chr.19: 12696352-12696352 on Build GRCh38
rs558882751 Chr.19: 12696381-12696381 on Build GRCh38
rs534991617 Chr.19: 12696388-12696388 on Build GRCh38
rs752333341 Chr.19: 12696129-12696129 on Build GRCh38
rs754133669 Chr.19: 12696491-12696491 on Build GRCh38
rs768026306 Chr.19: 12696464-12696464 on Build GRCh38
rs867996943 Chr.19: 12696266-12696266 on Build GRCh38
rs780168680 Chr.19: 12696512-12696512 on Build GRCh38
rs371715059 Chr.19: 12696207-12696207 on Build GRCh38
rs769274322 Chr.19: 12696431-12696431 on Build GRCh38
rs6511832 Chr.19: 12696252-12696252 on Build GRCh38
rs769844221 Chr.19: 12696150-12696156 on Build GRCh38
rs761257668 Chr.19: 12696371-12696371 on Build GRCh38
rs749602876 Chr.19: 12696360-12696361 on Build GRCh38
rs973917092 Chr.19: 12696321-12696321 on Build GRCh38

View Full Product Details