Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12899914-12900413 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCDH
Entrez Gene ID
2639
Gene Name
glutaryl-CoA dehydrogenase
Gene Aliases
ACAD5, GCD
UniGene
Hs.532699
Gene Chromosome Location
Chr.19: 12891129-12915345 on Build GRCh38
Transcripts
NM_000159.3 NM_013976.3 NR_102316.1 NR_102317.1 XM_006722721.2 XM_011527899.2 XM_011527900.2 XM_017026580.1 NM_001105578.1 XM_005259848.4 XM_011527882.2 XM_011527883.2

Gene Symbol
SYCE2
Entrez Gene ID
256126
Gene Name
synaptonemal complex central element protein 2
Gene Aliases
CESC1
UniGene
Hs.655173
Gene Chromosome Location
Chr.19: 12898229-12919674 on Build GRCh38
Transcripts
NM_000159.3 NM_013976.3 NR_102316.1 NR_102317.1 XM_006722721.2 XM_011527899.2 XM_011527900.2 XM_017026580.1 NM_001105578.1 XM_005259848.4 XM_011527882.2 XM_011527883.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6903086 c.535G>A p.G179R Chr.19: 12900081-12900081 on Build GRCh38
335838 c.498G>C p.Q166H Chr.19: 12900118-12900118 on Build GRCh38
6793691 c.519delG p.P174fs*>45 Chr.19: 12900097-12900097 on Build GRCh38
4074531 c.592G>A p.V198M Chr.19: 12900024-12900024 on Build GRCh38
1267071 c.577G>A p.V193M Chr.19: 12900039-12900039 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751102827 Chr.19: 12900048-12900048 on Build GRCh38
rs200424351 Chr.19: 12900160-12900160 on Build GRCh38
rs1028116966 Chr.19: 12900202-12900202 on Build GRCh38
rs757639544 Chr.19: 12899966-12899966 on Build GRCh38
rs186538441 Chr.19: 12900060-12900060 on Build GRCh38
rs946738645 Chr.19: 12900254-12900254 on Build GRCh38
rs747163093 Chr.19: 12900071-12900071 on Build GRCh38
rs776665629 Chr.19: 12900033-12900033 on Build GRCh38
rs759115177 Chr.19: 12900054-12900054 on Build GRCh38
rs368686817 Chr.19: 12900264-12900264 on Build GRCh38
rs758633712 Chr.19: 12900065-12900065 on Build GRCh38
rs189554230 Chr.19: 12900077-12900077 on Build GRCh38
rs374710434 Chr.19: 12899986-12899986 on Build GRCh38
rs752198411 Chr.19: 12900162-12900162 on Build GRCh38
rs371431562 Chr.19: 12899959-12899959 on Build GRCh38
rs769837115 Chr.19: 12900115-12900115 on Build GRCh38
rs916667246 Chr.19: 12900230-12900230 on Build GRCh38
rs775491315 Chr.19: 12899991-12899991 on Build GRCh38
rs866882259 Chr.19: 12900298-12900298 on Build GRCh38
rs1055184832 Chr.19: 12900026-12900026 on Build GRCh38
rs781403406 Chr.19: 12900091-12900091 on Build GRCh38
rs773131972 Chr.19: 12900039-12900039 on Build GRCh38
rs1045583479 Chr.19: 12899955-12899955 on Build GRCh38
rs567509626 Chr.19: 12900327-12900327 on Build GRCh38
rs1006976076 Chr.19: 12900366-12900366 on Build GRCh38
rs976973476 Chr.19: 12900354-12900354 on Build GRCh38
rs748518757 Chr.19: 12900323-12900323 on Build GRCh38
rs370920733 Chr.19: 12900066-12900066 on Build GRCh38
rs747127884 Chr.19: 12900011-12900011 on Build GRCh38
rs557289830 Chr.19: 12900204-12900204 on Build GRCh38
rs368767319 Chr.19: 12900151-12900151 on Build GRCh38
rs759087160 Chr.19: 12900152-12900152 on Build GRCh38
rs973762433 Chr.19: 12900125-12900125 on Build GRCh38
rs772335144 Chr.19: 12899987-12899987 on Build GRCh38
rs767156949 Chr.19: 12899957-12899957 on Build GRCh38
rs772497833 Chr.19: 12900357-12900357 on Build GRCh38
rs766886984 Chr.19: 12900055-12900055 on Build GRCh38
rs548701419 Chr.19: 12900295-12900295 on Build GRCh38
rs181777132 Chr.19: 12900120-12900120 on Build GRCh38
rs779227372 Chr.19: 12899977-12899977 on Build GRCh38
rs370021453 Chr.19: 12900040-12900040 on Build GRCh38
rs754390527 Chr.19: 12900056-12900056 on Build GRCh38
rs953119022 Chr.19: 12900226-12900226 on Build GRCh38
rs768555038 Chr.19: 12900031-12900031 on Build GRCh38
rs991313967 Chr.19: 12900227-12900227 on Build GRCh38
rs748159256 Chr.19: 12900103-12900103 on Build GRCh38
rs530280983 Chr.19: 12899942-12899942 on Build GRCh38
rs569203348 Chr.19: 12900215-12900215 on Build GRCh38
rs895630922 Chr.19: 12900123-12900123 on Build GRCh38
rs145442966 Chr.19: 12900061-12900061 on Build GRCh38
rs749753298 Chr.19: 12899963-12899963 on Build GRCh38
rs567697090 Chr.19: 12900256-12900256 on Build GRCh38
rs1007732125 Chr.19: 12900239-12900239 on Build GRCh38
rs953147544 Chr.19: 12899960-12899960 on Build GRCh38
rs568057721 Chr.19: 12900136-12900136 on Build GRCh38
rs375724605 Chr.19: 12900034-12900034 on Build GRCh38
rs1051260722 Chr.19: 12900325-12900325 on Build GRCh38
rs766066084 Chr.19: 12900047-12900047 on Build GRCh38
rs985626042 Chr.19: 12899975-12899975 on Build GRCh38
rs770589323 Chr.19: 12900149-12900149 on Build GRCh38

View Full Product Details