Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 13104808-13105301 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRMT1
Entrez Gene ID
55621
Gene Name
tRNA methyltransferase 1
Gene Aliases
TRM1
UniGene
Hs.515169
Gene Chromosome Location
Chr.19: 13104900-13117568 on Build GRCh38
Transcripts
NM_001136035.2 NM_001142554.1 NM_017722.3 XM_006722793.2 XM_011528124.1 XM_011528125.1 XM_017026944.1 XM_017026945.1 XM_017026946.1 XM_017026947.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1168867 c.1880C>T p.P627L Chr.19: 13105035-13105035 on Build GRCh38
6808929 c.1823G>T p.R608M Chr.19: 13105277-13105277 on Build GRCh38
6808945 c.1839C>A p.T613T Chr.19: 13105076-13105076 on Build GRCh38
2817217 c.1919C>T p.P640L Chr.19: 13104996-13104996 on Build GRCh38
6808946 c.1943_1944insC p.G649fs*>12 Chr.19: 13104971-13104972 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1042305901 Chr.19: 13104910-13104910 on Build GRCh38
rs764162192 Chr.19: 13105251-13105251 on Build GRCh38
rs754811457 Chr.19: 13105077-13105077 on Build GRCh38
rs140561557 Chr.19: 13105191-13105191 on Build GRCh38
rs373766611 Chr.19: 13105072-13105072 on Build GRCh38
rs34011570 Chr.19: 13105277-13105277 on Build GRCh38
rs146810234 Chr.19: 13105151-13105151 on Build GRCh38
rs773579799 Chr.19: 13104978-13104978 on Build GRCh38
rs926699943 Chr.19: 13104829-13104829 on Build GRCh38
rs767236600 Chr.19: 13104972-13104972 on Build GRCh38
rs759977633 Chr.19: 13105042-13105042 on Build GRCh38
rs755918592 Chr.19: 13104971-13104971 on Build GRCh38
rs770151631 Chr.19: 13105053-13105053 on Build GRCh38
rs373070378 Chr.19: 13105130-13105130 on Build GRCh38
rs768223348 Chr.19: 13104903-13104909 on Build GRCh38
rs985320190 Chr.19: 13104841-13104841 on Build GRCh38
rs755820220 Chr.19: 13105008-13105008 on Build GRCh38
rs761336576 Chr.19: 13105006-13105006 on Build GRCh38
rs766296246 Chr.19: 13105236-13105240 on Build GRCh38
rs201403342 Chr.19: 13105040-13105040 on Build GRCh38
rs779691146 Chr.19: 13105013-13105013 on Build GRCh38
rs770201054 Chr.19: 13105108-13105108 on Build GRCh38
rs774151642 Chr.19: 13104961-13104962 on Build GRCh38
rs377547397 Chr.19: 13105249-13105249 on Build GRCh38
rs767888114 Chr.19: 13104908-13104908 on Build GRCh38
rs552583718 Chr.19: 13105079-13105079 on Build GRCh38
rs542840419 Chr.19: 13105095-13105095 on Build GRCh38
rs949571778 Chr.19: 13105198-13105198 on Build GRCh38
rs7254704 Chr.19: 13105159-13105159 on Build GRCh38
rs767856855 Chr.19: 13105063-13105063 on Build GRCh38
rs1047998291 Chr.19: 13105257-13105257 on Build GRCh38
rs773036580 Chr.19: 13105034-13105035 on Build GRCh38
rs746162494 Chr.19: 13105107-13105107 on Build GRCh38
rs889531945 Chr.19: 13105166-13105166 on Build GRCh38
rs771237572 Chr.19: 13105039-13105039 on Build GRCh38
rs554080215 Chr.19: 13105069-13105069 on Build GRCh38
rs761141515 Chr.19: 13104911-13104911 on Build GRCh38
rs775888842 Chr.19: 13105109-13105109 on Build GRCh38
rs899289639 Chr.19: 13105242-13105242 on Build GRCh38
rs773667785 Chr.19: 13104909-13104909 on Build GRCh38
rs781499853 Chr.19: 13105099-13105099 on Build GRCh38
rs1032311760 Chr.19: 13105035-13105035 on Build GRCh38
rs756339174 Chr.19: 13105276-13105276 on Build GRCh38
rs766999804 Chr.19: 13105068-13105068 on Build GRCh38
rs780854924 Chr.19: 13104958-13104958 on Build GRCh38
rs763502997 Chr.19: 13104939-13104939 on Build GRCh38
rs148754749 Chr.19: 13105001-13105001 on Build GRCh38
rs911008707 Chr.19: 13104859-13104859 on Build GRCh38
rs376828486 Chr.19: 13104952-13104952 on Build GRCh38
rs376780373 Chr.19: 13104976-13104976 on Build GRCh38
rs989758277 Chr.19: 13104897-13104897 on Build GRCh38
rs941077859 Chr.19: 13105182-13105182 on Build GRCh38
rs752674785 Chr.19: 13105084-13105084 on Build GRCh38
rs778646912 Chr.19: 13105078-13105078 on Build GRCh38
rs772280411 Chr.19: 13105036-13105036 on Build GRCh38
rs759941917 Chr.19: 13104991-13104991 on Build GRCh38
rs774637943 Chr.19: 13104902-13104902 on Build GRCh38
rs199990153 Chr.19: 13105096-13105096 on Build GRCh38
rs759174916 Chr.19: 13105133-13105133 on Build GRCh38
rs34077952 Chr.19: 13105142-13105143 on Build GRCh38
rs753706241 Chr.19: 13105018-13105018 on Build GRCh38
rs756872858 Chr.19: 13104956-13104956 on Build GRCh38
rs761486084 Chr.19: 13104971-13104972 on Build GRCh38
rs748265622 Chr.19: 13104975-13104975 on Build GRCh38
rs772668267 Chr.19: 13105115-13105115 on Build GRCh38
rs369390870 Chr.19: 13105067-13105067 on Build GRCh38
rs755577994 Chr.19: 13104928-13104928 on Build GRCh38
rs943988555 Chr.19: 13104872-13104872 on Build GRCh38
rs774547806 Chr.19: 13104896-13104896 on Build GRCh38
rs747223151 Chr.19: 13105038-13105038 on Build GRCh38
rs755544907 Chr.19: 13105081-13105081 on Build GRCh38
rs753247652 Chr.19: 13105185-13105185 on Build GRCh38
rs371881044 Chr.19: 13104972-13104972 on Build GRCh38
rs368586523 Chr.19: 13105034-13105034 on Build GRCh38
rs937849760 Chr.19: 13104873-13104873 on Build GRCh38
rs749771351 Chr.19: 13105110-13105110 on Build GRCh38
rs142786869 Chr.19: 13105252-13105252 on Build GRCh38
rs577720271 Chr.19: 13105032-13105032 on Build GRCh38
rs35911417 Chr.19: 13105120-13105120 on Build GRCh38
rs778368877 Chr.19: 13105267-13105267 on Build GRCh38
rs745615872 Chr.19: 13104966-13104966 on Build GRCh38
rs765114566 Chr.19: 13105228-13105228 on Build GRCh38
rs1038139978 Chr.19: 13105205-13105205 on Build GRCh38
rs753523817 Chr.19: 13105075-13105075 on Build GRCh38
rs369638881 Chr.19: 13105112-13105112 on Build GRCh38
rs753972967 Chr.19: 13105264-13105265 on Build GRCh38
rs765885788 Chr.19: 13105128-13105128 on Build GRCh38
rs900809827 Chr.19: 13104915-13104915 on Build GRCh38
rs537821803 Chr.19: 13104916-13104916 on Build GRCh38
rs144546583 Chr.19: 13104973-13104973 on Build GRCh38
rs757372487 Chr.19: 13105258-13105258 on Build GRCh38
rs567404387 Chr.19: 13104903-13104903 on Build GRCh38
rs373190081 Chr.19: 13105264-13105264 on Build GRCh38
rs777116447 Chr.19: 13104984-13104984 on Build GRCh38
rs760567376 Chr.19: 13105131-13105132 on Build GRCh38
rs748795001 Chr.19: 13104885-13104891 on Build GRCh38
rs879339012 Chr.19: 13105194-13105194 on Build GRCh38
rs1049460638 Chr.19: 13104948-13104948 on Build GRCh38
rs556159504 Chr.19: 13105024-13105024 on Build GRCh38
rs1003525570 Chr.19: 13105266-13105266 on Build GRCh38
rs201263674 Chr.19: 13105092-13105092 on Build GRCh38
rs750070085 Chr.19: 13105007-13105007 on Build GRCh38
rs1006881598 Chr.19: 13105268-13105268 on Build GRCh38
rs35562739 Chr.19: 13105225-13105225 on Build GRCh38
rs760251487 Chr.19: 13105116-13105116 on Build GRCh38

View Full Product Details