Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 13795306-13795795 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZSWIM4
Entrez Gene ID
65249
Gene Name
zinc finger SWIM-type containing 4
Gene Aliases
-
UniGene
Hs.466015
Gene Chromosome Location
Chr.19: 13795424-13832254 on Build GRCh38
Transcripts
NM_023072.2 XM_005260023.3 XM_017027153.1 XM_017027154.1 XM_017027155.1 XM_017027156.1 XM_017027157.1

SNP Detail

SNP ID Chromosome Location
rs1002333943 Chr.19: 13795633-13795633 on Build GRCh38
rs906688318 Chr.19: 13795397-13795397 on Build GRCh38
rs3745458 Chr.19: 13795415-13795415 on Build GRCh38
rs921247151 Chr.19: 13795498-13795498 on Build GRCh38
rs750280643 Chr.19: 13795765-13795765 on Build GRCh38
rs1015953921 Chr.19: 13795660-13795660 on Build GRCh38
rs951141951 Chr.19: 13795359-13795359 on Build GRCh38
rs1009654953 Chr.19: 13795370-13795370 on Build GRCh38
rs936660741 Chr.19: 13795354-13795354 on Build GRCh38
rs993220253 Chr.19: 13795685-13795685 on Build GRCh38
rs971769666 Chr.19: 13795481-13795481 on Build GRCh38
rs552374783 Chr.19: 13795646-13795646 on Build GRCh38
rs570678207 Chr.19: 13795696-13795696 on Build GRCh38
rs983032408 Chr.19: 13795387-13795387 on Build GRCh38
rs971905544 Chr.19: 13795333-13795333 on Build GRCh38
rs566059647 Chr.19: 13795325-13795325 on Build GRCh38
rs969069195 Chr.19: 13795326-13795326 on Build GRCh38
rs933261846 Chr.19: 13795338-13795338 on Build GRCh38
rs923136156 Chr.19: 13795491-13795491 on Build GRCh38
rs1054279914 Chr.19: 13795356-13795356 on Build GRCh38
rs928455904 Chr.19: 13795396-13795396 on Build GRCh38
rs1027134735 Chr.19: 13795355-13795355 on Build GRCh38
rs749964814 Chr.19: 13795414-13795414 on Build GRCh38
rs112193788 Chr.19: 13795578-13795578 on Build GRCh38
rs372250403 Chr.19: 13795541-13795541 on Build GRCh38
rs761230823 Chr.19: 13795712-13795712 on Build GRCh38
rs1006213778 Chr.19: 13795656-13795656 on Build GRCh38
rs1030011400 Chr.19: 13795694-13795694 on Build GRCh38
rs867550761 Chr.19: 13795510-13795510 on Build GRCh38
rs892445011 Chr.19: 13795588-13795588 on Build GRCh38
rs958005854 Chr.19: 13795773-13795773 on Build GRCh38
rs961808537 Chr.19: 13795466-13795466 on Build GRCh38
rs1053823449 Chr.19: 13795584-13795584 on Build GRCh38
rs956557777 Chr.19: 13795436-13795436 on Build GRCh38
rs887201118 Chr.19: 13795548-13795548 on Build GRCh38
rs766866513 Chr.19: 13795721-13795721 on Build GRCh38
rs751179392 Chr.19: 13795739-13795739 on Build GRCh38
rs373636646 Chr.19: 13795351-13795351 on Build GRCh38
rs946648627 Chr.19: 13795449-13795449 on Build GRCh38
rs775001644 Chr.19: 13795724-13795724 on Build GRCh38
rs906719598 Chr.19: 13795616-13795617 on Build GRCh38
rs868144069 Chr.19: 13795335-13795335 on Build GRCh38
rs940527507 Chr.19: 13795331-13795331 on Build GRCh38
rs756950298 Chr.19: 13795748-13795748 on Build GRCh38
rs767283792 Chr.19: 13795757-13795757 on Build GRCh38
rs557063494 Chr.19: 13795413-13795413 on Build GRCh38
rs915122907 Chr.19: 13795443-13795443 on Build GRCh38
rs1055794360 Chr.19: 13795638-13795638 on Build GRCh38
rs763756010 Chr.19: 13795734-13795734 on Build GRCh38
rs892515668 Chr.19: 13795358-13795358 on Build GRCh38
rs988387015 Chr.19: 13795437-13795437 on Build GRCh38
rs534850044 Chr.19: 13795727-13795727 on Build GRCh38
rs1025173386 Chr.19: 13795373-13795373 on Build GRCh38
rs1051182351 Chr.19: 13795523-13795523 on Build GRCh38
rs530103423 Chr.19: 13795348-13795348 on Build GRCh38
rs1039625758 Chr.19: 13795471-13795471 on Build GRCh38
rs936674218 Chr.19: 13795577-13795577 on Build GRCh38
rs1046490121 Chr.19: 13795612-13795612 on Build GRCh38
rs1016335753 Chr.19: 13795438-13795438 on Build GRCh38
rs754566232 Chr.19: 13795334-13795334 on Build GRCh38
rs945473873 Chr.19: 13795592-13795592 on Build GRCh38
rs910367681 Chr.19: 13795732-13795732 on Build GRCh38

View Full Product Details