Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 45017115-45017625 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZSWIM5
Entrez Gene ID
57643
Gene Name
zinc finger SWIM-type containing 5
Gene Aliases
-
UniGene
Hs.656613
Gene Chromosome Location
Chr.1: 45016399-45206586 on Build GRCh38
Transcripts
NM_020883.1 XM_017001913.1

SNP Detail

SNP ID Chromosome Location
rs1015555904 Chr.1: 45017381-45017381 on Build GRCh38
rs892130550 Chr.1: 45017311-45017311 on Build GRCh38
rs369883773 Chr.1: 45017481-45017481 on Build GRCh38
rs372375955 Chr.1: 45017158-45017158 on Build GRCh38
rs142502451 Chr.1: 45017411-45017411 on Build GRCh38
rs925119082 Chr.1: 45017501-45017501 on Build GRCh38
rs757530871 Chr.1: 45017147-45017147 on Build GRCh38
rs983280025 Chr.1: 45017378-45017380 on Build GRCh38
rs1025262752 Chr.1: 45017342-45017342 on Build GRCh38
rs778001000 Chr.1: 45017348-45017348 on Build GRCh38
rs578076512 Chr.1: 45017159-45017159 on Build GRCh38
rs750541558 Chr.1: 45017555-45017556 on Build GRCh38
rs779951776 Chr.1: 45017439-45017439 on Build GRCh38
rs548453133 Chr.1: 45017349-45017349 on Build GRCh38
rs923214979 Chr.1: 45017445-45017445 on Build GRCh38
rs3831960 Chr.1: 45017546-45017546 on Build GRCh38
rs758499933 Chr.1: 45017261-45017261 on Build GRCh38
rs949059627 Chr.1: 45017563-45017563 on Build GRCh38
rs41306605 Chr.1: 45017583-45017583 on Build GRCh38
rs542904632 Chr.1: 45017407-45017407 on Build GRCh38
rs557939421 Chr.1: 45017148-45017148 on Build GRCh38
rs545203303 Chr.1: 45017503-45017503 on Build GRCh38
rs140223641 Chr.1: 45017387-45017387 on Build GRCh38
rs538433682 Chr.1: 45017138-45017138 on Build GRCh38
rs1050581881 Chr.1: 45017184-45017185 on Build GRCh38
rs780028759 Chr.1: 45017323-45017324 on Build GRCh38
rs1052652143 Chr.1: 45017529-45017529 on Build GRCh38
rs975959234 Chr.1: 45017393-45017393 on Build GRCh38
rs746941366 Chr.1: 45017378-45017379 on Build GRCh38
rs985954407 Chr.1: 45017483-45017483 on Build GRCh38
rs376744032 Chr.1: 45017192-45017192 on Build GRCh38
rs554256831 Chr.1: 45017360-45017360 on Build GRCh38
rs936432917 Chr.1: 45017250-45017250 on Build GRCh38
rs936397574 Chr.1: 45017506-45017511 on Build GRCh38
rs1027584105 Chr.1: 45017377-45017377 on Build GRCh38
rs531377323 Chr.1: 45017468-45017468 on Build GRCh38
rs761551799 Chr.1: 45017545-45017546 on Build GRCh38
rs771297366 Chr.1: 45017413-45017413 on Build GRCh38
rs903580741 Chr.1: 45017187-45017187 on Build GRCh38
rs150935572 Chr.1: 45017597-45017597 on Build GRCh38
rs905346254 Chr.1: 45017343-45017343 on Build GRCh38
rs540612673 Chr.1: 45017202-45017202 on Build GRCh38
rs796943882 Chr.1: 45017506-45017509 on Build GRCh38

View Full Product Details