Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 45515678-45516188 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRDX1
Entrez Gene ID
5052
Gene Name
peroxiredoxin 1
Gene Aliases
MSP23, NKEF-A, NKEFA, PAG, PAGA, PAGB, PRX1, PRXI, TDPX2
UniGene
Hs.180909
Gene Chromosome Location
Chr.1: 45511035-45522890 on Build GRCh38
Transcripts
NM_001202431.1 NM_002574.3 NM_181696.2 NM_181697.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7448931 c.183T>G p.D61E Chr.1: 45515731-45515731 on Build GRCh38
5551265 c.154T>A p.C52S Chr.1: 45515760-45515760 on Build GRCh38
6755241 c.208A>T p.N70Y Chr.1: 45515706-45515706 on Build GRCh38
352919 c.161C>T p.T54M Chr.1: 45515753-45515753 on Build GRCh38
4577395 c.165G>A p.E55E Chr.1: 45515749-45515749 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775525174 Chr.1: 45515833-45515833 on Build GRCh38
rs995701801 Chr.1: 45516133-45516133 on Build GRCh38
rs947255888 Chr.1: 45515897-45515897 on Build GRCh38
rs770218614 Chr.1: 45515711-45515718 on Build GRCh38
rs777940810 Chr.1: 45515705-45515705 on Build GRCh38
rs999603852 Chr.1: 45516024-45516024 on Build GRCh38
rs188866583 Chr.1: 45515968-45515968 on Build GRCh38
rs903121201 Chr.1: 45515961-45515961 on Build GRCh38
rs746890055 Chr.1: 45515839-45515839 on Build GRCh38
rs34034070 Chr.1: 45515730-45515730 on Build GRCh38
rs58257925 Chr.1: 45515845-45515846 on Build GRCh38
rs544397110 Chr.1: 45515997-45515997 on Build GRCh38
rs1039629416 Chr.1: 45515894-45515894 on Build GRCh38
rs776500101 Chr.1: 45515841-45515841 on Build GRCh38
rs959844084 Chr.1: 45515734-45515734 on Build GRCh38
rs538020423 Chr.1: 45515709-45515709 on Build GRCh38
rs757405965 Chr.1: 45515817-45515817 on Build GRCh38
rs902213721 Chr.1: 45516021-45516021 on Build GRCh38
rs944854932 Chr.1: 45515890-45515890 on Build GRCh38
rs564299496 Chr.1: 45516027-45516027 on Build GRCh38
rs895446589 Chr.1: 45516074-45516074 on Build GRCh38
rs759398353 Chr.1: 45515729-45515729 on Build GRCh38
rs761495506 Chr.1: 45515754-45515754 on Build GRCh38
rs974002513 Chr.1: 45516082-45516082 on Build GRCh38
rs1011185698 Chr.1: 45515936-45515936 on Build GRCh38
rs867120959 Chr.1: 45515742-45515742 on Build GRCh38
rs963123449 Chr.1: 45516101-45516101 on Build GRCh38
rs139756090 Chr.1: 45515822-45515822 on Build GRCh38
rs912525963 Chr.1: 45515871-45515871 on Build GRCh38
rs756503279 Chr.1: 45515814-45515814 on Build GRCh38
rs1023455735 Chr.1: 45516077-45516077 on Build GRCh38
rs796103629 Chr.1: 45516066-45516066 on Build GRCh38
rs145270179 Chr.1: 45515800-45515800 on Build GRCh38
rs1022757141 Chr.1: 45516086-45516086 on Build GRCh38
rs969512923 Chr.1: 45515777-45515777 on Build GRCh38
rs757483506 Chr.1: 45516100-45516100 on Build GRCh38
rs140856676 Chr.1: 45515746-45515746 on Build GRCh38
rs6699702 Chr.1: 45516162-45516162 on Build GRCh38
rs1021462796 Chr.1: 45515713-45515713 on Build GRCh38
rs1042868469 Chr.1: 45515937-45515937 on Build GRCh38
rs747134452 Chr.1: 45516081-45516081 on Build GRCh38
rs35635144 Chr.1: 45515842-45515843 on Build GRCh38
rs1021965997 Chr.1: 45515998-45515998 on Build GRCh38
rs778262329 Chr.1: 45515783-45515784 on Build GRCh38
rs1034733397 Chr.1: 45516042-45516042 on Build GRCh38
rs999163640 Chr.1: 45515982-45515982 on Build GRCh38
rs900741545 Chr.1: 45515914-45515914 on Build GRCh38
rs142590526 Chr.1: 45515744-45515744 on Build GRCh38
rs960753919 Chr.1: 45516053-45516053 on Build GRCh38
rs578052057 Chr.1: 45516054-45516054 on Build GRCh38
rs944155410 Chr.1: 45515879-45515879 on Build GRCh38
rs778050612 Chr.1: 45515881-45515881 on Build GRCh38
rs771640438 Chr.1: 45515829-45515830 on Build GRCh38
rs775210395 Chr.1: 45515735-45515735 on Build GRCh38
rs764841073 Chr.1: 45515755-45515755 on Build GRCh38
rs760500844 Chr.1: 45515737-45515737 on Build GRCh38
rs745328384 Chr.1: 45515810-45515810 on Build GRCh38
rs146706025 Chr.1: 45515779-45515779 on Build GRCh38
rs903090629 Chr.1: 45515717-45515717 on Build GRCh38
rs755494211 Chr.1: 45515781-45515781 on Build GRCh38
rs199654232 Chr.1: 45515816-45515816 on Build GRCh38
rs112565051 Chr.1: 45515758-45515758 on Build GRCh38
rs771956478 Chr.1: 45515829-45515829 on Build GRCh38
rs921103341 Chr.1: 45515885-45515885 on Build GRCh38
rs2356559 Chr.1: 45515840-45515840 on Build GRCh38
rs533609014 Chr.1: 45515749-45515749 on Build GRCh38
rs929290436 Chr.1: 45516122-45516122 on Build GRCh38
rs781707654 Chr.1: 45515782-45515782 on Build GRCh38
rs386630862 Chr.1: 45515840-45515843 on Build GRCh38
rs771022045 Chr.1: 45515721-45515721 on Build GRCh38

View Full Product Details