Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 13931003-13931505 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PODNL1
Entrez Gene ID
79883
Gene Name
podocan like 1
Gene Aliases
SLRR5B
UniGene
Hs.448497
Gene Chromosome Location
Chr.19: 13931187-13953392 on Build GRCh38
Transcripts
NM_001146254.1 NM_001146255.1 NM_024825.3 XM_006722901.1 XM_006722902.1 XM_006722903.1 XM_011528308.1 XM_011528309.2 XM_011528310.2 XM_011528311.1 XM_011528313.1 XM_011528314.1 XM_011528316.1

SNP Detail

SNP ID Chromosome Location
rs572001655 Chr.19: 13931178-13931178 on Build GRCh38
rs115774711 Chr.19: 13931157-13931157 on Build GRCh38
rs541133529 Chr.19: 13931314-13931314 on Build GRCh38
rs180792666 Chr.19: 13931105-13931105 on Build GRCh38
rs1043692639 Chr.19: 13931161-13931161 on Build GRCh38
rs568889467 Chr.19: 13931439-13931439 on Build GRCh38
rs980172078 Chr.19: 13931220-13931220 on Build GRCh38
rs77875973 Chr.19: 13931044-13931045 on Build GRCh38
rs552799386 Chr.19: 13931451-13931451 on Build GRCh38
rs1049892825 Chr.19: 13931221-13931221 on Build GRCh38
rs553986275 Chr.19: 13931464-13931464 on Build GRCh38
rs944510584 Chr.19: 13931290-13931290 on Build GRCh38
rs926450166 Chr.19: 13931431-13931431 on Build GRCh38
rs916815424 Chr.19: 13931198-13931198 on Build GRCh38
rs762119139 Chr.19: 13931286-13931286 on Build GRCh38
rs925254990 Chr.19: 13931467-13931467 on Build GRCh38
rs948422634 Chr.19: 13931219-13931220 on Build GRCh38
rs968541526 Chr.19: 13931112-13931112 on Build GRCh38
rs913802153 Chr.19: 13931127-13931127 on Build GRCh38
rs375450619 Chr.19: 13931432-13931432 on Build GRCh38
rs1047452208 Chr.19: 13931248-13931248 on Build GRCh38
rs903967888 Chr.19: 13931171-13931171 on Build GRCh38
rs886246302 Chr.19: 13931063-13931063 on Build GRCh38
rs960887373 Chr.19: 13931379-13931379 on Build GRCh38
rs951788715 Chr.19: 13931336-13931336 on Build GRCh38
rs763131579 Chr.19: 13931370-13931370 on Build GRCh38
rs528846717 Chr.19: 13931326-13931326 on Build GRCh38
rs1057193 Chr.19: 13931448-13931448 on Build GRCh38
rs563981952 Chr.19: 13931247-13931247 on Build GRCh38
rs533638531 Chr.19: 13931303-13931303 on Build GRCh38
rs781492808 Chr.19: 13931479-13931479 on Build GRCh38
rs563773228 Chr.19: 13931167-13931167 on Build GRCh38
rs561514816 Chr.19: 13931324-13931324 on Build GRCh38
rs992389205 Chr.19: 13931447-13931447 on Build GRCh38
rs886297708 Chr.19: 13931258-13931258 on Build GRCh38
rs186033965 Chr.19: 13931113-13931113 on Build GRCh38
rs150182802 Chr.19: 13931356-13931356 on Build GRCh38
rs545551701 Chr.19: 13931220-13931220 on Build GRCh38
rs994718274 Chr.19: 13931037-13931038 on Build GRCh38
rs1015880475 Chr.19: 13931307-13931307 on Build GRCh38
rs868685051 Chr.19: 13931106-13931106 on Build GRCh38
rs142880550 Chr.19: 13931060-13931060 on Build GRCh38
rs1000035776 Chr.19: 13931315-13931315 on Build GRCh38
rs565638018 Chr.19: 13931029-13931029 on Build GRCh38
rs140938573 Chr.19: 13931222-13931222 on Build GRCh38
rs978166411 Chr.19: 13931147-13931147 on Build GRCh38
rs891256201 Chr.19: 13931238-13931238 on Build GRCh38
rs7250453 Chr.19: 13931078-13931078 on Build GRCh38
rs776780685 Chr.19: 13931125-13931125 on Build GRCh38
rs979417611 Chr.19: 13931466-13931466 on Build GRCh38
rs373820012 Chr.19: 13931053-13931053 on Build GRCh38
rs960607346 Chr.19: 13931433-13931433 on Build GRCh38
rs776777461 Chr.19: 13931262-13931262 on Build GRCh38
rs961914882 Chr.19: 13931310-13931310 on Build GRCh38
rs898900745 Chr.19: 13931026-13931026 on Build GRCh38
rs980481933 Chr.19: 13931351-13931351 on Build GRCh38
rs764492948 Chr.19: 13931396-13931396 on Build GRCh38
rs1008838796 Chr.19: 13931070-13931070 on Build GRCh38
rs541792629 Chr.19: 13931082-13931082 on Build GRCh38

View Full Product Details