Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 13962862-13963391 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RFX1
Entrez Gene ID
5989
Gene Name
regulatory factor X1
Gene Aliases
EFC, RFX
UniGene
Hs.655215
Gene Chromosome Location
Chr.19: 13961530-14007514 on Build GRCh38
Transcripts
NM_002918.4 XM_011528165.2 XM_011528166.2 XM_011528167.2 XM_011528168.2 XM_011528169.2 XM_011528170.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4820876 c.2671G>A p.E891K Chr.19: 13963175-13963175 on Build GRCh38
6765350 c.2679C>T p.R893R Chr.19: 13963167-13963167 on Build GRCh38
3281469 c.2710G>A p.A904T Chr.19: 13963136-13963136 on Build GRCh38
3822090 c.2773G>T p.E925* Chr.19: 13962862-13962862 on Build GRCh38
3281470 c.2649_2651delCGA p.D883delD Chr.19: 13963195-13963197 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746451025 Chr.19: 13963032-13963032 on Build GRCh38
rs773470466 Chr.19: 13963327-13963327 on Build GRCh38
rs922451833 Chr.19: 13963007-13963015 on Build GRCh38
rs897048322 Chr.19: 13963218-13963218 on Build GRCh38
rs373669117 Chr.19: 13963166-13963166 on Build GRCh38
rs937391929 Chr.19: 13962948-13962948 on Build GRCh38
rs1057315952 Chr.19: 13962954-13962954 on Build GRCh38
rs954008509 Chr.19: 13963299-13963299 on Build GRCh38
rs751876803 Chr.19: 13963009-13963009 on Build GRCh38
rs543046067 Chr.19: 13962966-13962966 on Build GRCh38
rs760515783 Chr.19: 13963053-13963053 on Build GRCh38
rs771302231 Chr.19: 13963030-13963030 on Build GRCh38
rs370229283 Chr.19: 13963203-13963203 on Build GRCh38
rs1048363489 Chr.19: 13962915-13962915 on Build GRCh38
rs950299841 Chr.19: 13963106-13963106 on Build GRCh38
rs200524505 Chr.19: 13963117-13963117 on Build GRCh38
rs796123607 Chr.19: 13963002-13963002 on Build GRCh38
rs756579764 Chr.19: 13963103-13963103 on Build GRCh38
rs773021409 Chr.19: 13963052-13963052 on Build GRCh38
rs767980413 Chr.19: 13963086-13963086 on Build GRCh38
rs756457911 Chr.19: 13963149-13963149 on Build GRCh38
rs953834496 Chr.19: 13963033-13963033 on Build GRCh38
rs773074406 Chr.19: 13962900-13962900 on Build GRCh38
rs761967952 Chr.19: 13963132-13963132 on Build GRCh38
rs749285244 Chr.19: 13963197-13963197 on Build GRCh38
rs749089174 Chr.19: 13963301-13963301 on Build GRCh38
rs756891688 Chr.19: 13963159-13963159 on Build GRCh38
rs377026593 Chr.19: 13962901-13962901 on Build GRCh38
rs912356431 Chr.19: 13962892-13962892 on Build GRCh38
rs751815598 Chr.19: 13963082-13963082 on Build GRCh38
rs904776591 Chr.19: 13962981-13962981 on Build GRCh38
rs757967033 Chr.19: 13963156-13963156 on Build GRCh38
rs746716048 Chr.19: 13963111-13963111 on Build GRCh38
rs780774852 Chr.19: 13963115-13963115 on Build GRCh38
rs747615075 Chr.19: 13963046-13963046 on Build GRCh38
rs1033497501 Chr.19: 13963317-13963317 on Build GRCh38
rs752734917 Chr.19: 13962977-13962977 on Build GRCh38
rs965622033 Chr.19: 13963043-13963043 on Build GRCh38
rs750768887 Chr.19: 13963090-13963090 on Build GRCh38
rs549662297 Chr.19: 13963105-13963105 on Build GRCh38
rs757206875 Chr.19: 13963328-13963328 on Build GRCh38
rs925951941 Chr.19: 13962885-13962885 on Build GRCh38
rs763926758 Chr.19: 13963063-13963063 on Build GRCh38
rs780862548 Chr.19: 13963290-13963290 on Build GRCh38
rs774349669 Chr.19: 13963206-13963206 on Build GRCh38
rs780727282 Chr.19: 13963161-13963161 on Build GRCh38
rs772085139 Chr.19: 13963323-13963323 on Build GRCh38
rs771863371 Chr.19: 13962897-13962897 on Build GRCh38
rs745649726 Chr.19: 13963167-13963167 on Build GRCh38
rs759462894 Chr.19: 13963063-13963063 on Build GRCh38
rs115223700 Chr.19: 13963118-13963118 on Build GRCh38
rs201876870 Chr.19: 13963083-13963083 on Build GRCh38
rs371009810 Chr.19: 13963079-13963079 on Build GRCh38
rs758022031 Chr.19: 13963108-13963108 on Build GRCh38
rs754267216 Chr.19: 13963262-13963262 on Build GRCh38
rs560687844 Chr.19: 13963077-13963077 on Build GRCh38
rs539077393 Chr.19: 13963335-13963335 on Build GRCh38
rs778846577 Chr.19: 13963316-13963316 on Build GRCh38
rs769521923 Chr.19: 13963176-13963176 on Build GRCh38
rs374283010 Chr.19: 13962986-13962986 on Build GRCh38
rs754461637 Chr.19: 13963153-13963153 on Build GRCh38
rs781517197 Chr.19: 13963022-13963022 on Build GRCh38
rs755927474 Chr.19: 13963296-13963296 on Build GRCh38
rs751519337 Chr.19: 13963277-13963278 on Build GRCh38
rs765093401 Chr.19: 13963066-13963066 on Build GRCh38
rs892085739 Chr.19: 13962896-13962896 on Build GRCh38
rs937932698 Chr.19: 13962959-13962959 on Build GRCh38
rs771810258 Chr.19: 13963051-13963051 on Build GRCh38
rs547182582 Chr.19: 13963001-13963001 on Build GRCh38
rs377632154 Chr.19: 13963042-13963042 on Build GRCh38
rs779769264 Chr.19: 13963298-13963298 on Build GRCh38
rs778006176 Chr.19: 13963008-13963008 on Build GRCh38
rs369465188 Chr.19: 13963239-13963239 on Build GRCh38
rs773868586 Chr.19: 13962886-13962886 on Build GRCh38
rs766711898 Chr.19: 13963250-13963250 on Build GRCh38
rs757663788 Chr.19: 13963013-13963013 on Build GRCh38
rs767730679 Chr.19: 13963221-13963221 on Build GRCh38
rs762993936 Chr.19: 13963125-13963125 on Build GRCh38
rs1005618253 Chr.19: 13963185-13963185 on Build GRCh38
rs564828373 Chr.19: 13962970-13962970 on Build GRCh38
rs777563498 Chr.19: 13963110-13963110 on Build GRCh38
rs1006650772 Chr.19: 13963333-13963333 on Build GRCh38
rs961298661 Chr.19: 13962987-13962987 on Build GRCh38
rs377574659 Chr.19: 13962895-13962895 on Build GRCh38
rs199954679 Chr.19: 13962982-13962982 on Build GRCh38
rs768542253 Chr.19: 13963305-13963305 on Build GRCh38
rs927410874 Chr.19: 13962931-13962931 on Build GRCh38
rs761095917 Chr.19: 13963245-13963245 on Build GRCh38
rs770917280 Chr.19: 13963057-13963057 on Build GRCh38
rs776420729 Chr.19: 13963062-13963062 on Build GRCh38
rs768035994 Chr.19: 13963034-13963034 on Build GRCh38
rs753854399 Chr.19: 13962952-13962952 on Build GRCh38
rs766397295 Chr.19: 13962943-13962943 on Build GRCh38
rs370425816 Chr.19: 13963281-13963281 on Build GRCh38
rs199537041 Chr.19: 13963158-13963158 on Build GRCh38
rs775406087 Chr.19: 13963181-13963181 on Build GRCh38
rs750760412 Chr.19: 13963148-13963148 on Build GRCh38
rs765874097 Chr.19: 13963283-13963283 on Build GRCh38
rs113449727 Chr.19: 13962912-13962912 on Build GRCh38
rs752304559 Chr.19: 13963042-13963045 on Build GRCh38
rs376362982 Chr.19: 13963212-13963212 on Build GRCh38
rs571560628 Chr.19: 13963168-13963168 on Build GRCh38
rs764229128 Chr.19: 13963127-13963127 on Build GRCh38
rs1005258893 Chr.19: 13962983-13962983 on Build GRCh38
rs976529787 Chr.19: 13962995-13962995 on Build GRCh38
rs906260704 Chr.19: 13962913-13962913 on Build GRCh38
rs758149240 Chr.19: 13963046-13963048 on Build GRCh38
rs918494242 Chr.19: 13962899-13962899 on Build GRCh38
rs751062175 Chr.19: 13963285-13963285 on Build GRCh38
rs767639250 Chr.19: 13963144-13963144 on Build GRCh38
rs201438389 Chr.19: 13963116-13963116 on Build GRCh38
rs766888168 Chr.19: 13963151-13963151 on Build GRCh38

View Full Product Details