Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 16087906-16088403 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TPM4
Entrez Gene ID
7171
Gene Name
tropomyosin 4
Gene Aliases
HEL-S-108
UniGene
Hs.631618
Gene Chromosome Location
Chr.19: 16067507-16103005 on Build GRCh38
Transcripts
NM_001145160.1 NM_003290.2 XM_005260042.3 XM_006722865.2 XM_011528231.2 XM_011528232.1 XM_017027188.1 XM_017027189.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6806881 c.505C>T p.L169L Chr.19: 16088039-16088039 on Build GRCh38
6882284 c.500G>A p.R167H Chr.19: 16088034-16088034 on Build GRCh38
4279041 c.533G>A p.R178K Chr.19: 16088067-16088067 on Build GRCh38
992405 c.563+2T>C p.? Chr.19: 16088099-16088099 on Build GRCh38
5422005 c.499C>T p.R167C Chr.19: 16088033-16088033 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373195006 Chr.19: 16087997-16087997 on Build GRCh38
rs769001665 Chr.19: 16088144-16088144 on Build GRCh38
rs777074669 Chr.19: 16087989-16087989 on Build GRCh38
rs932802105 Chr.19: 16088095-16088095 on Build GRCh38
rs1003798356 Chr.19: 16088212-16088212 on Build GRCh38
rs879438508 Chr.19: 16088211-16088211 on Build GRCh38
rs774485804 Chr.19: 16087980-16087981 on Build GRCh38
rs114561872 Chr.19: 16087983-16087983 on Build GRCh38
rs905918941 Chr.19: 16088300-16088300 on Build GRCh38
rs754913516 Chr.19: 16088053-16088053 on Build GRCh38
rs370009172 Chr.19: 16088119-16088119 on Build GRCh38
rs752488156 Chr.19: 16088078-16088078 on Build GRCh38
rs747231292 Chr.19: 16088085-16088085 on Build GRCh38
rs1034790579 Chr.19: 16088217-16088217 on Build GRCh38
rs542893939 Chr.19: 16087948-16087948 on Build GRCh38
rs113423795 Chr.19: 16087969-16087969 on Build GRCh38
rs538902086 Chr.19: 16088079-16088079 on Build GRCh38
rs753766561 Chr.19: 16088039-16088039 on Build GRCh38
rs112747312 Chr.19: 16088098-16088098 on Build GRCh38
rs771571055 Chr.19: 16087988-16087988 on Build GRCh38
rs1030498508 Chr.19: 16087936-16087936 on Build GRCh38
rs1045813844 Chr.19: 16088285-16088285 on Build GRCh38
rs749938370 Chr.19: 16088006-16088006 on Build GRCh38
rs936207738 Chr.19: 16088150-16088150 on Build GRCh38
rs546470230 Chr.19: 16088137-16088137 on Build GRCh38
rs71334798 Chr.19: 16088074-16088074 on Build GRCh38
rs533128638 Chr.19: 16088122-16088122 on Build GRCh38
rs771195365 Chr.19: 16088104-16088104 on Build GRCh38
rs79700591 Chr.19: 16088179-16088179 on Build GRCh38
rs546839845 Chr.19: 16088308-16088308 on Build GRCh38
rs758146082 Chr.19: 16088344-16088344 on Build GRCh38
rs535626563 Chr.19: 16088167-16088167 on Build GRCh38
rs71334797 Chr.19: 16088072-16088072 on Build GRCh38
rs765574384 Chr.19: 16087996-16087996 on Build GRCh38
rs765105143 Chr.19: 16088049-16088049 on Build GRCh38
rs372712176 Chr.19: 16088083-16088083 on Build GRCh38
rs890175174 Chr.19: 16087937-16087937 on Build GRCh38
rs988841777 Chr.19: 16088299-16088299 on Build GRCh38
rs370753747 Chr.19: 16087999-16087999 on Build GRCh38
rs1043692580 Chr.19: 16088041-16088041 on Build GRCh38
rs201878921 Chr.19: 16088093-16088093 on Build GRCh38
rs772890544 Chr.19: 16087949-16087949 on Build GRCh38
rs913268098 Chr.19: 16088305-16088305 on Build GRCh38
rs960137679 Chr.19: 16088265-16088265 on Build GRCh38
rs988789645 Chr.19: 16088002-16088002 on Build GRCh38
rs533032636 Chr.19: 16088132-16088132 on Build GRCh38
rs371728238 Chr.19: 16088031-16088031 on Build GRCh38
rs765901581 Chr.19: 16087981-16087981 on Build GRCh38
rs969126760 Chr.19: 16088311-16088311 on Build GRCh38
rs376370608 Chr.19: 16087998-16087998 on Build GRCh38
rs1027417606 Chr.19: 16088337-16088337 on Build GRCh38
rs188115268 Chr.19: 16088145-16088145 on Build GRCh38
rs370683629 Chr.19: 16087984-16087984 on Build GRCh38
rs1023974814 Chr.19: 16088194-16088194 on Build GRCh38
rs548951390 Chr.19: 16088314-16088314 on Build GRCh38
rs748357243 Chr.19: 16087982-16087982 on Build GRCh38
rs142315721 Chr.19: 16087933-16087933 on Build GRCh38
rs374120813 Chr.19: 16088011-16088011 on Build GRCh38
rs530525344 Chr.19: 16087978-16087978 on Build GRCh38
rs760077627 Chr.19: 16087994-16087994 on Build GRCh38
rs761888380 Chr.19: 16088135-16088135 on Build GRCh38
rs367667507 Chr.19: 16088133-16088133 on Build GRCh38
rs200756013 Chr.19: 16088107-16088107 on Build GRCh38
rs757402835 Chr.19: 16088186-16088186 on Build GRCh38
rs978752950 Chr.19: 16088332-16088332 on Build GRCh38

View Full Product Details