Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 16577174-16577681 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MED26
Entrez Gene ID
9441
Gene Name
mediator complex subunit 26
Gene Aliases
CRSP7, CRSP70
UniGene
Hs.715040
Gene Chromosome Location
Chr.19: 16574907-16628204 on Build GRCh38
Transcripts
NM_004831.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4609731 c.577_578ins53 p.A193fs*5 Chr.19: 16577252-16577253 on Build GRCh38
6711000 c.576C>T p.F192F Chr.19: 16577254-16577254 on Build GRCh38
5651058 c.174C>T p.N58N Chr.19: 16577656-16577656 on Build GRCh38
228696 c.400C>T p.Q134* Chr.19: 16577430-16577430 on Build GRCh38
6212767 c.428G>A p.R143K Chr.19: 16577402-16577402 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755703214 Chr.19: 16577293-16577293 on Build GRCh38
rs781294294 Chr.19: 16577426-16577426 on Build GRCh38
rs760850314 Chr.19: 16577583-16577583 on Build GRCh38
rs765918261 Chr.19: 16577410-16577410 on Build GRCh38
rs369386204 Chr.19: 16577549-16577549 on Build GRCh38
rs202178044 Chr.19: 16577253-16577253 on Build GRCh38
rs761805169 Chr.19: 16577564-16577564 on Build GRCh38
rs748627855 Chr.19: 16577536-16577536 on Build GRCh38
rs540369553 Chr.19: 16577230-16577230 on Build GRCh38
rs375797169 Chr.19: 16577495-16577495 on Build GRCh38
rs754859528 Chr.19: 16577299-16577299 on Build GRCh38
rs752578360 Chr.19: 16577370-16577370 on Build GRCh38
rs755649730 Chr.19: 16577203-16577203 on Build GRCh38
rs200056042 Chr.19: 16577198-16577198 on Build GRCh38
rs368197948 Chr.19: 16577319-16577319 on Build GRCh38
rs576745640 Chr.19: 16577516-16577516 on Build GRCh38
rs543367189 Chr.19: 16577482-16577482 on Build GRCh38
rs200593026 Chr.19: 16577351-16577351 on Build GRCh38
rs763934151 Chr.19: 16577421-16577421 on Build GRCh38
rs771242857 Chr.19: 16577656-16577656 on Build GRCh38
rs958919483 Chr.19: 16577328-16577328 on Build GRCh38
rs750678868 Chr.19: 16577496-16577496 on Build GRCh38
rs773319730 Chr.19: 16577244-16577244 on Build GRCh38
rs778047964 Chr.19: 16577638-16577638 on Build GRCh38
rs754062611 Chr.19: 16577614-16577614 on Build GRCh38
rs776157495 Chr.19: 16577477-16577477 on Build GRCh38
rs372669311 Chr.19: 16577515-16577515 on Build GRCh38
rs367966430 Chr.19: 16577419-16577419 on Build GRCh38
rs767277361 Chr.19: 16577577-16577577 on Build GRCh38
rs759157826 Chr.19: 16577479-16577479 on Build GRCh38
rs756350916 Chr.19: 16577505-16577505 on Build GRCh38
rs910822942 Chr.19: 16577580-16577580 on Build GRCh38
rs897770888 Chr.19: 16577653-16577653 on Build GRCh38
rs772324321 Chr.19: 16577537-16577537 on Build GRCh38
rs190038139 Chr.19: 16577632-16577632 on Build GRCh38
rs769870130 Chr.19: 16577382-16577382 on Build GRCh38
rs1009194771 Chr.19: 16577453-16577453 on Build GRCh38
rs571147808 Chr.19: 16577391-16577391 on Build GRCh38
rs778569367 Chr.19: 16577221-16577221 on Build GRCh38
rs139528780 Chr.19: 16577578-16577578 on Build GRCh38
rs371027623 Chr.19: 16577320-16577320 on Build GRCh38
rs771812981 Chr.19: 16577316-16577316 on Build GRCh38
rs374700479 Chr.19: 16577281-16577281 on Build GRCh38
rs768503593 Chr.19: 16577219-16577219 on Build GRCh38
rs747864157 Chr.19: 16577303-16577303 on Build GRCh38
rs1019162749 Chr.19: 16577280-16577280 on Build GRCh38
rs748551829 Chr.19: 16577456-16577456 on Build GRCh38
rs760456340 Chr.19: 16577404-16577404 on Build GRCh38
rs868692980 Chr.19: 16577478-16577478 on Build GRCh38
rs199926336 Chr.19: 16577587-16577587 on Build GRCh38
rs770172101 Chr.19: 16577337-16577337 on Build GRCh38
rs760145084 Chr.19: 16577322-16577322 on Build GRCh38
rs779583130 Chr.19: 16577527-16577527 on Build GRCh38
rs777478207 Chr.19: 16577376-16577376 on Build GRCh38
rs749745511 Chr.19: 16577390-16577390 on Build GRCh38
rs757038907 Chr.19: 16577425-16577425 on Build GRCh38
rs745563556 Chr.19: 16577544-16577544 on Build GRCh38
rs776028210 Chr.19: 16577341-16577341 on Build GRCh38
rs746045701 Chr.19: 16577429-16577429 on Build GRCh38
rs567636032 Chr.19: 16577424-16577424 on Build GRCh38
rs777204502 Chr.19: 16577251-16577251 on Build GRCh38
rs763436711 Chr.19: 16577347-16577347 on Build GRCh38
rs779802540 Chr.19: 16577377-16577377 on Build GRCh38
rs375492411 Chr.19: 16577546-16577546 on Build GRCh38
rs779065125 Chr.19: 16577623-16577623 on Build GRCh38
rs371851199 Chr.19: 16577480-16577480 on Build GRCh38
rs548718062 Chr.19: 16577350-16577350 on Build GRCh38
rs202000537 Chr.19: 16577551-16577551 on Build GRCh38
rs781379441 Chr.19: 16577379-16577379 on Build GRCh38
rs146319284 Chr.19: 16577296-16577296 on Build GRCh38
rs764499531 Chr.19: 16577348-16577348 on Build GRCh38
rs757017468 Chr.19: 16577492-16577492 on Build GRCh38
rs951706626 Chr.19: 16577306-16577306 on Build GRCh38
rs765697215 Chr.19: 16577254-16577254 on Build GRCh38
rs980428416 Chr.19: 16577313-16577313 on Build GRCh38
rs778819363 Chr.19: 16577300-16577300 on Build GRCh38
rs528910232 Chr.19: 16577339-16577339 on Build GRCh38
rs139271259 Chr.19: 16577207-16577207 on Build GRCh38
rs756153294 Chr.19: 16577433-16577433 on Build GRCh38
rs752988166 Chr.19: 16577262-16577262 on Build GRCh38
rs775875361 Chr.19: 16577384-16577384 on Build GRCh38
rs531503971 Chr.19: 16577378-16577378 on Build GRCh38
rs772258425 Chr.19: 16577235-16577235 on Build GRCh38
rs774898510 Chr.19: 16577442-16577442 on Build GRCh38
rs777992148 Chr.19: 16577317-16577317 on Build GRCh38
rs750051834 Chr.19: 16577290-16577290 on Build GRCh38
rs551219804 Chr.19: 16577381-16577381 on Build GRCh38
rs758199176 Chr.19: 16577371-16577371 on Build GRCh38
rs112717816 Chr.19: 16577365-16577365 on Build GRCh38
rs774082179 Chr.19: 16577283-16577285 on Build GRCh38
rs769267106 Chr.19: 16577441-16577441 on Build GRCh38
rs533733191 Chr.19: 16577411-16577411 on Build GRCh38
rs936041032 Chr.19: 16577572-16577572 on Build GRCh38
rs765975570 Chr.19: 16577361-16577361 on Build GRCh38
rs768098000 Chr.19: 16577552-16577552 on Build GRCh38
rs200430258 Chr.19: 16577438-16577438 on Build GRCh38
rs145192786 Chr.19: 16577647-16577647 on Build GRCh38
rs766741681 Chr.19: 16577602-16577602 on Build GRCh38
rs201117259 Chr.19: 16577369-16577369 on Build GRCh38
rs774966415 Chr.19: 16577481-16577481 on Build GRCh38
rs745342341 Chr.19: 16577199-16577199 on Build GRCh38
rs764742324 Chr.19: 16577412-16577412 on Build GRCh38
rs778195628 Chr.19: 16577538-16577538 on Build GRCh38
rs760774365 Chr.19: 16577248-16577248 on Build GRCh38
rs1008835486 Chr.19: 16577270-16577270 on Build GRCh38
rs367848015 Chr.19: 16577509-16577509 on Build GRCh38
rs780334253 Chr.19: 16577508-16577508 on Build GRCh38
rs144231247 Chr.19: 16577617-16577617 on Build GRCh38
rs752635632 Chr.19: 16577418-16577418 on Build GRCh38
rs761716046 Chr.19: 16577513-16577524 on Build GRCh38
rs371433884 Chr.19: 16577264-16577264 on Build GRCh38
rs753268204 Chr.19: 16577362-16577362 on Build GRCh38
rs774277841 Chr.19: 16577554-16577554 on Build GRCh38
rs772429394 Chr.19: 16577466-16577466 on Build GRCh38
rs780281216 Chr.19: 16577436-16577436 on Build GRCh38
rs749122470 Chr.19: 16577208-16577208 on Build GRCh38
rs751451162 Chr.19: 16577491-16577491 on Build GRCh38

View Full Product Details