Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 17294512-17294913 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD8
Entrez Gene ID
79575
Gene Name
abhydrolase domain containing 8
Gene Aliases
-
UniGene
Hs.515664
Gene Chromosome Location
Chr.19: 17292131-17303473 on Build GRCh38
Transcripts
NM_024527.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5645181 c.882C>T p.P294P Chr.19: 17294725-17294725 on Build GRCh38
4632198 c.856A>G p.S286G Chr.19: 17294751-17294751 on Build GRCh38
4844505 c.873C>T p.F291F Chr.19: 17294734-17294734 on Build GRCh38
4075318 c.813G>T p.K271N Chr.19: 17294794-17294794 on Build GRCh38
4845514 c.771C>T p.F257F Chr.19: 17294836-17294836 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs956370634 Chr.19: 17294615-17294615 on Build GRCh38
rs368686278 Chr.19: 17294808-17294808 on Build GRCh38
rs770758079 Chr.19: 17294771-17294771 on Build GRCh38
rs745904932 Chr.19: 17294640-17294640 on Build GRCh38
rs780190388 Chr.19: 17294743-17294743 on Build GRCh38
rs983193074 Chr.19: 17294601-17294601 on Build GRCh38
rs376684191 Chr.19: 17294555-17294555 on Build GRCh38
rs760932860 Chr.19: 17294857-17294857 on Build GRCh38
rs764860195 Chr.19: 17294716-17294716 on Build GRCh38
rs564498276 Chr.19: 17294698-17294698 on Build GRCh38
rs150010983 Chr.19: 17294731-17294731 on Build GRCh38
rs756997082 Chr.19: 17294889-17294889 on Build GRCh38
rs929035128 Chr.19: 17294580-17294581 on Build GRCh38
rs780129673 Chr.19: 17294647-17294647 on Build GRCh38
rs767889101 Chr.19: 17294735-17294735 on Build GRCh38
rs760613065 Chr.19: 17294531-17294531 on Build GRCh38
rs896579823 Chr.19: 17294881-17294881 on Build GRCh38
rs753900162 Chr.19: 17294861-17294861 on Build GRCh38
rs200349196 Chr.19: 17294633-17294633 on Build GRCh38
rs528302809 Chr.19: 17294792-17294792 on Build GRCh38
rs145795042 Chr.19: 17294701-17294701 on Build GRCh38
rs766323396 Chr.19: 17294852-17294852 on Build GRCh38
rs758942089 Chr.19: 17294708-17294708 on Build GRCh38
rs981887148 Chr.19: 17294591-17294591 on Build GRCh38
rs1035061048 Chr.19: 17294807-17294807 on Build GRCh38
rs141675608 Chr.19: 17294793-17294793 on Build GRCh38
rs1030638552 Chr.19: 17294594-17294594 on Build GRCh38
rs752970908 Chr.19: 17294887-17294887 on Build GRCh38
rs537342843 Chr.19: 17294856-17294856 on Build GRCh38
rs371746284 Chr.19: 17294547-17294547 on Build GRCh38
rs771790599 Chr.19: 17294651-17294651 on Build GRCh38
rs764761766 Chr.19: 17294554-17294554 on Build GRCh38
rs368550001 Chr.19: 17294624-17294624 on Build GRCh38
rs77887041 Chr.19: 17294533-17294533 on Build GRCh38
rs756147430 Chr.19: 17294740-17294740 on Build GRCh38
rs753308937 Chr.19: 17294536-17294536 on Build GRCh38
rs1039187307 Chr.19: 17294667-17294667 on Build GRCh38
rs544329143 Chr.19: 17294602-17294602 on Build GRCh38
rs755088688 Chr.19: 17294857-17294857 on Build GRCh38
rs769117374 Chr.19: 17294791-17294791 on Build GRCh38
rs752006334 Chr.19: 17294730-17294730 on Build GRCh38
rs766719261 Chr.19: 17294860-17294861 on Build GRCh38
rs746897140 Chr.19: 17294661-17294661 on Build GRCh38
rs750832474 Chr.19: 17294824-17294824 on Build GRCh38
rs761261189 Chr.19: 17294842-17294842 on Build GRCh38
rs117124277 Chr.19: 17294803-17294803 on Build GRCh38
rs562055541 Chr.19: 17294560-17294560 on Build GRCh38
rs369571526 Chr.19: 17294710-17294710 on Build GRCh38
rs775000128 Chr.19: 17294718-17294718 on Build GRCh38
rs531189672 Chr.19: 17294563-17294563 on Build GRCh38
rs767996580 Chr.19: 17294811-17294811 on Build GRCh38
rs758822540 Chr.19: 17294556-17294556 on Build GRCh38
rs756267143 Chr.19: 17294641-17294641 on Build GRCh38
rs762698564 Chr.19: 17294719-17294719 on Build GRCh38
rs556716539 Chr.19: 17294873-17294873 on Build GRCh38
rs375423667 Chr.19: 17294703-17294703 on Build GRCh38
rs1047150851 Chr.19: 17294767-17294767 on Build GRCh38
rs781293459 Chr.19: 17294638-17294638 on Build GRCh38
rs762255829 Chr.19: 17294810-17294810 on Build GRCh38
rs745884795 Chr.19: 17294786-17294786 on Build GRCh38
rs764493398 Chr.19: 17294720-17294720 on Build GRCh38
rs749456670 Chr.19: 17294755-17294755 on Build GRCh38
rs750996957 Chr.19: 17294739-17294739 on Build GRCh38
rs746772447 Chr.19: 17294770-17294770 on Build GRCh38
rs200026977 Chr.19: 17294761-17294761 on Build GRCh38
rs749594333 Chr.19: 17294650-17294650 on Build GRCh38
rs776416280 Chr.19: 17294776-17294776 on Build GRCh38
rs777500896 Chr.19: 17294764-17294764 on Build GRCh38

View Full Product Details