Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 17605674-17606193 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UNC13A
Entrez Gene ID
23025
Gene Name
unc-13 homolog A
Gene Aliases
Munc13-1
UniGene
Hs.164502
Gene Chromosome Location
Chr.19: 17601328-17688344 on Build GRCh38
Transcripts
NM_001080421.2 XM_011527810.2 XM_011527811.2 XM_017026502.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4824689 c.5013C>G p.I1671M Chr.19: 17606153-17606153 on Build GRCh38
4824690 c.5013C>G p.I1671M Chr.19: 17606153-17606153 on Build GRCh38
5638184 c.5110T>C p.*1704Q Chr.19: 17606056-17606056 on Build GRCh38
5143267 c.5303T>C p.V1768A Chr.19: 17606127-17606127 on Build GRCh38
5638185 c.5110T>C p.*1704Q Chr.19: 17606056-17606056 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs959739434 Chr.19: 17605947-17605948 on Build GRCh38
rs756728476 Chr.19: 17605850-17605850 on Build GRCh38
rs971857991 Chr.19: 17605795-17605795 on Build GRCh38
rs757391788 Chr.19: 17606044-17606071 on Build GRCh38
rs755963373 Chr.19: 17606052-17606052 on Build GRCh38
rs904041977 Chr.19: 17605935-17605935 on Build GRCh38
rs7254755 Chr.19: 17605773-17605773 on Build GRCh38
rs951939949 Chr.19: 17605832-17605832 on Build GRCh38
rs565905132 Chr.19: 17606041-17606041 on Build GRCh38
rs768424710 Chr.19: 17606054-17606054 on Build GRCh38
rs12974304 Chr.19: 17605985-17605985 on Build GRCh38
rs993095382 Chr.19: 17605761-17605761 on Build GRCh38
rs772836083 Chr.19: 17606063-17606063 on Build GRCh38
rs868352249 Chr.19: 17606142-17606142 on Build GRCh38
rs771122749 Chr.19: 17605734-17605734 on Build GRCh38
rs780691845 Chr.19: 17605900-17605900 on Build GRCh38
rs754854349 Chr.19: 17606105-17606105 on Build GRCh38
rs890020457 Chr.19: 17606132-17606132 on Build GRCh38
rs929436088 Chr.19: 17605802-17605802 on Build GRCh38
rs766787115 Chr.19: 17606008-17606008 on Build GRCh38
rs529855318 Chr.19: 17605869-17605869 on Build GRCh38
rs372747454 Chr.19: 17606068-17606068 on Build GRCh38
rs771677125 Chr.19: 17606061-17606061 on Build GRCh38
rs763447275 Chr.19: 17606162-17606162 on Build GRCh38
rs1048185459 Chr.19: 17605804-17605804 on Build GRCh38
rs897212567 Chr.19: 17605996-17605996 on Build GRCh38
rs926275170 Chr.19: 17605859-17605859 on Build GRCh38
rs758033324 Chr.19: 17605768-17605768 on Build GRCh38
rs1022665698 Chr.19: 17606086-17606086 on Build GRCh38
rs1019639601 Chr.19: 17605986-17605986 on Build GRCh38
rs1008524024 Chr.19: 17605813-17605813 on Build GRCh38
rs1001143320 Chr.19: 17605728-17605728 on Build GRCh38
rs916182319 Chr.19: 17605984-17605984 on Build GRCh38
rs752671390 Chr.19: 17606042-17606042 on Build GRCh38
rs984686024 Chr.19: 17605840-17605840 on Build GRCh38
rs944325169 Chr.19: 17606156-17606156 on Build GRCh38
rs372458664 Chr.19: 17606094-17606094 on Build GRCh38
rs764297759 Chr.19: 17606084-17606084 on Build GRCh38
rs747304890 Chr.19: 17606048-17606048 on Build GRCh38
rs1028306712 Chr.19: 17605736-17605736 on Build GRCh38
rs936871633 Chr.19: 17605948-17605948 on Build GRCh38
rs747578104 Chr.19: 17606058-17606058 on Build GRCh38
rs924332272 Chr.19: 17605719-17605719 on Build GRCh38
rs1042564578 Chr.19: 17605932-17605932 on Build GRCh38
rs376327069 Chr.19: 17606099-17606099 on Build GRCh38
rs545293000 Chr.19: 17605700-17605700 on Build GRCh38
rs774392878 Chr.19: 17606057-17606057 on Build GRCh38
rs746545284 Chr.19: 17606138-17606138 on Build GRCh38
rs571325205 Chr.19: 17605949-17605949 on Build GRCh38
rs775853127 Chr.19: 17606150-17606150 on Build GRCh38
rs3746198 Chr.19: 17606087-17606087 on Build GRCh38
rs552847488 Chr.19: 17605997-17605997 on Build GRCh38
rs1026075064 Chr.19: 17606012-17606012 on Build GRCh38
rs937678282 Chr.19: 17605864-17605864 on Build GRCh38
rs945137788 Chr.19: 17605925-17605925 on Build GRCh38
rs549506604 Chr.19: 17605890-17605890 on Build GRCh38
rs201314772 Chr.19: 17605814-17605814 on Build GRCh38
rs765805519 Chr.19: 17606035-17606035 on Build GRCh38
rs778704405 Chr.19: 17606107-17606107 on Build GRCh38
rs1047811841 Chr.19: 17606006-17606006 on Build GRCh38
rs771909449 Chr.19: 17606125-17606125 on Build GRCh38
rs890082144 Chr.19: 17605961-17605961 on Build GRCh38
rs777207198 Chr.19: 17606136-17606136 on Build GRCh38
rs951890492 Chr.19: 17606013-17606013 on Build GRCh38
rs959352988 Chr.19: 17606043-17606043 on Build GRCh38
rs977475744 Chr.19: 17605701-17605701 on Build GRCh38
rs754214589 Chr.19: 17606019-17606019 on Build GRCh38
rs770250093 Chr.19: 17606143-17606143 on Build GRCh38
rs374274380 Chr.19: 17605983-17605983 on Build GRCh38
rs527499410 Chr.19: 17605769-17605769 on Build GRCh38
rs554779097 Chr.19: 17606075-17606075 on Build GRCh38
rs920796657 Chr.19: 17605992-17605992 on Build GRCh38
rs1021439830 Chr.19: 17605867-17605867 on Build GRCh38
rs751653585 Chr.19: 17606171-17606171 on Build GRCh38
rs950922481 Chr.19: 17606165-17606165 on Build GRCh38
rs779941342 Chr.19: 17606053-17606053 on Build GRCh38
rs557029134 Chr.19: 17606141-17606141 on Build GRCh38
rs758556863 Chr.19: 17606044-17606044 on Build GRCh38
rs899648496 Chr.19: 17605827-17605827 on Build GRCh38
rs1056173696 Chr.19: 17605870-17605870 on Build GRCh38
rs766023950 Chr.19: 17606066-17606066 on Build GRCh38
rs758392831 Chr.19: 17606076-17606076 on Build GRCh38
rs753658542 Chr.19: 17606102-17606102 on Build GRCh38
rs560948878 Chr.19: 17605805-17605805 on Build GRCh38
rs760261470 Chr.19: 17606064-17606064 on Build GRCh38
rs755411745 Chr.19: 17606033-17606033 on Build GRCh38
rs532653529 Chr.19: 17605991-17605991 on Build GRCh38
rs374975843 Chr.19: 17606074-17606074 on Build GRCh38
rs189794656 Chr.19: 17605749-17605749 on Build GRCh38
rs748184528 Chr.19: 17606120-17606120 on Build GRCh38
rs961516237 Chr.19: 17605782-17605782 on Build GRCh38

View Full Product Details