Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18743985-18744480 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRTC1
Entrez Gene ID
23373
Gene Name
CREB regulated transcription coactivator 1
Gene Aliases
MECT1, TORC-1, TORC1, WAMTP1
UniGene
Hs.371096
Gene Chromosome Location
Chr.19: 18683615-18782333 on Build GRCh38
Transcripts
NM_001098482.1 NM_015321.2 XM_005259833.3 XM_005259834.1 XM_005259835.3 XM_005259836.3 XM_006722710.3 XM_011527842.2 XM_017026536.1 XM_017026537.1 XM_017026538.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
993135 c.250G>A p.G84R Chr.19: 18744114-18744114 on Build GRCh38
6635770 c.271G>A p.A91T Chr.19: 18744135-18744135 on Build GRCh38
6839866 c.256C>A p.L86M Chr.19: 18744120-18744120 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs778357661 Chr.19: 18744187-18744187 on Build GRCh38
rs765892132 Chr.19: 18744068-18744068 on Build GRCh38
rs561984417 Chr.19: 18744006-18744006 on Build GRCh38
rs781595990 Chr.19: 18744192-18744192 on Build GRCh38
rs946963359 Chr.19: 18744195-18744195 on Build GRCh38
rs915815753 Chr.19: 18744113-18744113 on Build GRCh38
rs148153617 Chr.19: 18744253-18744253 on Build GRCh38
rs969125283 Chr.19: 18744025-18744025 on Build GRCh38
rs1028003480 Chr.19: 18744344-18744344 on Build GRCh38
rs913923170 Chr.19: 18744194-18744194 on Build GRCh38
rs944497392 Chr.19: 18744355-18744355 on Build GRCh38
rs185045652 Chr.19: 18744007-18744007 on Build GRCh38
rs778482371 Chr.19: 18744114-18744114 on Build GRCh38
rs761712603 Chr.19: 18744059-18744059 on Build GRCh38
rs913882600 Chr.19: 18744339-18744339 on Build GRCh38
rs996526585 Chr.19: 18744028-18744028 on Build GRCh38
rs746121895 Chr.19: 18744132-18744132 on Build GRCh38
rs746255596 Chr.19: 18744200-18744200 on Build GRCh38
rs932943164 Chr.19: 18744407-18744407 on Build GRCh38
rs373531190 Chr.19: 18744181-18744181 on Build GRCh38
rs1022767029 Chr.19: 18744323-18744323 on Build GRCh38
rs187261114 Chr.19: 18744043-18744043 on Build GRCh38
rs900026134 Chr.19: 18744027-18744027 on Build GRCh38
rs548405073 Chr.19: 18744279-18744279 on Build GRCh38
rs996977447 Chr.19: 18744215-18744215 on Build GRCh38
rs767478273 Chr.19: 18744061-18744061 on Build GRCh38
rs752305069 Chr.19: 18744090-18744090 on Build GRCh38
rs35421095 Chr.19: 18744027-18744028 on Build GRCh38
rs1043862852 Chr.19: 18744013-18744013 on Build GRCh38
rs199565562 Chr.19: 18744169-18744169 on Build GRCh38
rs1051236146 Chr.19: 18744217-18744217 on Build GRCh38
rs759205595 Chr.19: 18744182-18744182 on Build GRCh38
rs775512684 Chr.19: 18744140-18744140 on Build GRCh38
rs970041952 Chr.19: 18744148-18744149 on Build GRCh38
rs748745158 Chr.19: 18744413-18744413 on Build GRCh38
rs369756605 Chr.19: 18744091-18744091 on Build GRCh38
rs756999552 Chr.19: 18744049-18744049 on Build GRCh38
rs1022896340 Chr.19: 18744265-18744265 on Build GRCh38
rs899969077 Chr.19: 18744212-18744212 on Build GRCh38
rs868552200 Chr.19: 18744258-18744258 on Build GRCh38
rs1043919840 Chr.19: 18744361-18744361 on Build GRCh38
rs970306286 Chr.19: 18744327-18744327 on Build GRCh38
rs911666597 Chr.19: 18744224-18744224 on Build GRCh38
rs773149852 Chr.19: 18744165-18744165 on Build GRCh38
rs537391002 Chr.19: 18744328-18744328 on Build GRCh38
rs1029366934 Chr.19: 18744029-18744029 on Build GRCh38
rs895930973 Chr.19: 18744296-18744296 on Build GRCh38
rs988646500 Chr.19: 18744332-18744332 on Build GRCh38
rs1011600448 Chr.19: 18744009-18744009 on Build GRCh38
rs552391738 Chr.19: 18744228-18744228 on Build GRCh38
rs752138187 Chr.19: 18744190-18744190 on Build GRCh38
rs1043287578 Chr.19: 18744406-18744406 on Build GRCh38
rs769955090 Chr.19: 18744133-18744133 on Build GRCh38
rs557230113 Chr.19: 18744350-18744350 on Build GRCh38
rs376724061 Chr.19: 18744180-18744180 on Build GRCh38
rs369676089 Chr.19: 18744186-18744186 on Build GRCh38
rs765022815 Chr.19: 18744185-18744185 on Build GRCh38
rs570357542 Chr.19: 18744352-18744352 on Build GRCh38
rs932826266 Chr.19: 18744404-18744404 on Build GRCh38
rs1021666317 Chr.19: 18744014-18744014 on Build GRCh38
rs771988521 Chr.19: 18744155-18744155 on Build GRCh38
rs1015246727 Chr.19: 18744229-18744229 on Build GRCh38
rs200526787 Chr.19: 18744089-18744089 on Build GRCh38
rs563711316 Chr.19: 18744129-18744129 on Build GRCh38
rs747319119 Chr.19: 18744118-18744118 on Build GRCh38
rs771329642 Chr.19: 18744122-18744122 on Build GRCh38
rs763082203 Chr.19: 18744153-18744153 on Build GRCh38
rs969998279 Chr.19: 18744266-18744266 on Build GRCh38
rs969881959 Chr.19: 18744183-18744183 on Build GRCh38
rs757513143 Chr.19: 18744191-18744191 on Build GRCh38
rs779673983 Chr.19: 18744396-18744396 on Build GRCh38
rs1041231549 Chr.19: 18744231-18744231 on Build GRCh38
rs921507678 Chr.19: 18744366-18744366 on Build GRCh38
rs750219074 Chr.19: 18744064-18744064 on Build GRCh38
rs1043709314 Chr.19: 18744257-18744257 on Build GRCh38
rs760580508 Chr.19: 18744066-18744066 on Build GRCh38
rs191630786 Chr.19: 18744390-18744390 on Build GRCh38
rs890875860 Chr.19: 18744223-18744223 on Build GRCh38
rs900079897 Chr.19: 18744232-18744232 on Build GRCh38
rs528704414 Chr.19: 18744267-18744267 on Build GRCh38
rs754407799 Chr.19: 18744077-18744077 on Build GRCh38
rs753536314 Chr.19: 18744075-18744075 on Build GRCh38
rs1003872946 Chr.19: 18744057-18744057 on Build GRCh38
rs1014095502 Chr.19: 18744309-18744309 on Build GRCh38
rs568301556 Chr.19: 18744295-18744295 on Build GRCh38
rs538346056 Chr.19: 18744292-18744292 on Build GRCh38
rs769995235 Chr.19: 18744204-18744204 on Build GRCh38
rs983108869 Chr.19: 18744351-18744351 on Build GRCh38
rs530854652 Chr.19: 18744039-18744039 on Build GRCh38

View Full Product Details