Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 19878129-19878624 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF253
Entrez Gene ID
56242
Gene Name
zinc finger protein 253
Gene Aliases
BMZF-1, BMZF1, ZNF411
UniGene
Hs.301059
Gene Chromosome Location
Chr.19: 19865905-19893484 on Build GRCh38
Transcripts
NM_021047.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5855171 c.121G>A p.V41I Chr.19: 19878598-19878598 on Build GRCh38
5855172 c.121G>A p.V41I Chr.19: 19878598-19878598 on Build GRCh38
286772 c.34G>T p.E12* Chr.19: 19878511-19878511 on Build GRCh38
6264369 c.130+8C>T p.? Chr.19: 19878615-19878615 on Build GRCh38
6291524 c.118T>C p.L40L Chr.19: 19878595-19878595 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775594523 Chr.19: 19878460-19878461 on Build GRCh38
rs753613453 Chr.19: 19878553-19878553 on Build GRCh38
rs763061966 Chr.19: 19878505-19878505 on Build GRCh38
rs567707425 Chr.19: 19878200-19878200 on Build GRCh38
rs778448236 Chr.19: 19878440-19878440 on Build GRCh38
rs190693250 Chr.19: 19878337-19878337 on Build GRCh38
rs10406883 Chr.19: 19878486-19878486 on Build GRCh38
rs1026275824 Chr.19: 19878272-19878272 on Build GRCh38
rs1016150009 Chr.19: 19878260-19878260 on Build GRCh38
rs574877794 Chr.19: 19878463-19878463 on Build GRCh38
rs398101123 Chr.19: 19878317-19878317 on Build GRCh38
rs1038484200 Chr.19: 19878162-19878162 on Build GRCh38
rs549999132 Chr.19: 19878291-19878291 on Build GRCh38
rs564010866 Chr.19: 19878587-19878587 on Build GRCh38
rs757945607 Chr.19: 19878575-19878575 on Build GRCh38
rs112610611 Chr.19: 19878458-19878459 on Build GRCh38
rs771452536 Chr.19: 19878470-19878470 on Build GRCh38
rs964368973 Chr.19: 19878296-19878296 on Build GRCh38
rs915004483 Chr.19: 19878331-19878331 on Build GRCh38
rs748940996 Chr.19: 19878436-19878436 on Build GRCh38
rs770185159 Chr.19: 19878459-19878460 on Build GRCh38
rs928149469 Chr.19: 19878439-19878439 on Build GRCh38
rs372446403 Chr.19: 19878559-19878559 on Build GRCh38
rs754689149 Chr.19: 19878561-19878561 on Build GRCh38
rs948246485 Chr.19: 19878413-19878413 on Build GRCh38
rs1006288948 Chr.19: 19878258-19878258 on Build GRCh38
rs916563956 Chr.19: 19878473-19878473 on Build GRCh38
rs755484158 Chr.19: 19878554-19878554 on Build GRCh38
rs3841054 Chr.19: 19878316-19878316 on Build GRCh38
rs886160364 Chr.19: 19878485-19878485 on Build GRCh38
rs867845559 Chr.19: 19878267-19878267 on Build GRCh38
rs766171578 Chr.19: 19878544-19878544 on Build GRCh38
rs977950340 Chr.19: 19878345-19878345 on Build GRCh38
rs190030835 Chr.19: 19878487-19878487 on Build GRCh38
rs773746571 Chr.19: 19878460-19878460 on Build GRCh38
rs533681486 Chr.19: 19878292-19878292 on Build GRCh38
rs974440344 Chr.19: 19878313-19878313 on Build GRCh38
rs767406118 Chr.19: 19878535-19878535 on Build GRCh38
rs898583222 Chr.19: 19878517-19878517 on Build GRCh38
rs775503342 Chr.19: 19878501-19878501 on Build GRCh38
rs1046606571 Chr.19: 19878427-19878427 on Build GRCh38
rs771509649 Chr.19: 19878590-19878590 on Build GRCh38
rs747660045 Chr.19: 19878572-19878572 on Build GRCh38
rs779779250 Chr.19: 19878434-19878434 on Build GRCh38
rs978240673 Chr.19: 19878307-19878307 on Build GRCh38
rs778596289 Chr.19: 19878567-19878567 on Build GRCh38
rs923805722 Chr.19: 19878375-19878375 on Build GRCh38
rs530237782 Chr.19: 19878242-19878242 on Build GRCh38
rs750399985 Chr.19: 19878536-19878536 on Build GRCh38
rs185435335 Chr.19: 19878293-19878293 on Build GRCh38
rs777193750 Chr.19: 19878599-19878599 on Build GRCh38
rs995971111 Chr.19: 19878277-19878277 on Build GRCh38
rs747556846 Chr.19: 19878576-19878576 on Build GRCh38
rs778036763 Chr.19: 19878576-19878577 on Build GRCh38
rs566090712 Chr.19: 19878441-19878441 on Build GRCh38
rs754564655 Chr.19: 19878437-19878437 on Build GRCh38
rs989451406 Chr.19: 19878339-19878339 on Build GRCh38
rs185323620 Chr.19: 19878459-19878459 on Build GRCh38
rs765542664 Chr.19: 19878488-19878488 on Build GRCh38
rs900434925 Chr.19: 19878491-19878491 on Build GRCh38
rs60758402 Chr.19: 19878473-19878474 on Build GRCh38
rs886143325 Chr.19: 19878253-19878253 on Build GRCh38
rs755975940 Chr.19: 19878538-19878538 on Build GRCh38
rs180711659 Chr.19: 19878445-19878445 on Build GRCh38

View Full Product Details