Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 20542943-20543453 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF737
Entrez Gene ID
100129842
Gene Name
zinc finger protein 737
Gene Aliases
ZNF102
UniGene
Hs.515696
Gene Chromosome Location
Chr.19: 20531230-20565821 on Build GRCh38
Transcripts
NM_001159293.1 XM_005259697.3 XM_006722597.3 XM_011527598.2 XM_011527599.2 XM_017026116.1 XM_017026117.1

SNP Detail

SNP ID Chromosome Location
rs551063123 Chr.19: 20543168-20543168 on Build GRCh38
rs34996698 Chr.19: 20543412-20543413 on Build GRCh38
rs7249742 Chr.19: 20543001-20543001 on Build GRCh38
rs146092302 Chr.19: 20543204-20543205 on Build GRCh38
rs188738542 Chr.19: 20543065-20543065 on Build GRCh38
rs201461779 Chr.19: 20543202-20543202 on Build GRCh38
rs1032119125 Chr.19: 20543095-20543095 on Build GRCh38
rs754578280 Chr.19: 20543260-20543260 on Build GRCh38
rs926895674 Chr.19: 20543181-20543181 on Build GRCh38
rs782450692 Chr.19: 20543269-20543269 on Build GRCh38
rs923045205 Chr.19: 20543253-20543253 on Build GRCh38
rs781847664 Chr.19: 20543343-20543343 on Build GRCh38
rs7250879 Chr.19: 20543173-20543173 on Build GRCh38
rs377391439 Chr.19: 20543205-20543206 on Build GRCh38
rs1000684851 Chr.19: 20543066-20543066 on Build GRCh38
rs138645537 Chr.19: 20543132-20543132 on Build GRCh38
rs185428336 Chr.19: 20543051-20543051 on Build GRCh38
rs984346965 Chr.19: 20543206-20543206 on Build GRCh38
rs782758742 Chr.19: 20543124-20543124 on Build GRCh38
rs577474228 Chr.19: 20543284-20543284 on Build GRCh38
rs926893348 Chr.19: 20542980-20542980 on Build GRCh38
rs1009316605 Chr.19: 20543399-20543399 on Build GRCh38
rs1053603622 Chr.19: 20543354-20543354 on Build GRCh38
rs781791215 Chr.19: 20543236-20543236 on Build GRCh38
rs181083170 Chr.19: 20543228-20543228 on Build GRCh38
rs75325097 Chr.19: 20543213-20543213 on Build GRCh38
rs1036087014 Chr.19: 20543021-20543021 on Build GRCh38
rs940441915 Chr.19: 20543005-20543005 on Build GRCh38
rs141146794 Chr.19: 20543002-20543002 on Build GRCh38
rs144909492 Chr.19: 20543061-20543061 on Build GRCh38
rs780990762 Chr.19: 20543391-20543394 on Build GRCh38
rs10410284 Chr.19: 20543027-20543027 on Build GRCh38
rs931504715 Chr.19: 20542987-20542987 on Build GRCh38
rs1025025490 Chr.19: 20543153-20543153 on Build GRCh38
rs189949731 Chr.19: 20543325-20543325 on Build GRCh38
rs188201094 Chr.19: 20543220-20543220 on Build GRCh38
rs200295916 Chr.19: 20543204-20543204 on Build GRCh38
rs184855071 Chr.19: 20543281-20543281 on Build GRCh38
rs199817753 Chr.19: 20543157-20543157 on Build GRCh38
rs142458524 Chr.19: 20543176-20543176 on Build GRCh38
rs892309053 Chr.19: 20543369-20543369 on Build GRCh38
rs7249873 Chr.19: 20543157-20543157 on Build GRCh38
rs33930 Chr.19: 20543062-20543062 on Build GRCh38
rs57449077 Chr.19: 20543205-20543205 on Build GRCh38
rs544734594 Chr.19: 20543342-20543342 on Build GRCh38

View Full Product Details