Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 21426033-21426540 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF493
Entrez Gene ID
284443
Gene Name
zinc finger protein 493
Gene Aliases
-
UniGene
Hs.656558
Gene Chromosome Location
Chr.19: 21397119-21427495 on Build GRCh38
Transcripts
NM_001076678.2 NM_175910.6

SNP Detail

SNP ID Chromosome Location
rs943512217 Chr.19: 21426066-21426066 on Build GRCh38
rs11878436 Chr.19: 21426098-21426098 on Build GRCh38
rs1055804719 Chr.19: 21426410-21426410 on Build GRCh38
rs938742468 Chr.19: 21426392-21426392 on Build GRCh38
rs568780261 Chr.19: 21426343-21426343 on Build GRCh38
rs975311004 Chr.19: 21426382-21426382 on Build GRCh38
rs532130025 Chr.19: 21426227-21426227 on Build GRCh38
rs374100427 Chr.19: 21426267-21426267 on Build GRCh38
rs745684499 Chr.19: 21426424-21426424 on Build GRCh38
rs990365967 Chr.19: 21426306-21426306 on Build GRCh38
rs753877932 Chr.19: 21426239-21426239 on Build GRCh38
rs1025868336 Chr.19: 21426495-21426495 on Build GRCh38
rs866907931 Chr.19: 21426183-21426183 on Build GRCh38
rs112557514 Chr.19: 21426118-21426118 on Build GRCh38
rs772172254 Chr.19: 21426219-21426219 on Build GRCh38
rs1012082308 Chr.19: 21426073-21426073 on Build GRCh38
rs780382373 Chr.19: 21426510-21426510 on Build GRCh38
rs946221200 Chr.19: 21426322-21426322 on Build GRCh38
rs902152462 Chr.19: 21426158-21426161 on Build GRCh38
rs780170819 Chr.19: 21426220-21426220 on Build GRCh38
rs971469865 Chr.19: 21426212-21426212 on Build GRCh38
rs923056229 Chr.19: 21426429-21426429 on Build GRCh38
rs923003722 Chr.19: 21426112-21426112 on Build GRCh38
rs144922029 Chr.19: 21426158-21426162 on Build GRCh38
rs185870636 Chr.19: 21426117-21426117 on Build GRCh38
rs894991531 Chr.19: 21426189-21426189 on Build GRCh38
rs757286134 Chr.19: 21426301-21426301 on Build GRCh38
rs928750292 Chr.19: 21426391-21426391 on Build GRCh38
rs955166958 Chr.19: 21426503-21426503 on Build GRCh38
rs1016733093 Chr.19: 21426075-21426075 on Build GRCh38
rs772128148 Chr.19: 21426493-21426493 on Build GRCh38
rs758040119 Chr.19: 21426206-21426207 on Build GRCh38
rs745344373 Chr.19: 21426174-21426174 on Build GRCh38
rs1023073707 Chr.19: 21426193-21426193 on Build GRCh38
rs1038798588 Chr.19: 21426090-21426090 on Build GRCh38
rs534238001 Chr.19: 21426472-21426472 on Build GRCh38
rs1054087929 Chr.19: 21426170-21426170 on Build GRCh38
rs184193003 Chr.19: 21426452-21426452 on Build GRCh38
rs867874987 Chr.19: 21426492-21426492 on Build GRCh38
rs914819201 Chr.19: 21426317-21426317 on Build GRCh38
rs371298986 Chr.19: 21426161-21426165 on Build GRCh38
rs1038292201 Chr.19: 21426485-21426485 on Build GRCh38
rs548489125 Chr.19: 21426441-21426441 on Build GRCh38
rs76902667 Chr.19: 21426411-21426411 on Build GRCh38
rs964291785 Chr.19: 21426275-21426275 on Build GRCh38
rs113355038 Chr.19: 21426371-21426371 on Build GRCh38
rs191854891 Chr.19: 21426271-21426271 on Build GRCh38
rs1012480087 Chr.19: 21426192-21426192 on Build GRCh38
rs778815424 Chr.19: 21426310-21426310 on Build GRCh38
rs528136369 Chr.19: 21426059-21426059 on Build GRCh38

View Full Product Details