Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 29529598-29530106 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VSTM2B
Entrez Gene ID
342865
Gene Name
V-set and transmembrane domain containing 2B
Gene Aliases
-
UniGene
Hs.451618
Gene Chromosome Location
Chr.19: 29525431-29564555 on Build GRCh38
Transcripts
NM_001146339.1 XM_005258856.3 XM_011526902.2 XM_011526903.2 XM_011526904.2 XM_011526905.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6416317 c.406G>T p.D136Y Chr.19: 29529927-29529927 on Build GRCh38
1612010 c.575C>A p.T192N Chr.19: 29530096-29530096 on Build GRCh38
5430560 c.461C>T p.P154L Chr.19: 29529982-29529982 on Build GRCh38
1392382 c.406G>A p.D136N Chr.19: 29529927-29529927 on Build GRCh38
5190486 c.390G>A p.S130S Chr.19: 29529911-29529911 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760404780 Chr.19: 29529619-29529619 on Build GRCh38
rs773973987 Chr.19: 29529821-29529821 on Build GRCh38
rs909083162 Chr.19: 29529909-29529909 on Build GRCh38
rs895291463 Chr.19: 29529779-29529779 on Build GRCh38
rs377294454 Chr.19: 29529853-29529853 on Build GRCh38
rs541615528 Chr.19: 29529743-29529743 on Build GRCh38
rs985010156 Chr.19: 29529908-29529908 on Build GRCh38
rs572517670 Chr.19: 29529740-29529740 on Build GRCh38
rs773053753 Chr.19: 29529998-29529998 on Build GRCh38
rs751790250 Chr.19: 29529884-29529884 on Build GRCh38
rs901314473 Chr.19: 29529737-29529737 on Build GRCh38
rs776400235 Chr.19: 29530031-29530031 on Build GRCh38
rs547562401 Chr.19: 29529968-29529968 on Build GRCh38
rs754418111 Chr.19: 29529966-29529966 on Build GRCh38
rs1034680204 Chr.19: 29529899-29529899 on Build GRCh38
rs750884925 Chr.19: 29530085-29530085 on Build GRCh38
rs775005192 Chr.19: 29530033-29530033 on Build GRCh38
rs561770422 Chr.19: 29529860-29529860 on Build GRCh38
rs1022486040 Chr.19: 29529804-29529804 on Build GRCh38
rs770949464 Chr.19: 29530029-29530029 on Build GRCh38
rs909973669 Chr.19: 29529670-29529670 on Build GRCh38
rs771802826 Chr.19: 29529825-29529825 on Build GRCh38
rs748881464 Chr.19: 29529962-29529962 on Build GRCh38
rs984095511 Chr.19: 29529902-29529902 on Build GRCh38
rs747738641 Chr.19: 29529970-29529970 on Build GRCh38
rs369065882 Chr.19: 29529822-29529822 on Build GRCh38
rs940075693 Chr.19: 29529720-29529720 on Build GRCh38
rs765667340 Chr.19: 29530056-29530056 on Build GRCh38
rs533404730 Chr.19: 29529886-29529886 on Build GRCh38
rs773143728 Chr.19: 29529826-29529826 on Build GRCh38
rs527457087 Chr.19: 29529885-29529885 on Build GRCh38
rs184979600 Chr.19: 29529768-29529768 on Build GRCh38
rs973209969 Chr.19: 29529967-29529967 on Build GRCh38
rs922824686 Chr.19: 29529980-29529980 on Build GRCh38
rs780416543 Chr.19: 29529931-29529931 on Build GRCh38
rs760194183 Chr.19: 29530044-29530044 on Build GRCh38
rs771855777 Chr.19: 29529991-29529991 on Build GRCh38
rs61731469 Chr.19: 29529845-29529845 on Build GRCh38
rs951926793 Chr.19: 29529901-29529901 on Build GRCh38
rs1032069835 Chr.19: 29530084-29530084 on Build GRCh38
rs767937037 Chr.19: 29529890-29529890 on Build GRCh38
rs748743395 Chr.19: 29529793-29529793 on Build GRCh38
rs760590847 Chr.19: 29529834-29529834 on Build GRCh38
rs938007575 Chr.19: 29529636-29529636 on Build GRCh38
rs749710750 Chr.19: 29529662-29529662 on Build GRCh38
rs1037216292 Chr.19: 29529734-29529734 on Build GRCh38
rs966039137 Chr.19: 29529874-29529874 on Build GRCh38
rs931820474 Chr.19: 29529664-29529664 on Build GRCh38
rs35700860 Chr.19: 29529771-29529771 on Build GRCh38
rs746762719 Chr.19: 29530024-29530024 on Build GRCh38
rs1010015789 Chr.19: 29530063-29530063 on Build GRCh38
rs756711400 Chr.19: 29529920-29529920 on Build GRCh38
rs562453487 Chr.19: 29529774-29529774 on Build GRCh38
rs934285512 Chr.19: 29529739-29529739 on Build GRCh38
rs1036435750 Chr.19: 29529751-29529751 on Build GRCh38
rs570604608 Chr.19: 29530076-29530076 on Build GRCh38
rs961461576 Chr.19: 29529945-29529945 on Build GRCh38
rs906911623 Chr.19: 29529757-29529757 on Build GRCh38
rs368179274 Chr.19: 29529750-29529750 on Build GRCh38
rs771486149 Chr.19: 29529855-29529855 on Build GRCh38
rs763122960 Chr.19: 29530088-29530088 on Build GRCh38
rs750971303 Chr.19: 29529892-29529892 on Build GRCh38
rs1049882593 Chr.19: 29530002-29530002 on Build GRCh38
rs1045880572 Chr.19: 29530065-29530065 on Build GRCh38
rs1054139160 Chr.19: 29530034-29530034 on Build GRCh38
rs768417641 Chr.19: 29529812-29529812 on Build GRCh38

View Full Product Details