Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 29942503-29942996 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
URI1
Entrez Gene ID
8725
Gene Name
URI1, prefoldin like chaperone
Gene Aliases
C19orf2, NNX3, PPP1R19, RMP, URI
UniGene
Hs.466391
Gene Chromosome Location
Chr.19: 29923644-30016612 on Build GRCh38
Transcripts
NM_001252641.1 NM_003796.3 NR_045557.1 XM_005259362.2 XM_005259363.4 XM_011527435.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6017421 c.26C>T p.P9L Chr.19: 29942573-29942573 on Build GRCh38
6373758 c.82C>G p.P28A Chr.19: 29942629-29942629 on Build GRCh38
5365306 c.102C>G p.R34R Chr.19: 29942649-29942649 on Build GRCh38
6416318 c.112G>T p.E38* Chr.19: 29942659-29942659 on Build GRCh38
5729575 c.3G>T p.M1I Chr.19: 29942550-29942550 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768976489 Chr.19: 29942637-29942637 on Build GRCh38
rs1023869839 Chr.19: 29942944-29942944 on Build GRCh38
rs531169826 Chr.19: 29942629-29942629 on Build GRCh38
rs780569338 Chr.19: 29942701-29942701 on Build GRCh38
rs767019685 Chr.19: 29942604-29942604 on Build GRCh38
rs1990903 Chr.19: 29942761-29942761 on Build GRCh38
rs777942561 Chr.19: 29942621-29942621 on Build GRCh38
rs12977890 Chr.19: 29942633-29942633 on Build GRCh38
rs773692912 Chr.19: 29942597-29942597 on Build GRCh38
rs780659800 Chr.19: 29942579-29942580 on Build GRCh38
rs891052951 Chr.19: 29942705-29942705 on Build GRCh38
rs969075996 Chr.19: 29942608-29942608 on Build GRCh38
rs1029932076 Chr.19: 29942932-29942932 on Build GRCh38
rs919513121 Chr.19: 29942821-29942821 on Build GRCh38
rs902954609 Chr.19: 29942869-29942869 on Build GRCh38
rs146108454 Chr.19: 29942903-29942903 on Build GRCh38
rs189187 Chr.19: 29942612-29942612 on Build GRCh38
rs776091266 Chr.19: 29942953-29942953 on Build GRCh38
rs929785945 Chr.19: 29942777-29942777 on Build GRCh38
rs962333516 Chr.19: 29942954-29942954 on Build GRCh38
rs1041460784 Chr.19: 29942867-29942867 on Build GRCh38
rs779352501 Chr.19: 29942704-29942704 on Build GRCh38
rs1052962033 Chr.19: 29942907-29942907 on Build GRCh38
rs772777085 Chr.19: 29942652-29942652 on Build GRCh38
rs370296751 Chr.19: 29942613-29942613 on Build GRCh38
rs78911708 Chr.19: 29942970-29942970 on Build GRCh38
rs747281228 Chr.19: 29942548-29942548 on Build GRCh38
rs777855740 Chr.19: 29942748-29942748 on Build GRCh38
rs942167206 Chr.19: 29942576-29942576 on Build GRCh38
rs751262232 Chr.19: 29942669-29942669 on Build GRCh38
rs556712974 Chr.19: 29942797-29942797 on Build GRCh38
rs748654326 Chr.19: 29942707-29942707 on Build GRCh38
rs942607865 Chr.19: 29942802-29942802 on Build GRCh38
rs932313666 Chr.19: 29942544-29942544 on Build GRCh38
rs112296993 Chr.19: 29942536-29942536 on Build GRCh38
rs962655589 Chr.19: 29942949-29942949 on Build GRCh38
rs1012894927 Chr.19: 29942931-29942931 on Build GRCh38
rs773924540 Chr.19: 29942638-29942638 on Build GRCh38
rs993558337 Chr.19: 29942578-29942578 on Build GRCh38
rs867276372 Chr.19: 29942855-29942855 on Build GRCh38
rs528472731 Chr.19: 29942549-29942549 on Build GRCh38
rs375986315 Chr.19: 29942598-29942598 on Build GRCh38
rs757515487 Chr.19: 29942627-29942627 on Build GRCh38
rs765031707 Chr.19: 29942683-29942683 on Build GRCh38
rs369305215 Chr.19: 29942835-29942835 on Build GRCh38
rs1057492614 Chr.19: 29942848-29942848 on Build GRCh38
rs1029404722 Chr.19: 29942756-29942756 on Build GRCh38
rs57838780 Chr.19: 29942860-29942860 on Build GRCh38
rs766185899 Chr.19: 29942667-29942667 on Build GRCh38
rs777824517 Chr.19: 29942731-29942731 on Build GRCh38
rs769372927 Chr.19: 29942601-29942602 on Build GRCh38
rs761282463 Chr.19: 29942641-29942641 on Build GRCh38
rs757348476 Chr.19: 29942685-29942685 on Build GRCh38
rs565573145 Chr.19: 29942717-29942717 on Build GRCh38
rs1039666137 Chr.19: 29942847-29942847 on Build GRCh38
rs763718000 Chr.19: 29942590-29942590 on Build GRCh38
rs748866650 Chr.19: 29942666-29942667 on Build GRCh38
rs974061358 Chr.19: 29942957-29942957 on Build GRCh38
rs781327419 Chr.19: 29942628-29942628 on Build GRCh38
rs772658083 Chr.19: 29942715-29942715 on Build GRCh38
rs746997794 Chr.19: 29942626-29942626 on Build GRCh38
rs750072934 Chr.19: 29942605-29942605 on Build GRCh38
rs961250782 Chr.19: 29942753-29942753 on Build GRCh38
rs918682522 Chr.19: 29942770-29942770 on Build GRCh38
rs536515936 Chr.19: 29942703-29942703 on Build GRCh38
rs752667629 Chr.19: 29942684-29942684 on Build GRCh38
rs770324421 Chr.19: 29942632-29942632 on Build GRCh38
rs552351202 Chr.19: 29942796-29942796 on Build GRCh38
rs777695537 Chr.19: 29942524-29942524 on Build GRCh38
rs565839895 Chr.19: 29942857-29942857 on Build GRCh38
rs763524587 Chr.19: 29942585-29942585 on Build GRCh38
rs756362839 Chr.19: 29942700-29942700 on Build GRCh38
rs572078888 Chr.19: 29942593-29942594 on Build GRCh38
rs1013336536 Chr.19: 29942582-29942582 on Build GRCh38
rs761530942 Chr.19: 29942599-29942599 on Build GRCh38
rs563729123 Chr.19: 29942734-29942734 on Build GRCh38
rs200928000 Chr.19: 29942649-29942649 on Build GRCh38
rs1042735293 Chr.19: 29942770-29942783 on Build GRCh38
rs753819220 Chr.19: 29942614-29942614 on Build GRCh38
rs1035255251 Chr.19: 29942752-29942752 on Build GRCh38
rs770271018 Chr.19: 29942577-29942577 on Build GRCh38
rs117820971 Chr.19: 29942711-29942711 on Build GRCh38
rs12984378 Chr.19: 29942634-29942634 on Build GRCh38
rs951551693 Chr.19: 29942971-29942971 on Build GRCh38
rs192142422 Chr.19: 29942755-29942755 on Build GRCh38
rs779612515 Chr.19: 29942663-29942664 on Build GRCh38
rs777063203 Chr.19: 29942565-29942565 on Build GRCh38
rs902671562 Chr.19: 29942738-29942738 on Build GRCh38
rs922261203 Chr.19: 29942526-29942526 on Build GRCh38
rs746143448 Chr.19: 29942693-29942693 on Build GRCh38
rs978554388 Chr.19: 29942610-29942610 on Build GRCh38
rs372525017 Chr.19: 29942676-29942676 on Build GRCh38
rs922345674 Chr.19: 29942625-29942625 on Build GRCh38
rs976434833 Chr.19: 29942619-29942619 on Build GRCh38
rs545412543 Chr.19: 29942951-29942951 on Build GRCh38
rs760295998 Chr.19: 29942660-29942660 on Build GRCh38
rs1057179595 Chr.19: 29942712-29942712 on Build GRCh38
rs542591670 Chr.19: 29942883-29942883 on Build GRCh38
rs909515761 Chr.19: 29942782-29942783 on Build GRCh38
rs781301927 Chr.19: 29942692-29942692 on Build GRCh38
rs1023588425 Chr.19: 29942758-29942758 on Build GRCh38
rs551935451 Chr.19: 29942581-29942581 on Build GRCh38
rs929458310 Chr.19: 29942842-29942842 on Build GRCh38
rs888806 Chr.19: 29942926-29942926 on Build GRCh38
rs892844041 Chr.19: 29942718-29942718 on Build GRCh38
rs953921694 Chr.19: 29942936-29942936 on Build GRCh38
rs752991171 Chr.19: 29942549-29942563 on Build GRCh38
rs754863175 Chr.19: 29942618-29942618 on Build GRCh38
rs746162017 Chr.19: 29942575-29942575 on Build GRCh38
rs758793515 Chr.19: 29942580-29942588 on Build GRCh38
rs974113829 Chr.19: 29942766-29942766 on Build GRCh38

View Full Product Details